Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 109614172-109614675 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RANBP2
Entrez Gene ID
5903
Gene Name
RAN binding protein 2
Gene Aliases
ADANE, ANE1, IIAE3, NUP358, TRP1, TRP2
UniGene
Hs.199561
Gene Chromosome Location
Chr.2: 108719446-109842301 on Build GRCh38
Transcripts
XM_017004623.1 NM_001321496.1 NM_001321498.1 NM_001321499.1 NM_001321500.1 NM_001321501.1 NM_001321502.1 NM_001321503.1 NM_001321504.1 NM_001321505.1 NM_001321506.1 NM_001321507.1 NM_001321508.1 NM_001321509.1 NM_001321510.1 NM_001321511.1 NM_001321512.1 NM_144710.4 NM_178584.3 XM_006712317.1 XM_011510700.2 XM_011510701.2 XM_011510702.2 XM_011510704.2 NM_023016.3

Gene Symbol
SEPT10
Entrez Gene ID
151011
Gene Name
septin 10
Gene Aliases
-
UniGene
Hs.469615
Gene Chromosome Location
Chr.2: 109542797-109614247 on Build GRCh38
Transcripts
XM_017004623.1 NM_001321496.1 NM_001321498.1 NM_001321499.1 NM_001321500.1 NM_001321501.1 NM_001321502.1 NM_001321503.1 NM_001321504.1 NM_001321505.1 NM_001321506.1 NM_001321507.1 NM_001321508.1 NM_001321509.1 NM_001321510.1 NM_001321511.1 NM_001321512.1 NM_144710.4 NM_178584.3 XM_006712317.1 XM_011510700.2 XM_011510701.2 XM_011510702.2 XM_011510704.2 NM_023016.3

Gene Symbol
SOWAHC
Entrez Gene ID
65124
Gene Name
sosondowah ankyrin repeat domain family member C
Gene Aliases
ANKRD57, C2orf26
UniGene
Hs.355455
Gene Chromosome Location
Chr.2: 109614334-109618987 on Build GRCh38
Transcripts
XM_017004623.1 NM_001321496.1 NM_001321498.1 NM_001321499.1 NM_001321500.1 NM_001321501.1 NM_001321502.1 NM_001321503.1 NM_001321504.1 NM_001321505.1 NM_001321506.1 NM_001321507.1 NM_001321508.1 NM_001321509.1 NM_001321510.1 NM_001321511.1 NM_001321512.1 NM_144710.4 NM_178584.3 XM_006712317.1 XM_011510700.2 XM_011510701.2 XM_011510702.2 XM_011510704.2 NM_023016.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3708811 c.85G>T p.A29S Chr.2: 109614574-109614574 on Build GRCh38
4132984 c.18G>T p.E6D Chr.2: 109614507-109614507 on Build GRCh38
3757577 c.126A>G p.L42L Chr.2: 109614615-109614615 on Build GRCh38
5497452 c.112T>G p.F38V Chr.2: 109614601-109614601 on Build GRCh38
4781487 c.90G>C p.L30L Chr.2: 109614579-109614579 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs781518026 Chr.2: 109614510-109614527 on Build GRCh38
rs756794665 Chr.2: 109614626-109614626 on Build GRCh38
rs1055282737 Chr.2: 109614634-109614639 on Build GRCh38
rs910383929 Chr.2: 109614369-109614369 on Build GRCh38
rs986328159 Chr.2: 109614368-109614368 on Build GRCh38
rs945151049 Chr.2: 109614577-109614577 on Build GRCh38
rs979632101 Chr.2: 109614596-109614596 on Build GRCh38
rs866709706 Chr.2: 109614380-109614380 on Build GRCh38
rs374818848 Chr.2: 109614406-109614406 on Build GRCh38
rs1017556901 Chr.2: 109614530-109614530 on Build GRCh38
rs557527053 Chr.2: 109614571-109614571 on Build GRCh38
rs960902729 Chr.2: 109614317-109614317 on Build GRCh38
rs937704680 Chr.2: 109614625-109614625 on Build GRCh38
rs7603756 Chr.2: 109614275-109614275 on Build GRCh38
rs892124960 Chr.2: 109614273-109614273 on Build GRCh38
rs1033857246 Chr.2: 109614308-109614308 on Build GRCh38
rs114226290 Chr.2: 109614235-109614235 on Build GRCh38
rs920476695 Chr.2: 109614358-109614358 on Build GRCh38
rs775456750 Chr.2: 109614607-109614607 on Build GRCh38
rs961453395 Chr.2: 109614455-109614455 on Build GRCh38
rs776092558 Chr.2: 109614348-109614348 on Build GRCh38
rs748417031 Chr.2: 109614633-109614634 on Build GRCh38
rs533846532 Chr.2: 109614585-109614585 on Build GRCh38
rs528832503 Chr.2: 109614403-109614403 on Build GRCh38
rs751208518 Chr.2: 109614622-109614622 on Build GRCh38
rs1015606541 Chr.2: 109614429-109614429 on Build GRCh38
rs566482097 Chr.2: 109614401-109614401 on Build GRCh38
rs924966782 Chr.2: 109614601-109614601 on Build GRCh38
rs546656675 Chr.2: 109614363-109614363 on Build GRCh38
rs779366267 Chr.2: 109614650-109614650 on Build GRCh38
rs749853971 Chr.2: 109614645-109614645 on Build GRCh38
rs1040186015 Chr.2: 109614248-109614248 on Build GRCh38
rs191122904 Chr.2: 109614321-109614321 on Build GRCh38
rs537679125 Chr.2: 109614468-109614468 on Build GRCh38
rs1037166889 Chr.2: 109614654-109614654 on Build GRCh38
rs920543417 Chr.2: 109614216-109614216 on Build GRCh38
rs767068330 Chr.2: 109614639-109614639 on Build GRCh38
rs944440301 Chr.2: 109614243-109614243 on Build GRCh38
rs961339104 Chr.2: 109614354-109614354 on Build GRCh38
rs548824903 Chr.2: 109614423-109614423 on Build GRCh38
rs913672090 Chr.2: 109614388-109614388 on Build GRCh38
rs573939693 Chr.2: 109614234-109614234 on Build GRCh38
rs571248763 Chr.2: 109614582-109614582 on Build GRCh38
rs545570999 Chr.2: 109614297-109614297 on Build GRCh38
rs945009545 Chr.2: 109614393-109614393 on Build GRCh38
rs553603778 Chr.2: 109614602-109614602 on Build GRCh38
rs965845386 Chr.2: 109614542-109614542 on Build GRCh38
rs765212941 Chr.2: 109614605-109614605 on Build GRCh38
rs1044344882 Chr.2: 109614288-109614288 on Build GRCh38
rs944556451 Chr.2: 109614370-109614370 on Build GRCh38
rs896758930 Chr.2: 109614641-109614641 on Build GRCh38
rs748564799 Chr.2: 109614651-109614651 on Build GRCh38
rs565603707 Chr.2: 109614301-109614301 on Build GRCh38
rs6594048 Chr.2: 109614615-109614615 on Build GRCh38
rs865806655 Chr.2: 109614413-109614413 on Build GRCh38
rs954225570 Chr.2: 109614505-109614505 on Build GRCh38
rs755513800 Chr.2: 109614649-109614649 on Build GRCh38
rs1026988809 Chr.2: 109614475-109614475 on Build GRCh38
rs762810046 Chr.2: 109614614-109614614 on Build GRCh38
rs986970962 Chr.2: 109614291-109614303 on Build GRCh38
rs974117537 Chr.2: 109614357-109614357 on Build GRCh38
rs562973394 Chr.2: 109614241-109614241 on Build GRCh38

View Full Product Details