Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 109615773-109616270 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RANBP2
Entrez Gene ID
5903
Gene Name
RAN binding protein 2
Gene Aliases
ADANE, ANE1, IIAE3, NUP358, TRP1, TRP2
UniGene
Hs.199561
Gene Chromosome Location
Chr.2: 108719446-109842301 on Build GRCh38
Transcripts
XM_017004623.1 NM_023016.3

Gene Symbol
SOWAHC
Entrez Gene ID
65124
Gene Name
sosondowah ankyrin repeat domain family member C
Gene Aliases
ANKRD57, C2orf26
UniGene
Hs.355455
Gene Chromosome Location
Chr.2: 109614334-109618987 on Build GRCh38
Transcripts
XM_017004623.1 NM_023016.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7159195 c.1368A>G p.P456P Chr.2: 109615857-109615857 on Build GRCh38
3673458 c.1496G>T p.G499V Chr.2: 109615985-109615985 on Build GRCh38
5750116 c.1418A>G p.N473S Chr.2: 109615907-109615907 on Build GRCh38
1005338 c.1373G>A p.R458H Chr.2: 109615862-109615862 on Build GRCh38
1005337 c.1284C>T p.L428L Chr.2: 109615773-109615773 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766957308 Chr.2: 109615982-109615982 on Build GRCh38
rs138108589 Chr.2: 109616113-109616113 on Build GRCh38
rs760688907 Chr.2: 109615914-109615914 on Build GRCh38
rs749846926 Chr.2: 109615841-109615861 on Build GRCh38
rs761216764 Chr.2: 109616044-109616044 on Build GRCh38
rs189662109 Chr.2: 109615907-109615907 on Build GRCh38
rs762532284 Chr.2: 109615966-109615967 on Build GRCh38
rs899536425 Chr.2: 109616095-109616095 on Build GRCh38
rs773994069 Chr.2: 109616104-109616104 on Build GRCh38
rs1010774132 Chr.2: 109615999-109615999 on Build GRCh38
rs1005598640 Chr.2: 109616011-109616011 on Build GRCh38
rs953898350 Chr.2: 109616082-109616082 on Build GRCh38
rs746541329 Chr.2: 109615799-109615807 on Build GRCh38
rs757964739 Chr.2: 109615819-109615819 on Build GRCh38
rs769713274 Chr.2: 109616034-109616034 on Build GRCh38
rs368848145 Chr.2: 109615946-109615946 on Build GRCh38
rs763672136 Chr.2: 109615957-109615957 on Build GRCh38
rs572554520 Chr.2: 109615927-109615927 on Build GRCh38
rs766190498 Chr.2: 109615922-109615922 on Build GRCh38
rs752137412 Chr.2: 109615981-109615982 on Build GRCh38
rs774543157 Chr.2: 109615874-109615874 on Build GRCh38
rs545187598 Chr.2: 109616194-109616195 on Build GRCh38
rs1045837066 Chr.2: 109615830-109615830 on Build GRCh38
rs1029947223 Chr.2: 109616158-109616158 on Build GRCh38
rs759468662 Chr.2: 109616022-109616022 on Build GRCh38
rs570033664 Chr.2: 109615861-109615861 on Build GRCh38
rs147455127 Chr.2: 109615888-109615888 on Build GRCh38
rs763614175 Chr.2: 109616041-109616041 on Build GRCh38
rs777944347 Chr.2: 109615990-109615990 on Build GRCh38
rs771698451 Chr.2: 109616119-109616119 on Build GRCh38
rs139093622 Chr.2: 109615823-109615823 on Build GRCh38
rs768112213 Chr.2: 109616008-109616008 on Build GRCh38
rs772917894 Chr.2: 109615998-109615998 on Build GRCh38
rs200515481 Chr.2: 109615811-109615811 on Build GRCh38
rs776927279 Chr.2: 109616139-109616139 on Build GRCh38
rs761395714 Chr.2: 109616107-109616107 on Build GRCh38
rs759378842 Chr.2: 109615937-109615937 on Build GRCh38
rs972975351 Chr.2: 109616171-109616171 on Build GRCh38
rs755359836 Chr.2: 109615983-109615997 on Build GRCh38
rs755334105 Chr.2: 109615978-109615978 on Build GRCh38
rs779274606 Chr.2: 109616106-109616106 on Build GRCh38
rs1033482802 Chr.2: 109616198-109616198 on Build GRCh38
rs981585534 Chr.2: 109616234-109616234 on Build GRCh38
rs553369737 Chr.2: 109615915-109615915 on Build GRCh38
rs146665390 Chr.2: 109615983-109615983 on Build GRCh38
rs991518487 Chr.2: 109615945-109615945 on Build GRCh38
rs747196511 Chr.2: 109615802-109615802 on Build GRCh38
rs376775773 Chr.2: 109615882-109615882 on Build GRCh38
rs139858463 Chr.2: 109615971-109615971 on Build GRCh38
rs147020337 Chr.2: 109616083-109616083 on Build GRCh38
rs749612872 Chr.2: 109615854-109615854 on Build GRCh38
rs752390824 Chr.2: 109615944-109615944 on Build GRCh38
rs142589827 Chr.2: 109615970-109615970 on Build GRCh38
rs747032452 Chr.2: 109615996-109615996 on Build GRCh38
rs143655240 Chr.2: 109616128-109616128 on Build GRCh38
rs766938529 Chr.2: 109616045-109616045 on Build GRCh38
rs762687415 Chr.2: 109616038-109616038 on Build GRCh38
rs752302736 Chr.2: 109615815-109615815 on Build GRCh38
rs780488804 Chr.2: 109615838-109615838 on Build GRCh38
rs201902797 Chr.2: 109616001-109616001 on Build GRCh38
rs781613187 Chr.2: 109615820-109615820 on Build GRCh38
rs76992194 Chr.2: 109615891-109615891 on Build GRCh38
rs4953847 Chr.2: 109616228-109616228 on Build GRCh38
rs765038860 Chr.2: 109615938-109615938 on Build GRCh38
rs997995403 Chr.2: 109616029-109616029 on Build GRCh38
rs564670087 Chr.2: 109616230-109616230 on Build GRCh38
rs745666093 Chr.2: 109615960-109615960 on Build GRCh38
rs751120356 Chr.2: 109616043-109616043 on Build GRCh38
rs141926388 Chr.2: 109615821-109615821 on Build GRCh38
rs902342622 Chr.2: 109616027-109616027 on Build GRCh38
rs764790790 Chr.2: 109615798-109615799 on Build GRCh38
rs918758732 Chr.2: 109615797-109615797 on Build GRCh38
rs750985353 Chr.2: 109615959-109615959 on Build GRCh38
rs200528877 Chr.2: 109615800-109615800 on Build GRCh38
rs1029831500 Chr.2: 109615809-109615809 on Build GRCh38

View Full Product Details