Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 110641564-110642073 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BUB1
Entrez Gene ID
699
Gene Name
BUB1 mitotic checkpoint serine/threonine kinase
Gene Aliases
BUB1A, BUB1L, hBUB1
UniGene
Hs.469649
Gene Chromosome Location
Chr.2: 110637698-110678107 on Build GRCh38
Transcripts
NM_001278616.1 NM_001278617.1 NM_004336.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5892702 c.2575C>T p.Q859* Chr.2: 110641692-110641692 on Build GRCh38
6153722 c.2518A>T p.R840* Chr.2: 110641749-110641749 on Build GRCh38
3565204 c.2467C>T p.Q823* Chr.2: 110641800-110641800 on Build GRCh38
4481735 c.2539C>T p.H847Y Chr.2: 110641728-110641728 on Build GRCh38
3565203 c.2482C>T p.P828S Chr.2: 110641785-110641785 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs929317381 Chr.2: 110641997-110641997 on Build GRCh38
rs368828519 Chr.2: 110641738-110641738 on Build GRCh38
rs189704008 Chr.2: 110641870-110641870 on Build GRCh38
rs769962935 Chr.2: 110641801-110641801 on Build GRCh38
rs763444715 Chr.2: 110641818-110641818 on Build GRCh38
rs965378193 Chr.2: 110641877-110641877 on Build GRCh38
rs369687502 Chr.2: 110641979-110641979 on Build GRCh38
rs145104409 Chr.2: 110641730-110641730 on Build GRCh38
rs976013628 Chr.2: 110641892-110641892 on Build GRCh38
rs772054070 Chr.2: 110641622-110641622 on Build GRCh38
rs905374664 Chr.2: 110641837-110641837 on Build GRCh38
rs940408647 Chr.2: 110641709-110641709 on Build GRCh38
rs753345713 Chr.2: 110641854-110641854 on Build GRCh38
rs141293590 Chr.2: 110641786-110641786 on Build GRCh38
rs1020018307 Chr.2: 110641876-110641876 on Build GRCh38
rs745799765 Chr.2: 110641618-110641618 on Build GRCh38
rs199546885 Chr.2: 110641839-110641839 on Build GRCh38
rs776930549 Chr.2: 110641667-110641667 on Build GRCh38
rs200251349 Chr.2: 110641784-110641784 on Build GRCh38
rs376562921 Chr.2: 110641890-110641892 on Build GRCh38
rs941252699 Chr.2: 110642031-110642031 on Build GRCh38
rs777879454 Chr.2: 110641972-110641972 on Build GRCh38
rs879858517 Chr.2: 110641673-110641673 on Build GRCh38
rs185227295 Chr.2: 110641856-110641856 on Build GRCh38
rs778754030 Chr.2: 110641763-110641763 on Build GRCh38
rs950129788 Chr.2: 110641600-110641600 on Build GRCh38
rs984541230 Chr.2: 110641620-110641620 on Build GRCh38
rs752793469 Chr.2: 110641867-110641867 on Build GRCh38
rs55731145 Chr.2: 110641789-110641789 on Build GRCh38
rs888375715 Chr.2: 110642025-110642025 on Build GRCh38
rs577822032 Chr.2: 110642016-110642016 on Build GRCh38
rs773509073 Chr.2: 110641702-110641702 on Build GRCh38
rs200481903 Chr.2: 110641769-110641769 on Build GRCh38
rs758297724 Chr.2: 110641774-110641774 on Build GRCh38
rs984620829 Chr.2: 110642040-110642040 on Build GRCh38
rs568383960 Chr.2: 110641890-110641890 on Build GRCh38
rs888341586 Chr.2: 110641733-110641733 on Build GRCh38
rs758512869 Chr.2: 110641896-110641896 on Build GRCh38
rs773682232 Chr.2: 110641803-110641803 on Build GRCh38
rs747129448 Chr.2: 110641640-110641640 on Build GRCh38
rs908954703 Chr.2: 110641660-110641660 on Build GRCh38
rs767034942 Chr.2: 110641725-110641725 on Build GRCh38
rs74442210 Chr.2: 110641875-110641875 on Build GRCh38
rs1033679397 Chr.2: 110641869-110641869 on Build GRCh38
rs375503157 Chr.2: 110641601-110641601 on Build GRCh38
rs763595804 Chr.2: 110641746-110641746 on Build GRCh38
rs1039598735 Chr.2: 110642035-110642035 on Build GRCh38
rs780083216 Chr.2: 110641626-110641626 on Build GRCh38
rs142192868 Chr.2: 110641762-110641762 on Build GRCh38
rs950159496 Chr.2: 110641909-110641909 on Build GRCh38
rs753532592 Chr.2: 110641758-110641758 on Build GRCh38
rs778648975 Chr.2: 110641591-110641591 on Build GRCh38
rs1049016929 Chr.2: 110642010-110642010 on Build GRCh38
rs762020192 Chr.2: 110641678-110641678 on Build GRCh38
rs375547743 Chr.2: 110641840-110641840 on Build GRCh38
rs563351644 Chr.2: 110641792-110641792 on Build GRCh38
rs1028875497 Chr.2: 110641930-110641930 on Build GRCh38
rs770194928 Chr.2: 110641692-110641692 on Build GRCh38
rs72942112 Chr.2: 110641938-110641938 on Build GRCh38
rs748392521 Chr.2: 110641794-110641794 on Build GRCh38
rs919320174 Chr.2: 110641973-110641973 on Build GRCh38
rs763534753 Chr.2: 110641706-110641706 on Build GRCh38
rs775012737 Chr.2: 110641835-110641835 on Build GRCh38
rs570878972 Chr.2: 110642042-110642042 on Build GRCh38
rs771516020 Chr.2: 110641820-110641820 on Build GRCh38
rs768716644 Chr.2: 110641642-110641642 on Build GRCh38

View Full Product Details