Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 119604787-119605282 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CFAP221
Entrez Gene ID
200373
Gene Name
cilia and flagella associated protein 221
Gene Aliases
FAP221, PCDP1
UniGene
Hs.496753
Gene Chromosome Location
Chr.2: 119544392-119688610 on Build GRCh38
Transcripts
NM_001271049.1 NR_073132.1 NR_073133.1 XM_006712353.3 XM_017003550.1 XM_017003551.1 XM_017003552.1 XM_017003553.1 XM_017003554.1 XM_017003555.1 XM_017003556.1 XM_017003557.1 XM_017003558.1 XM_017003559.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1399065 c.219C>A p.L73L Chr.2: 119605233-119605233 on Build GRCh38
1291287 c.54G>A p.K18K Chr.2: 119604792-119604792 on Build GRCh38
4854040 c.250G>T p.E84* Chr.2: 119605264-119605264 on Build GRCh38
3894637 c.137G>A p.G46E Chr.2: 119604958-119604958 on Build GRCh38
441053 c.219C>G p.L73L Chr.2: 119605233-119605233 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762827089 Chr.2: 119605250-119605250 on Build GRCh38
rs768916817 Chr.2: 119604837-119604837 on Build GRCh38
rs920818332 Chr.2: 119605140-119605140 on Build GRCh38
rs780260965 Chr.2: 119605094-119605094 on Build GRCh38
rs768013621 Chr.2: 119605012-119605012 on Build GRCh38
rs536158307 Chr.2: 119605233-119605233 on Build GRCh38
rs369018788 Chr.2: 119605161-119605161 on Build GRCh38
rs373961421 Chr.2: 119605255-119605255 on Build GRCh38
rs374566349 Chr.2: 119604821-119604821 on Build GRCh38
rs148375672 Chr.2: 119604916-119604916 on Build GRCh38
rs774914393 Chr.2: 119605000-119605000 on Build GRCh38
rs199910398 Chr.2: 119604832-119604833 on Build GRCh38
rs955723377 Chr.2: 119605152-119605152 on Build GRCh38
rs148985492 Chr.2: 119605238-119605238 on Build GRCh38
rs766045453 Chr.2: 119605251-119605251 on Build GRCh38
rs762155520 Chr.2: 119604872-119604873 on Build GRCh38
rs868605053 Chr.2: 119604987-119604987 on Build GRCh38
rs970054266 Chr.2: 119605144-119605144 on Build GRCh38
rs780063197 Chr.2: 119604929-119604929 on Build GRCh38
rs755113026 Chr.2: 119605188-119605188 on Build GRCh38
rs752977384 Chr.2: 119605194-119605194 on Build GRCh38
rs763866162 Chr.2: 119604813-119604813 on Build GRCh38
rs764044968 Chr.2: 119604961-119604961 on Build GRCh38
rs748498705 Chr.2: 119604862-119604862 on Build GRCh38
rs182462823 Chr.2: 119605101-119605101 on Build GRCh38
rs778221051 Chr.2: 119604978-119604978 on Build GRCh38
rs765074385 Chr.2: 119605142-119605142 on Build GRCh38
rs911514076 Chr.2: 119604899-119604899 on Build GRCh38
rs760287563 Chr.2: 119604889-119604889 on Build GRCh38
rs771543800 Chr.2: 119604876-119604876 on Build GRCh38
rs778325278 Chr.2: 119605197-119605197 on Build GRCh38
rs140168134 Chr.2: 119604942-119604942 on Build GRCh38
rs1014983228 Chr.2: 119605100-119605100 on Build GRCh38
rs757302475 Chr.2: 119605063-119605063 on Build GRCh38
rs1008931258 Chr.2: 119605071-119605071 on Build GRCh38
rs376003657 Chr.2: 119605217-119605217 on Build GRCh38
rs1014127494 Chr.2: 119604984-119604984 on Build GRCh38
rs955564274 Chr.2: 119604856-119604856 on Build GRCh38
rs776271818 Chr.2: 119605021-119605021 on Build GRCh38
rs372652619 Chr.2: 119604818-119604818 on Build GRCh38
rs754902967 Chr.2: 119604815-119604815 on Build GRCh38
rs775064928 Chr.2: 119604885-119604885 on Build GRCh38
rs61738913 Chr.2: 119604917-119604917 on Build GRCh38
rs141477875 Chr.2: 119604958-119604958 on Build GRCh38
rs1009923485 Chr.2: 119605127-119605127 on Build GRCh38
rs771441829 Chr.2: 119604995-119604995 on Build GRCh38
rs904455754 Chr.2: 119604988-119604988 on Build GRCh38
rs757236664 Chr.2: 119604817-119604817 on Build GRCh38
rs772986208 Chr.2: 119605147-119605147 on Build GRCh38
rs776107118 Chr.2: 119605236-119605236 on Build GRCh38
rs767142120 Chr.2: 119604847-119604847 on Build GRCh38
rs1568540 Chr.2: 119604831-119604831 on Build GRCh38
rs773882710 Chr.2: 119604870-119604870 on Build GRCh38
rs141590203 Chr.2: 119605038-119605038 on Build GRCh38
rs985655812 Chr.2: 119605160-119605160 on Build GRCh38
rs753441574 Chr.2: 119604912-119604912 on Build GRCh38
rs758342076 Chr.2: 119604925-119604925 on Build GRCh38
rs200710868 Chr.2: 119604869-119604869 on Build GRCh38
rs759330690 Chr.2: 119605256-119605256 on Build GRCh38
rs920860066 Chr.2: 119604829-119604829 on Build GRCh38
rs746553415 Chr.2: 119605002-119605002 on Build GRCh38
rs772927999 Chr.2: 119605245-119605245 on Build GRCh38
rs779507937 Chr.2: 119605231-119605231 on Build GRCh38
rs770283561 Chr.2: 119604972-119604972 on Build GRCh38
rs370358555 Chr.2: 119605230-119605230 on Build GRCh38
rs751401128 Chr.2: 119605165-119605165 on Build GRCh38
rs763483467 Chr.2: 119604871-119604871 on Build GRCh38
rs1056559397 Chr.2: 119605241-119605241 on Build GRCh38
rs749830944 Chr.2: 119604994-119604994 on Build GRCh38
rs763782582 Chr.2: 119604911-119604911 on Build GRCh38
rs550127561 Chr.2: 119605126-119605126 on Build GRCh38
rs751625859 Chr.2: 119604934-119604934 on Build GRCh38
rs781448027 Chr.2: 119604859-119604859 on Build GRCh38
rs372472009 Chr.2: 119605163-119605163 on Build GRCh38
rs974449771 Chr.2: 119605221-119605221 on Build GRCh38
rs752923328 Chr.2: 119605003-119605003 on Build GRCh38
rs905788046 Chr.2: 119605057-119605057 on Build GRCh38
rs267598852 Chr.2: 119605185-119605185 on Build GRCh38
rs200170187 Chr.2: 119605211-119605211 on Build GRCh38
rs780124628 Chr.2: 119604827-119604827 on Build GRCh38
rs767634123 Chr.2: 119605190-119605190 on Build GRCh38
rs750321404 Chr.2: 119604820-119604820 on Build GRCh38
rs962210446 Chr.2: 119605120-119605120 on Build GRCh38
rs926997216 Chr.2: 119604887-119604887 on Build GRCh38
rs769404674 Chr.2: 119605239-119605239 on Build GRCh38

View Full Product Details