Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 18078157-18078650 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PEX26
Entrez Gene ID
55670
Gene Name
peroxisomal biogenesis factor 26
Gene Aliases
PBD7A, PBD7B, PEX26M1T, Pex26pM1T
UniGene
Hs.517400
Gene Chromosome Location
Chr.22: 18077920-18091031 on Build GRCh38
Transcripts
NM_001127649.2 NM_001199319.1 NM_017929.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6385343 c.76C>A p.R26S Chr.22: 18078452-18078452 on Build GRCh38
1307939 c.230+1G>C p.? Chr.22: 18078607-18078607 on Build GRCh38
6378408 c.224C>T p.A75V Chr.22: 18078600-18078600 on Build GRCh38
3708127 c.179G>A p.R60Q Chr.22: 18078555-18078555 on Build GRCh38
7316149 c.118G>T p.E40* Chr.22: 18078494-18078494 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs771847925 Chr.22: 18078447-18078447 on Build GRCh38
rs778782682 Chr.22: 18078512-18078512 on Build GRCh38
rs766581207 Chr.22: 18078584-18078584 on Build GRCh38
rs61752130 Chr.22: 18078413-18078414 on Build GRCh38
rs267608190 Chr.22: 18078607-18078607 on Build GRCh38
rs767551808 Chr.22: 18078492-18078492 on Build GRCh38
rs200178985 Chr.22: 18078442-18078442 on Build GRCh38
rs747273337 Chr.22: 18078370-18078370 on Build GRCh38
rs886057245 Chr.22: 18078614-18078614 on Build GRCh38
rs765484850 Chr.22: 18078412-18078412 on Build GRCh38
rs150895887 Chr.22: 18078506-18078506 on Build GRCh38
rs747165381 Chr.22: 18078247-18078247 on Build GRCh38
rs1006819708 Chr.22: 18078253-18078253 on Build GRCh38
rs558287136 Chr.22: 18078332-18078332 on Build GRCh38
rs751509006 Chr.22: 18078333-18078336 on Build GRCh38
rs914485463 Chr.22: 18078349-18078349 on Build GRCh38
rs61752131 Chr.22: 18078507-18078507 on Build GRCh38
rs1015977997 Chr.22: 18078568-18078568 on Build GRCh38
rs762891761 Chr.22: 18078265-18078265 on Build GRCh38
rs1051110181 Chr.22: 18078419-18078419 on Build GRCh38
rs910106149 Chr.22: 18078180-18078180 on Build GRCh38
rs768425084 Chr.22: 18078405-18078405 on Build GRCh38
rs754809897 Chr.22: 18078431-18078431 on Build GRCh38
rs766917354 Chr.22: 18078322-18078322 on Build GRCh38
rs786205556 Chr.22: 18078604-18078604 on Build GRCh38
rs372192286 Chr.22: 18078581-18078581 on Build GRCh38
rs748194244 Chr.22: 18078238-18078238 on Build GRCh38
rs552957173 Chr.22: 18078264-18078264 on Build GRCh38
rs769260247 Chr.22: 18078545-18078545 on Build GRCh38
rs373434404 Chr.22: 18078327-18078327 on Build GRCh38
rs748386776 Chr.22: 18078343-18078343 on Build GRCh38
rs770555305 Chr.22: 18078453-18078453 on Build GRCh38
rs374290931 Chr.22: 18078574-18078574 on Build GRCh38
rs758213085 Chr.22: 18078528-18078528 on Build GRCh38
rs757375227 Chr.22: 18078479-18078479 on Build GRCh38
rs754201816 Chr.22: 18078341-18078341 on Build GRCh38
rs61752129 Chr.22: 18078410-18078411 on Build GRCh38
rs747831264 Chr.22: 18078518-18078518 on Build GRCh38
rs988740478 Chr.22: 18078336-18078336 on Build GRCh38
rs777633990 Chr.22: 18078529-18078529 on Build GRCh38
rs992575746 Chr.22: 18078398-18078398 on Build GRCh38
rs746066587 Chr.22: 18078430-18078430 on Build GRCh38
rs752047094 Chr.22: 18078476-18078476 on Build GRCh38
rs780849690 Chr.22: 18078626-18078626 on Build GRCh38
rs764244307 Chr.22: 18078424-18078424 on Build GRCh38
rs751562812 Chr.22: 18078603-18078603 on Build GRCh38
rs775203834 Chr.22: 18078551-18078551 on Build GRCh38
rs779148634 Chr.22: 18078189-18078189 on Build GRCh38
rs774393110 Chr.22: 18078406-18078406 on Build GRCh38
rs758542848 Chr.22: 18078357-18078357 on Build GRCh38
rs267608189 Chr.22: 18078568-18078592 on Build GRCh38
rs984532105 Chr.22: 18078385-18078385 on Build GRCh38
rs1054584769 Chr.22: 18078433-18078433 on Build GRCh38
rs758179855 Chr.22: 18078593-18078593 on Build GRCh38
rs535852316 Chr.22: 18078245-18078245 on Build GRCh38
rs769604096 Chr.22: 18078250-18078250 on Build GRCh38
rs763285100 Chr.22: 18078553-18078553 on Build GRCh38
rs764495911 Chr.22: 18078555-18078555 on Build GRCh38
rs749146718 Chr.22: 18078399-18078399 on Build GRCh38
rs760205716 Chr.22: 18078331-18078331 on Build GRCh38
rs76345477 Chr.22: 18078270-18078270 on Build GRCh38
rs770013891 Chr.22: 18078393-18078393 on Build GRCh38
rs779828551 Chr.22: 18078394-18078394 on Build GRCh38
rs867918324 Chr.22: 18078281-18078281 on Build GRCh38
rs903287940 Chr.22: 18078256-18078256 on Build GRCh38
rs777671183 Chr.22: 18078358-18078358 on Build GRCh38
rs774876537 Chr.22: 18078563-18078563 on Build GRCh38
rs1010208134 Chr.22: 18078496-18078496 on Build GRCh38
rs757324021 Chr.22: 18078426-18078426 on Build GRCh38
rs963166801 Chr.22: 18078571-18078571 on Build GRCh38
rs1002922692 Chr.22: 18078481-18078481 on Build GRCh38
rs745673881 Chr.22: 18078554-18078554 on Build GRCh38
rs201884779 Chr.22: 18078576-18078576 on Build GRCh38
rs754081996 Chr.22: 18078590-18078590 on Build GRCh38
rs761627505 Chr.22: 18078407-18078407 on Build GRCh38
rs1025994616 Chr.22: 18078345-18078345 on Build GRCh38
rs771079248 Chr.22: 18078375-18078375 on Build GRCh38
rs924635830 Chr.22: 18078428-18078428 on Build GRCh38
rs774150589 Chr.22: 18078276-18078276 on Build GRCh38
rs956044242 Chr.22: 18078315-18078315 on Build GRCh38
rs965187527 Chr.22: 18078279-18078279 on Build GRCh38
rs781474596 Chr.22: 18078292-18078292 on Build GRCh38
rs368026843 Chr.22: 18078182-18078182 on Build GRCh38
rs374630564 Chr.22: 18078455-18078455 on Build GRCh38
rs779022472 Chr.22: 18078209-18078212 on Build GRCh38
rs781449474 Chr.22: 18078427-18078427 on Build GRCh38
rs772042346 Chr.22: 18078243-18078243 on Build GRCh38
rs199668827 Chr.22: 18078409-18078409 on Build GRCh38
rs778534478 Chr.22: 18078441-18078441 on Build GRCh38
rs547969346 Chr.22: 18078460-18078460 on Build GRCh38
rs918352794 Chr.22: 18078616-18078616 on Build GRCh38
rs746924588 Chr.22: 18078531-18078531 on Build GRCh38
rs745525570 Chr.22: 18078544-18078544 on Build GRCh38
rs766873426 Chr.22: 18078338-18078338 on Build GRCh38
rs981132384 Chr.22: 18078339-18078339 on Build GRCh38
rs1051578101 Chr.22: 18078255-18078255 on Build GRCh38
rs755550753 Chr.22: 18078342-18078342 on Build GRCh38
rs753275056 Chr.22: 18078415-18078415 on Build GRCh38
rs927136983 Chr.22: 18078362-18078362 on Build GRCh38
rs746587582 Chr.22: 18078451-18078451 on Build GRCh38
rs200279475 Chr.22: 18078408-18078408 on Build GRCh38
rs375516973 Chr.22: 18078583-18078583 on Build GRCh38
rs780983228 Chr.22: 18078509-18078511 on Build GRCh38
rs61752132 Chr.22: 18078510-18078510 on Build GRCh38
rs74315506 Chr.22: 18078378-18078378 on Build GRCh38
rs775656539 Chr.22: 18078467-18078467 on Build GRCh38
rs866064101 Chr.22: 18078178-18078178 on Build GRCh38
rs777682317 Chr.22: 18078594-18078594 on Build GRCh38
rs899826237 Chr.22: 18078191-18078191 on Build GRCh38
rs758778915 Chr.22: 18078418-18078418 on Build GRCh38
rs776229764 Chr.22: 18078539-18078539 on Build GRCh38
rs765654181 Chr.22: 18078465-18078465 on Build GRCh38
rs757200331 Chr.22: 18078609-18078609 on Build GRCh38
rs892997707 Chr.22: 18078282-18078282 on Build GRCh38
rs759221571 Chr.22: 18078463-18078463 on Build GRCh38
rs753539295 Chr.22: 18078410-18078410 on Build GRCh38
rs968643646 Chr.22: 18078355-18078355 on Build GRCh38
rs910687564 Chr.22: 18078413-18078413 on Build GRCh38
rs753133378 Chr.22: 18078353-18078353 on Build GRCh38
rs368118099 Chr.22: 18078474-18078474 on Build GRCh38
rs751527539 Chr.22: 18078425-18078425 on Build GRCh38

View Full Product Details