Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 19454670-19455180 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
UFD1L
Entrez Gene ID
7353
Gene Name
ubiquitin fusion degradation 1 like (yeast)
Gene Aliases
UFD1
UniGene
Hs.474213
Gene Chromosome Location
Chr.22: 19449941-19479215 on Build GRCh38
Transcripts
NM_001035247.2 NM_005659.6 XM_011530361.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4848046 c.813C>T p.I271I Chr.22: 19454785-19454785 on Build GRCh38
7190051 c.828_829insC p.L277fs*6 Chr.22: 19454769-19454770 on Build GRCh38
5675737 c.838A>G p.K280E Chr.22: 19454760-19454760 on Build GRCh38
6468041 c.815G>A p.R272K Chr.22: 19454783-19454783 on Build GRCh38
4102582 c.782A>G p.Y261C Chr.22: 19454816-19454816 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs752708647 Chr.22: 19454730-19454730 on Build GRCh38
rs994423721 Chr.22: 19455085-19455085 on Build GRCh38
rs777533484 Chr.22: 19454704-19454704 on Build GRCh38
rs745388140 Chr.22: 19454726-19454726 on Build GRCh38
rs910208142 Chr.22: 19454896-19454897 on Build GRCh38
rs755952623 Chr.22: 19454910-19454910 on Build GRCh38
rs184179976 Chr.22: 19455055-19455055 on Build GRCh38
rs1057386166 Chr.22: 19454926-19454926 on Build GRCh38
rs776751114 Chr.22: 19454850-19454851 on Build GRCh38
rs780807680 Chr.22: 19454718-19454718 on Build GRCh38
rs966861162 Chr.22: 19454839-19454839 on Build GRCh38
rs751584851 Chr.22: 19454707-19454707 on Build GRCh38
rs985719414 Chr.22: 19455136-19455136 on Build GRCh38
rs912724252 Chr.22: 19454729-19454729 on Build GRCh38
rs1030500060 Chr.22: 19454847-19454847 on Build GRCh38
rs368965112 Chr.22: 19454722-19454731 on Build GRCh38
rs762421866 Chr.22: 19454774-19454774 on Build GRCh38
rs760116554 Chr.22: 19455054-19455054 on Build GRCh38
rs772436238 Chr.22: 19454808-19454808 on Build GRCh38
rs575096282 Chr.22: 19454933-19454933 on Build GRCh38
rs1001450548 Chr.22: 19454923-19454923 on Build GRCh38
rs758761200 Chr.22: 19454707-19454709 on Build GRCh38
rs773729986 Chr.22: 19454815-19454815 on Build GRCh38
rs546150653 Chr.22: 19454913-19454913 on Build GRCh38
rs745909546 Chr.22: 19454871-19454871 on Build GRCh38
rs928056966 Chr.22: 19454927-19454927 on Build GRCh38
rs138869651 Chr.22: 19454854-19454854 on Build GRCh38
rs748889125 Chr.22: 19454757-19454757 on Build GRCh38
rs758205273 Chr.22: 19454701-19454701 on Build GRCh38
rs572830693 Chr.22: 19454859-19454859 on Build GRCh38
rs1029803994 Chr.22: 19455144-19455144 on Build GRCh38
rs763920053 Chr.22: 19454865-19454865 on Build GRCh38
rs771225321 Chr.22: 19454845-19454846 on Build GRCh38
rs563224418 Chr.22: 19454920-19454920 on Build GRCh38
rs73877067 Chr.22: 19455106-19455106 on Build GRCh38
rs769199616 Chr.22: 19454763-19454763 on Build GRCh38
rs954477802 Chr.22: 19455118-19455118 on Build GRCh38
rs1004608741 Chr.22: 19454993-19454993 on Build GRCh38
rs1053169950 Chr.22: 19454765-19454765 on Build GRCh38
rs929147653 Chr.22: 19454919-19454919 on Build GRCh38
rs377614768 Chr.22: 19454717-19454717 on Build GRCh38
rs747177448 Chr.22: 19454839-19454844 on Build GRCh38
rs751292936 Chr.22: 19454868-19454868 on Build GRCh38
rs1804674 Chr.22: 19454759-19454759 on Build GRCh38
rs757024403 Chr.22: 19454878-19454878 on Build GRCh38
rs895633555 Chr.22: 19454944-19454944 on Build GRCh38
rs753624901 Chr.22: 19455026-19455026 on Build GRCh38
rs998376248 Chr.22: 19455129-19455129 on Build GRCh38
rs531795752 Chr.22: 19455074-19455074 on Build GRCh38
rs912748013 Chr.22: 19455141-19455141 on Build GRCh38
rs567211282 Chr.22: 19454766-19454766 on Build GRCh38
rs536261331 Chr.22: 19454844-19454844 on Build GRCh38
rs199863554 Chr.22: 19454793-19454793 on Build GRCh38
rs1021173785 Chr.22: 19454746-19454746 on Build GRCh38
rs543883905 Chr.22: 19455047-19455085 on Build GRCh38
rs113116276 Chr.22: 19454911-19454911 on Build GRCh38
rs200084725 Chr.22: 19454738-19454738 on Build GRCh38
rs760675798 Chr.22: 19454833-19454833 on Build GRCh38

View Full Product Details