Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 20889960-20890460 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SNAP29
Entrez Gene ID
9342
Gene Name
synaptosome associated protein 29
Gene Aliases
CEDNIK, SNAP-29
UniGene
Hs.108002
Gene Chromosome Location
Chr.22: 20859004-20891214 on Build GRCh38
Transcripts
NM_004782.3

SNP Detail

SNP ID Chromosome Location
rs1040963696 Chr.22: 20890139-20890139 on Build GRCh38
rs984894233 Chr.22: 20889999-20889999 on Build GRCh38
rs978822617 Chr.22: 20890085-20890085 on Build GRCh38
rs139989203 Chr.22: 20890247-20890247 on Build GRCh38
rs536474432 Chr.22: 20890001-20890001 on Build GRCh38
rs528779987 Chr.22: 20890248-20890248 on Build GRCh38
rs917269858 Chr.22: 20890353-20890353 on Build GRCh38
rs965629445 Chr.22: 20890037-20890037 on Build GRCh38
rs926683465 Chr.22: 20890067-20890067 on Build GRCh38
rs756798079 Chr.22: 20890207-20890207 on Build GRCh38
rs146027253 Chr.22: 20890041-20890041 on Build GRCh38
rs1045115539 Chr.22: 20890060-20890060 on Build GRCh38
rs755372395 Chr.22: 20890036-20890036 on Build GRCh38
rs1020375078 Chr.22: 20890266-20890266 on Build GRCh38
rs543535379 Chr.22: 20890116-20890116 on Build GRCh38
rs564712017 Chr.22: 20890304-20890304 on Build GRCh38
rs563469455 Chr.22: 20890142-20890142 on Build GRCh38
rs901120462 Chr.22: 20890141-20890141 on Build GRCh38
rs888104939 Chr.22: 20890022-20890022 on Build GRCh38
rs918434313 Chr.22: 20890178-20890178 on Build GRCh38
rs73162856 Chr.22: 20890387-20890387 on Build GRCh38
rs112984030 Chr.22: 20890337-20890337 on Build GRCh38
rs1053884107 Chr.22: 20890421-20890422 on Build GRCh38
rs918200774 Chr.22: 20890032-20890032 on Build GRCh38
rs901410064 Chr.22: 20890250-20890250 on Build GRCh38
rs866349039 Chr.22: 20890010-20890010 on Build GRCh38
rs1020428754 Chr.22: 20890046-20890046 on Build GRCh38
rs918159439 Chr.22: 20890291-20890291 on Build GRCh38
rs191563769 Chr.22: 20890434-20890434 on Build GRCh38
rs967511223 Chr.22: 20890271-20890271 on Build GRCh38
rs532316531 Chr.22: 20890317-20890317 on Build GRCh38
rs165744 Chr.22: 20890097-20890097 on Build GRCh38
rs534968594 Chr.22: 20890096-20890096 on Build GRCh38
rs1016809067 Chr.22: 20890251-20890251 on Build GRCh38
rs1000408198 Chr.22: 20890283-20890283 on Build GRCh38
rs886057274 Chr.22: 20890140-20890140 on Build GRCh38
rs186251828 Chr.22: 20890243-20890243 on Build GRCh38
rs936701336 Chr.22: 20890419-20890419 on Build GRCh38
rs115923524 Chr.22: 20890113-20890113 on Build GRCh38
rs972682815 Chr.22: 20890282-20890282 on Build GRCh38
rs958845967 Chr.22: 20890330-20890330 on Build GRCh38
rs573867053 Chr.22: 20890151-20890151 on Build GRCh38
rs970999413 Chr.22: 20890292-20890292 on Build GRCh38
rs949293280 Chr.22: 20890122-20890122 on Build GRCh38
rs1006598562 Chr.22: 20890026-20890026 on Build GRCh38
rs886057273 Chr.22: 20890004-20890004 on Build GRCh38
rs529877604 Chr.22: 20890371-20890371 on Build GRCh38
rs1046237598 Chr.22: 20890131-20890131 on Build GRCh38
rs567385933 Chr.22: 20890358-20890358 on Build GRCh38
rs998395400 Chr.22: 20890254-20890254 on Build GRCh38
rs553271593 Chr.22: 20890015-20890015 on Build GRCh38
rs775605635 Chr.22: 20890105-20890105 on Build GRCh38
rs367880062 Chr.22: 20890320-20890320 on Build GRCh38
rs967396570 Chr.22: 20890074-20890074 on Build GRCh38
rs886057275 Chr.22: 20890400-20890401 on Build GRCh38
rs566704425 Chr.22: 20890424-20890424 on Build GRCh38
rs1027957112 Chr.22: 20890225-20890225 on Build GRCh38
rs929831198 Chr.22: 20890202-20890202 on Build GRCh38
rs760927522 Chr.22: 20890224-20890224 on Build GRCh38
rs972349238 Chr.22: 20890022-20890026 on Build GRCh38
rs552092485 Chr.22: 20890280-20890280 on Build GRCh38

View Full Product Details