Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 49999003-49999516 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IL17REL
Entrez Gene ID
400935
Gene Name
interleukin 17 receptor E like
Gene Aliases
-
UniGene
Hs.526712
Gene Chromosome Location
Chr.22: 49993578-50013827 on Build GRCh38
Transcripts
NM_001001694.2 XM_011530689.2 XM_017028798.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6685032 c.574C>T p.R192W Chr.22: 49999318-49999318 on Build GRCh38
1566406 c.570G>A p.A190A Chr.22: 49999322-49999322 on Build GRCh38
6533317 c.554C>G p.S185C Chr.22: 49999338-49999338 on Build GRCh38
363526 c.595G>A p.E199K Chr.22: 49999297-49999297 on Build GRCh38
7290482 c.583A>T p.I195F Chr.22: 49999309-49999309 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs773763375 Chr.22: 49999270-49999270 on Build GRCh38
rs879016393 Chr.22: 49999364-49999364 on Build GRCh38
rs375620326 Chr.22: 49999380-49999380 on Build GRCh38
rs745975001 Chr.22: 49999353-49999353 on Build GRCh38
rs746487946 Chr.22: 49999254-49999254 on Build GRCh38
rs978886012 Chr.22: 49999391-49999391 on Build GRCh38
rs890077318 Chr.22: 49999165-49999165 on Build GRCh38
rs771398857 Chr.22: 49999277-49999277 on Build GRCh38
rs778002617 Chr.22: 49999330-49999330 on Build GRCh38
rs756978055 Chr.22: 49999337-49999338 on Build GRCh38
rs747039026 Chr.22: 49999415-49999415 on Build GRCh38
rs775910823 Chr.22: 49999357-49999357 on Build GRCh38
rs549525300 Chr.22: 49999316-49999316 on Build GRCh38
rs143798666 Chr.22: 49999318-49999318 on Build GRCh38
rs775743205 Chr.22: 49999413-49999413 on Build GRCh38
rs767198914 Chr.22: 49999344-49999344 on Build GRCh38
rs144306366 Chr.22: 49999098-49999099 on Build GRCh38
rs754544291 Chr.22: 49999382-49999382 on Build GRCh38
rs760984247 Chr.22: 49999362-49999362 on Build GRCh38
rs749963630 Chr.22: 49999348-49999349 on Build GRCh38
rs752132627 Chr.22: 49999389-49999389 on Build GRCh38
rs780804044 Chr.22: 49999257-49999257 on Build GRCh38
rs779436951 Chr.22: 49999251-49999251 on Build GRCh38
rs886841030 Chr.22: 49999193-49999193 on Build GRCh38
rs752224497 Chr.22: 49999452-49999452 on Build GRCh38
rs374689248 Chr.22: 49999268-49999268 on Build GRCh38
rs200581652 Chr.22: 49999282-49999282 on Build GRCh38
rs762726616 Chr.22: 49999432-49999432 on Build GRCh38
rs557425200 Chr.22: 49999063-49999063 on Build GRCh38
rs776885429 Chr.22: 49999429-49999429 on Build GRCh38
rs763380481 Chr.22: 49999273-49999273 on Build GRCh38
rs533412083 Chr.22: 49999212-49999212 on Build GRCh38
rs13056254 Chr.22: 49999232-49999232 on Build GRCh38
rs369338278 Chr.22: 49999135-49999135 on Build GRCh38
rs934030903 Chr.22: 49999117-49999117 on Build GRCh38
rs933810967 Chr.22: 49999164-49999164 on Build GRCh38
rs770750992 Chr.22: 49999437-49999437 on Build GRCh38
rs911423631 Chr.22: 49999394-49999394 on Build GRCh38
rs757619950 Chr.22: 49999482-49999482 on Build GRCh38
rs1050893708 Chr.22: 49999175-49999175 on Build GRCh38
rs557149104 Chr.22: 49999345-49999345 on Build GRCh38
rs148168797 Chr.22: 49999334-49999334 on Build GRCh38
rs371867167 Chr.22: 49999459-49999459 on Build GRCh38
rs925917765 Chr.22: 49999131-49999131 on Build GRCh38
rs758997309 Chr.22: 49999491-49999491 on Build GRCh38
rs749834372 Chr.22: 49999442-49999446 on Build GRCh38
rs1031778986 Chr.22: 49999397-49999397 on Build GRCh38
rs772904947 Chr.22: 49999374-49999374 on Build GRCh38
rs760152952 Chr.22: 49999458-49999458 on Build GRCh38
rs759470319 Chr.22: 49999447-49999447 on Build GRCh38
rs779354978 Chr.22: 49999403-49999403 on Build GRCh38
rs923956789 Chr.22: 49999107-49999107 on Build GRCh38
rs386822623 Chr.22: 49999096-49999098 on Build GRCh38
rs368928921 Chr.22: 49999433-49999433 on Build GRCh38
rs202125932 Chr.22: 49999412-49999412 on Build GRCh38
rs755628537 Chr.22: 49999426-49999426 on Build GRCh38
rs1021422002 Chr.22: 49999317-49999317 on Build GRCh38
rs768810910 Chr.22: 49999427-49999427 on Build GRCh38
rs370778377 Chr.22: 49999264-49999264 on Build GRCh38
rs561432969 Chr.22: 49999157-49999157 on Build GRCh38
rs915325258 Chr.22: 49999039-49999039 on Build GRCh38
rs766291643 Chr.22: 49999304-49999304 on Build GRCh38
rs201338275 Chr.22: 49999385-49999385 on Build GRCh38
rs71196391 Chr.22: 49999113-49999114 on Build GRCh38
rs766271959 Chr.22: 49999378-49999378 on Build GRCh38
rs992958158 Chr.22: 49999030-49999033 on Build GRCh38
rs527669587 Chr.22: 49999307-49999307 on Build GRCh38
rs768464422 Chr.22: 49999099-49999100 on Build GRCh38
rs74802784 Chr.22: 49999096-49999096 on Build GRCh38
rs529473785 Chr.22: 49999184-49999184 on Build GRCh38
rs763692273 Chr.22: 49999460-49999460 on Build GRCh38
rs780467586 Chr.22: 49999438-49999439 on Build GRCh38
rs768216136 Chr.22: 49999256-49999256 on Build GRCh38
rs201952246 Chr.22: 49999355-49999355 on Build GRCh38
rs562858324 Chr.22: 49999211-49999211 on Build GRCh38
rs765139228 Chr.22: 49999292-49999292 on Build GRCh38
rs746254074 Chr.22: 49999281-49999281 on Build GRCh38
rs375899805 Chr.22: 49999409-49999409 on Build GRCh38
rs749662208 Chr.22: 49999399-49999399 on Build GRCh38
rs898126508 Chr.22: 49999208-49999208 on Build GRCh38
rs779347746 Chr.22: 49999487-49999487 on Build GRCh38
rs777431910 Chr.22: 49999398-49999398 on Build GRCh38
rs373983547 Chr.22: 49999446-49999446 on Build GRCh38
rs535659851 Chr.22: 49999326-49999326 on Build GRCh38
rs750168754 Chr.22: 49999298-49999298 on Build GRCh38
rs898211911 Chr.22: 49999214-49999214 on Build GRCh38
rs572796126 Chr.22: 49999124-49999124 on Build GRCh38
rs758299121 Chr.22: 49999248-49999248 on Build GRCh38
rs978129162 Chr.22: 49999325-49999325 on Build GRCh38
rs746362770 Chr.22: 49999405-49999405 on Build GRCh38
rs758298700 Chr.22: 49999302-49999302 on Build GRCh38
rs753522201 Chr.22: 49999464-49999464 on Build GRCh38
rs371022898 Chr.22: 49999367-49999367 on Build GRCh38
rs1038548527 Chr.22: 49999196-49999196 on Build GRCh38
rs750777726 Chr.22: 49999489-49999489 on Build GRCh38
rs1035004146 Chr.22: 49999244-49999244 on Build GRCh38
rs745948886 Chr.22: 49999100-49999101 on Build GRCh38
rs751344348 Chr.22: 49999312-49999329 on Build GRCh38
rs921146978 Chr.22: 49999163-49999163 on Build GRCh38
rs946805944 Chr.22: 49999083-49999083 on Build GRCh38
rs761765229 Chr.22: 49999286-49999286 on Build GRCh38
rs1024795530 Chr.22: 49999091-49999091 on Build GRCh38
rs762787734 Chr.22: 49999377-49999377 on Build GRCh38
rs199924155 Chr.22: 49999401-49999401 on Build GRCh38
rs374977841 Chr.22: 49999195-49999195 on Build GRCh38
rs747789553 Chr.22: 49999324-49999324 on Build GRCh38

View Full Product Details