Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 50275687-50276180 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PLXNB2
Entrez Gene ID
23654
Gene Name
plexin B2
Gene Aliases
MM1, Nbla00445, PLEXB2, dJ402G11.3
UniGene
Hs.3989
Gene Chromosome Location
Chr.22: 50274979-50307637 on Build GRCh38
Transcripts
NM_012401.3 XM_005261909.1 XM_005261910.2 XM_005261911.4 XM_006724413.2 XM_011530682.2 XM_017028703.1 XM_017028704.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7154796 c.34G>C p.E12Q Chr.22: 50275707-50275707 on Build GRCh38
4300696 c.135+1C>A p.? Chr.22: 50275809-50275809 on Build GRCh38
4300694 c.106G>A p.G36S Chr.22: 50275779-50275779 on Build GRCh38
4185492 c.65G>A p.G22D Chr.22: 50275738-50275738 on Build GRCh38
4105088 c.147A>G p.V49V Chr.22: 50275900-50275900 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs756799761 Chr.22: 50275747-50275748 on Build GRCh38
rs35380399 Chr.22: 50275836-50275837 on Build GRCh38
rs745450391 Chr.22: 50275895-50275895 on Build GRCh38
rs745539999 Chr.22: 50275846-50275846 on Build GRCh38
rs753662439 Chr.22: 50275943-50275943 on Build GRCh38
rs73891205 Chr.22: 50275821-50275821 on Build GRCh38
rs769097063 Chr.22: 50275858-50275858 on Build GRCh38
rs761096392 Chr.22: 50275809-50275809 on Build GRCh38
rs544368857 Chr.22: 50275877-50275877 on Build GRCh38
rs866516387 Chr.22: 50275738-50275738 on Build GRCh38
rs6010225 Chr.22: 50276112-50276112 on Build GRCh38
rs748113245 Chr.22: 50275758-50275758 on Build GRCh38
rs964427325 Chr.22: 50276155-50276155 on Build GRCh38
rs769581020 Chr.22: 50275766-50275766 on Build GRCh38
rs779042031 Chr.22: 50275731-50275731 on Build GRCh38
rs887084893 Chr.22: 50276038-50276038 on Build GRCh38
rs767661287 Chr.22: 50275940-50275940 on Build GRCh38
rs987161922 Chr.22: 50276138-50276138 on Build GRCh38
rs776039270 Chr.22: 50275862-50275862 on Build GRCh38
rs575107761 Chr.22: 50275749-50275749 on Build GRCh38
rs372835571 Chr.22: 50275961-50275961 on Build GRCh38
rs774389993 Chr.22: 50275981-50275981 on Build GRCh38
rs539166417 Chr.22: 50276033-50276033 on Build GRCh38
rs1056611218 Chr.22: 50275852-50275852 on Build GRCh38
rs748709582 Chr.22: 50275911-50275911 on Build GRCh38
rs778090678 Chr.22: 50275975-50275975 on Build GRCh38
rs199581722 Chr.22: 50275869-50275869 on Build GRCh38
rs765312834 Chr.22: 50275998-50275998 on Build GRCh38
rs554298985 Chr.22: 50276137-50276137 on Build GRCh38
rs968566526 Chr.22: 50276111-50276111 on Build GRCh38
rs771983044 Chr.22: 50275988-50275988 on Build GRCh38
rs565447693 Chr.22: 50275980-50275983 on Build GRCh38
rs373745510 Chr.22: 50275976-50275976 on Build GRCh38
rs991382066 Chr.22: 50275715-50275715 on Build GRCh38
rs765620801 Chr.22: 50275866-50275866 on Build GRCh38
rs763150093 Chr.22: 50275936-50275936 on Build GRCh38
rs1031604462 Chr.22: 50276129-50276129 on Build GRCh38
rs544293369 Chr.22: 50275837-50275837 on Build GRCh38
rs764450803 Chr.22: 50275814-50275814 on Build GRCh38
rs377049625 Chr.22: 50276012-50276012 on Build GRCh38
rs752565989 Chr.22: 50275835-50275835 on Build GRCh38
rs771653017 Chr.22: 50275853-50275853 on Build GRCh38
rs190255875 Chr.22: 50275782-50275782 on Build GRCh38
rs761824594 Chr.22: 50275994-50275994 on Build GRCh38
rs775709318 Chr.22: 50275990-50275990 on Build GRCh38
rs5771118 Chr.22: 50275860-50275860 on Build GRCh38
rs751114927 Chr.22: 50276007-50276007 on Build GRCh38
rs746546029 Chr.22: 50275905-50275905 on Build GRCh38
rs1039892759 Chr.22: 50275859-50275859 on Build GRCh38
rs201524381 Chr.22: 50275815-50275815 on Build GRCh38
rs758189415 Chr.22: 50275898-50275898 on Build GRCh38
rs373532871 Chr.22: 50275908-50275908 on Build GRCh38
rs1004221325 Chr.22: 50276046-50276046 on Build GRCh38
rs887144487 Chr.22: 50275880-50275880 on Build GRCh38
rs750555906 Chr.22: 50275736-50275736 on Build GRCh38
rs754031063 Chr.22: 50275718-50275718 on Build GRCh38
rs1022788349 Chr.22: 50276095-50276095 on Build GRCh38
rs113019596 Chr.22: 50275833-50275833 on Build GRCh38
rs978700987 Chr.22: 50275969-50275969 on Build GRCh38
rs763978926 Chr.22: 50275876-50275876 on Build GRCh38
rs747544496 Chr.22: 50275856-50275856 on Build GRCh38
rs554851279 Chr.22: 50275737-50275737 on Build GRCh38
rs376653010 Chr.22: 50275881-50275881 on Build GRCh38
rs1006353931 Chr.22: 50276076-50276076 on Build GRCh38
rs766637848 Chr.22: 50275937-50275937 on Build GRCh38
rs750299230 Chr.22: 50275952-50275952 on Build GRCh38
rs770085526 Chr.22: 50275913-50275913 on Build GRCh38
rs766217852 Chr.22: 50276005-50276005 on Build GRCh38
rs1023659153 Chr.22: 50276077-50276077 on Build GRCh38
rs113343570 Chr.22: 50275811-50275811 on Build GRCh38
rs376188298 Chr.22: 50275950-50275950 on Build GRCh38
rs750209367 Chr.22: 50275999-50275999 on Build GRCh38
rs773634596 Chr.22: 50275925-50275925 on Build GRCh38
rs180698014 Chr.22: 50275960-50275960 on Build GRCh38
rs746436046 Chr.22: 50275857-50275858 on Build GRCh38
rs921471788 Chr.22: 50276029-50276029 on Build GRCh38
rs562376418 Chr.22: 50275874-50275874 on Build GRCh38
rs988300231 Chr.22: 50276119-50276122 on Build GRCh38
rs372016901 Chr.22: 50275837-50275837 on Build GRCh38
rs149124212 Chr.22: 50275891-50275891 on Build GRCh38
rs780128535 Chr.22: 50275822-50275822 on Build GRCh38
rs527548070 Chr.22: 50276024-50276024 on Build GRCh38
rs779693958 Chr.22: 50275900-50275900 on Build GRCh38
rs561298929 Chr.22: 50275857-50275857 on Build GRCh38
rs759161065 Chr.22: 50275788-50275788 on Build GRCh38
rs368362707 Chr.22: 50275836-50275836 on Build GRCh38
rs1026394421 Chr.22: 50276057-50276057 on Build GRCh38
rs778866905 Chr.22: 50275892-50275892 on Build GRCh38
rs759820199 Chr.22: 50275984-50275984 on Build GRCh38
rs749647201 Chr.22: 50275976-50275976 on Build GRCh38
rs770422474 Chr.22: 50275858-50275863 on Build GRCh38
rs375691632 Chr.22: 50275977-50275977 on Build GRCh38
rs200418901 Chr.22: 50275818-50275818 on Build GRCh38
rs754749475 Chr.22: 50275968-50275968 on Build GRCh38
rs376948709 Chr.22: 50275844-50275844 on Build GRCh38
rs757423052 Chr.22: 50275725-50275725 on Build GRCh38
rs563052435 Chr.22: 50275907-50275907 on Build GRCh38
rs111787707 Chr.22: 50275965-50275965 on Build GRCh38
rs6010224 Chr.22: 50276062-50276062 on Build GRCh38
rs143473905 Chr.22: 50276032-50276032 on Build GRCh38
rs758097644 Chr.22: 50275955-50275955 on Build GRCh38
rs1057454430 Chr.22: 50276054-50276054 on Build GRCh38
rs368982068 Chr.22: 50275845-50275845 on Build GRCh38
rs539241813 Chr.22: 50276133-50276133 on Build GRCh38
rs927599302 Chr.22: 50275875-50275875 on Build GRCh38
rs537976056 Chr.22: 50276037-50276037 on Build GRCh38
rs960358406 Chr.22: 50276117-50276117 on Build GRCh38
rs777436314 Chr.22: 50275777-50275777 on Build GRCh38
rs911646641 Chr.22: 50276000-50276000 on Build GRCh38
rs779890038 Chr.22: 50275956-50275956 on Build GRCh38
rs572649940 Chr.22: 50275819-50275819 on Build GRCh38
rs911580916 Chr.22: 50276152-50276152 on Build GRCh38
rs762794919 Chr.22: 50276001-50276001 on Build GRCh38
rs762124319 Chr.22: 50275864-50275864 on Build GRCh38
rs1022211560 Chr.22: 50275757-50275757 on Build GRCh38
rs773372832 Chr.22: 50275867-50275867 on Build GRCh38
rs751650772 Chr.22: 50275873-50275873 on Build GRCh38
rs748535043 Chr.22: 50275967-50275967 on Build GRCh38
rs375392362 Chr.22: 50275779-50275779 on Build GRCh38
rs1013047603 Chr.22: 50276081-50276081 on Build GRCh38
rs955743186 Chr.22: 50275986-50275986 on Build GRCh38
rs970323795 Chr.22: 50276094-50276095 on Build GRCh38
rs944176002 Chr.22: 50276019-50276019 on Build GRCh38
rs776094413 Chr.22: 50275797-50275797 on Build GRCh38
rs748967251 Chr.22: 50275778-50275778 on Build GRCh38

View Full Product Details