Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 3847220-3847723 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LRRN1
Entrez Gene ID
57633
Gene Name
leucine rich repeat neuronal 1
Gene Aliases
FIGLER3, NLRR-1
UniGene
Hs.163244
Gene Chromosome Location
Chr.3: 3799431-3849834 on Build GRCh38
Transcripts
NM_001324188.1 NM_001324189.1 NM_020873.6

SNP Detail

SNP ID Chromosome Location
rs944201499 Chr.3: 3847634-3847634 on Build GRCh38
rs116619315 Chr.3: 3847330-3847330 on Build GRCh38
rs71040094 Chr.3: 3847397-3847397 on Build GRCh38
rs925558585 Chr.3: 3847548-3847548 on Build GRCh38
rs1007864535 Chr.3: 3847550-3847551 on Build GRCh38
rs575949688 Chr.3: 3847387-3847387 on Build GRCh38
rs913478457 Chr.3: 3847326-3847328 on Build GRCh38
rs779648042 Chr.3: 3847641-3847641 on Build GRCh38
rs199777973 Chr.3: 3847384-3847387 on Build GRCh38
rs574826691 Chr.3: 3847385-3847387 on Build GRCh38
rs957806599 Chr.3: 3847607-3847607 on Build GRCh38
rs927309048 Chr.3: 3847663-3847663 on Build GRCh38
rs71623146 Chr.3: 3847395-3847397 on Build GRCh38
rs981456446 Chr.3: 3847658-3847658 on Build GRCh38
rs771745178 Chr.3: 3847500-3847502 on Build GRCh38
rs1872417 Chr.3: 3847538-3847538 on Build GRCh38
rs144143754 Chr.3: 3847267-3847267 on Build GRCh38
rs1002055624 Chr.3: 3847373-3847373 on Build GRCh38
rs913636796 Chr.3: 3847472-3847472 on Build GRCh38
rs869223121 Chr.3: 3847396-3847397 on Build GRCh38
rs1037444877 Chr.3: 3847285-3847285 on Build GRCh38
rs529461758 Chr.3: 3847499-3847499 on Build GRCh38
rs747192443 Chr.3: 3847567-3847567 on Build GRCh38
rs897194250 Chr.3: 3847296-3847296 on Build GRCh38
rs1049894544 Chr.3: 3847561-3847561 on Build GRCh38
rs898348050 Chr.3: 3847439-3847439 on Build GRCh38
rs764562635 Chr.3: 3847386-3847386 on Build GRCh38
rs200876834 Chr.3: 3847395-3847395 on Build GRCh38
rs991883471 Chr.3: 3847450-3847450 on Build GRCh38
rs905967691 Chr.3: 3847344-3847345 on Build GRCh38
rs201623975 Chr.3: 3847384-3847386 on Build GRCh38
rs78761252 Chr.3: 3847385-3847385 on Build GRCh38
rs747617826 Chr.3: 3847365-3847365 on Build GRCh38
rs200715434 Chr.3: 3847384-3847385 on Build GRCh38
rs769020032 Chr.3: 3847583-3847583 on Build GRCh38
rs187357681 Chr.3: 3847665-3847665 on Build GRCh38
rs933733530 Chr.3: 3847372-3847372 on Build GRCh38
rs995499779 Chr.3: 3847324-3847324 on Build GRCh38
rs932877846 Chr.3: 3847550-3847550 on Build GRCh38
rs562016493 Chr.3: 3847402-3847402 on Build GRCh38
rs963668632 Chr.3: 3847564-3847564 on Build GRCh38
rs573571918 Chr.3: 3847342-3847342 on Build GRCh38
rs966434632 Chr.3: 3847480-3847480 on Build GRCh38
rs913478191 Chr.3: 3847623-3847623 on Build GRCh38
rs1025989146 Chr.3: 3847524-3847528 on Build GRCh38
rs34173470 Chr.3: 3847384-3847384 on Build GRCh38
rs542616562 Chr.3: 3847421-3847421 on Build GRCh38
rs879173511 Chr.3: 3847385-3847385 on Build GRCh38
rs950308467 Chr.3: 3847627-3847627 on Build GRCh38
rs988961151 Chr.3: 3847621-3847621 on Build GRCh38
rs576768169 Chr.3: 3847600-3847600 on Build GRCh38
rs762468360 Chr.3: 3847385-3847386 on Build GRCh38
rs960745405 Chr.3: 3847427-3847427 on Build GRCh38

View Full Product Details