Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 9761548-9762008 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CAMK1
Entrez Gene ID
8536
Gene Name
calcium/calmodulin dependent protein kinase I
Gene Aliases
CAMKI
UniGene
Hs.434875
Gene Chromosome Location
Chr.3: 9757345-9769987 on Build GRCh38
Transcripts
NM_003656.4 XM_005265516.1 XM_005265517.2 XM_017007354.1 NM_016821.2 NM_016826.2 NM_016827.2 NM_016828.2 NM_016829.2 XM_011533760.1 XM_017006494.1 XM_017006496.1 XM_017006497.1 XM_017006498.1 XM_017006499.1

Gene Symbol
OGG1
Entrez Gene ID
4968
Gene Name
8-oxoguanine DNA glycosylase
Gene Aliases
HMMH, HOGG1, MUTM, OGH1
UniGene
Hs.380271
Gene Chromosome Location
Chr.3: 9749944-9788246 on Build GRCh38
Transcripts
NM_003656.4 XM_005265516.1 XM_005265517.2 XM_017007354.1 NM_016821.2 NM_016826.2 NM_016827.2 NM_016828.2 NM_016829.2 XM_011533760.1 XM_017006494.1 XM_017006496.1 XM_017006497.1 XM_017006498.1 XM_017006499.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
447124 c.433G>A p.E145K Chr.3: 9761754-9761754 on Build GRCh38
4435873 c.430-4C>T p.? Chr.3: 9761761-9761761 on Build GRCh38
176914 c.435G>T p.E145D Chr.3: 9761752-9761752 on Build GRCh38
6619155 c.512C>T p.P171L Chr.3: 9761675-9761675 on Build GRCh38
4668930 c.476T>C p.M159T Chr.3: 9761711-9761711 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs772524967 Chr.3: 9761599-9761599 on Build GRCh38
rs973698645 Chr.3: 9761810-9761810 on Build GRCh38
rs564224182 Chr.3: 9761952-9761952 on Build GRCh38
rs753815172 Chr.3: 9761743-9761743 on Build GRCh38
rs9822279 Chr.3: 9761903-9761903 on Build GRCh38
rs749295716 Chr.3: 9761592-9761592 on Build GRCh38
rs373479502 Chr.3: 9761611-9761611 on Build GRCh38
rs747407167 Chr.3: 9761780-9761780 on Build GRCh38
rs959800434 Chr.3: 9761729-9761729 on Build GRCh38
rs532570739 Chr.3: 9761972-9761972 on Build GRCh38
rs925718348 Chr.3: 9761703-9761703 on Build GRCh38
rs768260156 Chr.3: 9761608-9761608 on Build GRCh38
rs761368031 Chr.3: 9761959-9761959 on Build GRCh38
rs293794 Chr.3: 9761943-9761943 on Build GRCh38
rs968739975 Chr.3: 9761902-9761902 on Build GRCh38
rs747319122 Chr.3: 9761679-9761679 on Build GRCh38
rs760416645 Chr.3: 9761724-9761724 on Build GRCh38
rs573587111 Chr.3: 9761623-9761623 on Build GRCh38
rs963174279 Chr.3: 9761822-9761822 on Build GRCh38
rs370403231 Chr.3: 9761626-9761626 on Build GRCh38
rs955513524 Chr.3: 9761893-9761893 on Build GRCh38
rs752506707 Chr.3: 9761719-9761719 on Build GRCh38
rs764630090 Chr.3: 9761614-9761614 on Build GRCh38
rs770520271 Chr.3: 9761704-9761704 on Build GRCh38
rs753550953 Chr.3: 9761658-9761659 on Build GRCh38
rs777891809 Chr.3: 9761584-9761584 on Build GRCh38
rs778506528 Chr.3: 9761958-9761959 on Build GRCh38
rs769307935 Chr.3: 9761683-9761683 on Build GRCh38
rs769181183 Chr.3: 9761787-9761787 on Build GRCh38
rs778829061 Chr.3: 9761665-9761665 on Build GRCh38
rs540971111 Chr.3: 9761655-9761655 on Build GRCh38
rs765977141 Chr.3: 9761625-9761625 on Build GRCh38
rs752480324 Chr.3: 9761642-9761642 on Build GRCh38
rs975576227 Chr.3: 9761607-9761607 on Build GRCh38
rs997360108 Chr.3: 9761876-9761876 on Build GRCh38
rs572307672 Chr.3: 9761760-9761760 on Build GRCh38
rs757121601 Chr.3: 9761746-9761746 on Build GRCh38
rs376170684 Chr.3: 9761575-9761575 on Build GRCh38
rs767399471 Chr.3: 9761639-9761639 on Build GRCh38
rs1053073 Chr.3: 9761801-9761801 on Build GRCh38
rs550091798 Chr.3: 9761936-9761936 on Build GRCh38
rs895671081 Chr.3: 9761786-9761786 on Build GRCh38
rs559224420 Chr.3: 9761705-9761705 on Build GRCh38
rs746152391 Chr.3: 9761674-9761674 on Build GRCh38
rs369444327 Chr.3: 9761596-9761596 on Build GRCh38
rs902884716 Chr.3: 9761654-9761654 on Build GRCh38
rs529514510 Chr.3: 9761983-9761983 on Build GRCh38
rs973320104 Chr.3: 9761823-9761823 on Build GRCh38
rs756037100 Chr.3: 9761577-9761577 on Build GRCh38
rs746763417 Chr.3: 9761960-9761960 on Build GRCh38
rs764052828 Chr.3: 9761735-9761735 on Build GRCh38
rs947553130 Chr.3: 9761946-9761946 on Build GRCh38
rs373676357 Chr.3: 9761627-9761627 on Build GRCh38
rs780569645 Chr.3: 9761675-9761675 on Build GRCh38
rs1014096001 Chr.3: 9761819-9761820 on Build GRCh38
rs756053756 Chr.3: 9761645-9761645 on Build GRCh38
rs780373289 Chr.3: 9761779-9761779 on Build GRCh38
rs1029808638 Chr.3: 9761884-9761884 on Build GRCh38
rs912529292 Chr.3: 9761895-9761895 on Build GRCh38
rs188672077 Chr.3: 9761621-9761621 on Build GRCh38
rs757321703 Chr.3: 9761659-9761659 on Build GRCh38
rs914155532 Chr.3: 9761898-9761898 on Build GRCh38
rs145978414 Chr.3: 9761635-9761635 on Build GRCh38
rs1022819308 Chr.3: 9761737-9761737 on Build GRCh38
rs546193931 Chr.3: 9761824-9761824 on Build GRCh38
rs762376373 Chr.3: 9761694-9761694 on Build GRCh38
rs777236576 Chr.3: 9761684-9761684 on Build GRCh38
rs1039727841 Chr.3: 9761918-9761918 on Build GRCh38
rs771069252 Chr.3: 9761594-9761594 on Build GRCh38
rs775695293 Chr.3: 9761608-9761608 on Build GRCh38
rs934347724 Chr.3: 9761964-9761964 on Build GRCh38
rs758659775 Chr.3: 9761774-9761774 on Build GRCh38
rs951489369 Chr.3: 9761842-9761842 on Build GRCh38
rs1053072 Chr.3: 9761766-9761766 on Build GRCh38
rs564395339 Chr.3: 9761856-9761856 on Build GRCh38
rs767015539 Chr.3: 9761712-9761712 on Build GRCh38
rs945677826 Chr.3: 9761667-9761667 on Build GRCh38
rs937082797 Chr.3: 9761954-9761954 on Build GRCh38
rs1012649189 Chr.3: 9761730-9761730 on Build GRCh38
rs377236766 Chr.3: 9761656-9761656 on Build GRCh38
rs1005547212 Chr.3: 9761831-9761831 on Build GRCh38
rs919173833 Chr.3: 9761838-9761838 on Build GRCh38
rs746247676 Chr.3: 9761597-9761597 on Build GRCh38

View Full Product Details