Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 443579-444094 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABCA11P
Entrez Gene ID
79963
Gene Name
ATP binding cassette subfamily A member 11, pseudogene
Gene Aliases
ABCA11, EST1133530
UniGene
Hs.677493
Gene Chromosome Location
Chr.4: 425435-474209 on Build GRCh38
Transcripts
NR_002451.2 NM_133474.3

Gene Symbol
ZNF721
Entrez Gene ID
170960
Gene Name
zinc finger protein 721
Gene Aliases
-
UniGene
Hs.428360
Gene Chromosome Location
Chr.4: 439984-499171 on Build GRCh38
Transcripts
NR_002451.2 NM_133474.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
587469 c.168T>C p.F56F Chr.4: 443609-443609 on Build GRCh38
265138 c.471A>C p.K157N Chr.4: 443996-443996 on Build GRCh38
394798 c.492T>C p.P164P Chr.4: 443975-443975 on Build GRCh38
346752 c.647C>G p.T216S Chr.4: 443820-443820 on Build GRCh38
4571713 c.522T>A p.F174L Chr.4: 443945-443945 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs782478719 Chr.4: 444029-444029 on Build GRCh38
rs782529181 Chr.4: 443752-443752 on Build GRCh38
rs188454118 Chr.4: 443978-443978 on Build GRCh38
rs372601849 Chr.4: 443822-443822 on Build GRCh38
rs782267392 Chr.4: 443951-443951 on Build GRCh38
rs782410956 Chr.4: 443871-443871 on Build GRCh38
rs781958248 Chr.4: 443992-443992 on Build GRCh38
rs549384435 Chr.4: 444015-444015 on Build GRCh38
rs374614076 Chr.4: 443647-443647 on Build GRCh38
rs781971482 Chr.4: 443884-443884 on Build GRCh38
rs782499763 Chr.4: 443935-443935 on Build GRCh38
rs782645432 Chr.4: 443764-443764 on Build GRCh38
rs781953802 Chr.4: 443607-443607 on Build GRCh38
rs782390817 Chr.4: 444056-444056 on Build GRCh38
rs782742309 Chr.4: 443826-443826 on Build GRCh38
rs1016552350 Chr.4: 443772-443772 on Build GRCh38
rs781795838 Chr.4: 443924-443924 on Build GRCh38
rs782410415 Chr.4: 443981-443981 on Build GRCh38
rs192018843 Chr.4: 443759-443759 on Build GRCh38
rs993646191 Chr.4: 444052-444052 on Build GRCh38
rs782139949 Chr.4: 444000-444000 on Build GRCh38
rs782656713 Chr.4: 443860-443860 on Build GRCh38
rs782245693 Chr.4: 443765-443765 on Build GRCh38
rs781896562 Chr.4: 443748-443748 on Build GRCh38
rs781818614 Chr.4: 443829-443829 on Build GRCh38
rs368280658 Chr.4: 443813-443813 on Build GRCh38
rs782507651 Chr.4: 443743-443743 on Build GRCh38
rs1023065306 Chr.4: 443751-443751 on Build GRCh38
rs781827787 Chr.4: 443733-443733 on Build GRCh38
rs781912933 Chr.4: 443669-443669 on Build GRCh38
rs782206149 Chr.4: 443769-443769 on Build GRCh38
rs782769152 Chr.4: 443988-443988 on Build GRCh38
rs782225038 Chr.4: 443970-443970 on Build GRCh38
rs782155360 Chr.4: 443814-443814 on Build GRCh38
rs782499459 Chr.4: 443835-443835 on Build GRCh38
rs782273290 Chr.4: 443653-443653 on Build GRCh38
rs116742561 Chr.4: 443931-443931 on Build GRCh38
rs1026910746 Chr.4: 444053-444053 on Build GRCh38
rs182954427 Chr.4: 443838-443838 on Build GRCh38
rs1002436872 Chr.4: 443761-443761 on Build GRCh38
rs782803961 Chr.4: 443710-443710 on Build GRCh38
rs367900463 Chr.4: 443731-443731 on Build GRCh38
rs782349297 Chr.4: 443858-443858 on Build GRCh38
rs375487945 Chr.4: 443808-443808 on Build GRCh38
rs782261910 Chr.4: 444035-444035 on Build GRCh38
rs919308563 Chr.4: 443785-443785 on Build GRCh38
rs202057586 Chr.4: 444033-444033 on Build GRCh38
rs201712232 Chr.4: 443836-443836 on Build GRCh38
rs370188046 Chr.4: 443904-443904 on Build GRCh38
rs782458115 Chr.4: 443960-443960 on Build GRCh38
rs782452507 Chr.4: 443851-443851 on Build GRCh38
rs782315918 Chr.4: 443667-443667 on Build GRCh38
rs200898264 Chr.4: 444041-444041 on Build GRCh38
rs782810116 Chr.4: 444013-444013 on Build GRCh38
rs781972052 Chr.4: 443780-443780 on Build GRCh38
rs1044174140 Chr.4: 444018-444018 on Build GRCh38
rs373050502 Chr.4: 443612-443612 on Build GRCh38
rs782080575 Chr.4: 443890-443890 on Build GRCh38
rs972680956 Chr.4: 443777-443777 on Build GRCh38
rs781989897 Chr.4: 443704-443704 on Build GRCh38
rs781880405 Chr.4: 443934-443934 on Build GRCh38
rs781793550 Chr.4: 443629-443629 on Build GRCh38
rs200907568 Chr.4: 443773-443773 on Build GRCh38
rs782147187 Chr.4: 443899-443899 on Build GRCh38
rs371331043 Chr.4: 443929-443929 on Build GRCh38
rs369596095 Chr.4: 444040-444040 on Build GRCh38
rs782091479 Chr.4: 443743-443743 on Build GRCh38
rs782173767 Chr.4: 443706-443706 on Build GRCh38
rs782766447 Chr.4: 443891-443891 on Build GRCh38
rs376114031 Chr.4: 444014-444014 on Build GRCh38
rs782390093 Chr.4: 443787-443787 on Build GRCh38
rs782589467 Chr.4: 443631-443631 on Build GRCh38
rs782139734 Chr.4: 443611-443611 on Build GRCh38
rs782575300 Chr.4: 443853-443853 on Build GRCh38
rs782685953 Chr.4: 443604-443604 on Build GRCh38
rs782756351 Chr.4: 443747-443747 on Build GRCh38
rs781929386 Chr.4: 444020-444020 on Build GRCh38
rs781990772 Chr.4: 443684-443684 on Build GRCh38
rs781809624 Chr.4: 443817-443817 on Build GRCh38
rs782312254 Chr.4: 443878-443878 on Build GRCh38
rs369127753 Chr.4: 443972-443972 on Build GRCh38
rs543482721 Chr.4: 444030-444030 on Build GRCh38
rs531329096 Chr.4: 443865-443865 on Build GRCh38
rs200530483 Chr.4: 443754-443754 on Build GRCh38
rs371495220 Chr.4: 443648-443648 on Build GRCh38
rs781941891 Chr.4: 443982-443982 on Build GRCh38
rs376631876 Chr.4: 443736-443736 on Build GRCh38
rs904349723 Chr.4: 443628-443628 on Build GRCh38
rs782237235 Chr.4: 443856-443856 on Build GRCh38
rs782729202 Chr.4: 443683-443683 on Build GRCh38
rs782385298 Chr.4: 443716-443716 on Build GRCh38
rs781975068 Chr.4: 444054-444054 on Build GRCh38
rs781941325 Chr.4: 443712-443712 on Build GRCh38
rs374982400 Chr.4: 443692-443692 on Build GRCh38
rs376255771 Chr.4: 443621-443621 on Build GRCh38
rs782048085 Chr.4: 443983-443983 on Build GRCh38
rs200542139 Chr.4: 443839-443839 on Build GRCh38
rs199512799 Chr.4: 443632-443632 on Build GRCh38
rs550304830 Chr.4: 443965-443965 on Build GRCh38
rs782057251 Chr.4: 443714-443714 on Build GRCh38
rs782493892 Chr.4: 443926-443926 on Build GRCh38
rs145798212 Chr.4: 444043-444043 on Build GRCh38
rs782536694 Chr.4: 443615-443615 on Build GRCh38
rs782669527 Chr.4: 443768-443768 on Build GRCh38
rs72501950 Chr.4: 444042-444042 on Build GRCh38
rs569899886 Chr.4: 443971-443971 on Build GRCh38
rs372873028 Chr.4: 444013-444013 on Build GRCh38
rs781979057 Chr.4: 443797-443797 on Build GRCh38
rs782216868 Chr.4: 444055-444055 on Build GRCh38
rs782084183 Chr.4: 443786-443786 on Build GRCh38
rs782301352 Chr.4: 443943-443943 on Build GRCh38
rs782418578 Chr.4: 443766-443766 on Build GRCh38
rs200730702 Chr.4: 443863-443863 on Build GRCh38
rs781904164 Chr.4: 443893-443893 on Build GRCh38
rs547673991 Chr.4: 443968-443968 on Build GRCh38
rs370185676 Chr.4: 443680-443680 on Build GRCh38
rs781885961 Chr.4: 443848-443848 on Build GRCh38

View Full Product Details