Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 73870807-73871356 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CXCL1
Entrez Gene ID
2919
Gene Name
C-X-C motif chemokine ligand 1
Gene Aliases
FSP, GRO1, GROa, MGSA, MGSA-a, NAP-3, SCYB1
UniGene
Hs.708652
Gene Chromosome Location
Chr.4: 73869392-73871302 on Build GRCh38
Transcripts
NM_001511.3 NR_046035.1

SNP Detail

SNP ID Chromosome Location
rs1049669382 Chr.4: 73870931-73870931 on Build GRCh38
rs111301726 Chr.4: 73871297-73871297 on Build GRCh38
rs373569191 Chr.4: 73870986-73870987 on Build GRCh38
rs542911276 Chr.4: 73871171-73871171 on Build GRCh38
rs566583028 Chr.4: 73870974-73870975 on Build GRCh38
rs889217196 Chr.4: 73871307-73871307 on Build GRCh38
rs113554285 Chr.4: 73870918-73870918 on Build GRCh38
rs909080167 Chr.4: 73871098-73871098 on Build GRCh38
rs917548131 Chr.4: 73871177-73871177 on Build GRCh38
rs375408676 Chr.4: 73870928-73870929 on Build GRCh38
rs949788399 Chr.4: 73870852-73870852 on Build GRCh38
rs893329023 Chr.4: 73871234-73871234 on Build GRCh38
rs1029056180 Chr.4: 73871014-73871014 on Build GRCh38
rs544472543 Chr.4: 73871266-73871266 on Build GRCh38
rs990646743 Chr.4: 73871322-73871322 on Build GRCh38
rs889182995 Chr.4: 73871068-73871068 on Build GRCh38
rs902133247 Chr.4: 73871257-73871257 on Build GRCh38
rs532221985 Chr.4: 73871295-73871295 on Build GRCh38
rs371415385 Chr.4: 73870932-73870932 on Build GRCh38
rs1054597920 Chr.4: 73871226-73871226 on Build GRCh38
rs1020199401 Chr.4: 73871127-73871127 on Build GRCh38
rs386676045 Chr.4: 73870974-73870986 on Build GRCh38
rs183378609 Chr.4: 73871281-73871281 on Build GRCh38
rs111913680 Chr.4: 73870996-73870996 on Build GRCh38
rs757128461 Chr.4: 73870847-73870847 on Build GRCh38
rs917600673 Chr.4: 73870924-73870924 on Build GRCh38
rs1042092379 Chr.4: 73871186-73871186 on Build GRCh38
rs959205342 Chr.4: 73871304-73871304 on Build GRCh38
rs766192794 Chr.4: 73870884-73870884 on Build GRCh38
rs981229793 Chr.4: 73870863-73870863 on Build GRCh38
rs971076090 Chr.4: 73871141-73871141 on Build GRCh38
rs573316994 Chr.4: 73871198-73871198 on Build GRCh38
rs997578059 Chr.4: 73871059-73871059 on Build GRCh38
rs1017596481 Chr.4: 73871002-73871002 on Build GRCh38
rs562459448 Chr.4: 73871300-73871300 on Build GRCh38
rs909132111 Chr.4: 73870904-73870904 on Build GRCh38
rs984981659 Chr.4: 73871246-73871246 on Build GRCh38
rs540839136 Chr.4: 73871245-73871245 on Build GRCh38
rs1045177189 Chr.4: 73870945-73870945 on Build GRCh38
rs909339303 Chr.4: 73871258-73871258 on Build GRCh38
rs990699236 Chr.4: 73871145-73871145 on Build GRCh38
rs997923762 Chr.4: 73871280-73871280 on Build GRCh38
rs868255147 Chr.4: 73871311-73871311 on Build GRCh38
rs1001317244 Chr.4: 73871314-73871314 on Build GRCh38
rs112791438 Chr.4: 73870986-73870986 on Build GRCh38
rs556105521 Chr.4: 73871084-73871084 on Build GRCh38
rs1006691466 Chr.4: 73871069-73871069 on Build GRCh38
rs1957068 Chr.4: 73871188-73871188 on Build GRCh38
rs937953701 Chr.4: 73870907-73870907 on Build GRCh38
rs989099195 Chr.4: 73871118-73871118 on Build GRCh38
rs762690344 Chr.4: 73870878-73870878 on Build GRCh38
rs902292477 Chr.4: 73870939-73870939 on Build GRCh38
rs949063688 Chr.4: 73871185-73871185 on Build GRCh38
rs957312289 Chr.4: 73871092-73871092 on Build GRCh38
rs200339008 Chr.4: 73870934-73870934 on Build GRCh38
rs966039050 Chr.4: 73871030-73871030 on Build GRCh38
rs116303802 Chr.4: 73870997-73870997 on Build GRCh38
rs940901483 Chr.4: 73870973-73870973 on Build GRCh38

View Full Product Details