Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 75913783-75914321 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NAAA
Entrez Gene ID
27163
Gene Name
N-acylethanolamine acid amidase
Gene Aliases
ASAHL, PLT
UniGene
Hs.437365
Gene Chromosome Location
Chr.4: 75910655-75941013 on Build GRCh38
Transcripts
NM_001042402.1 NM_014435.3 XM_005262920.3 XM_005262923.2 XM_005262924.2 XM_006714180.2 XM_017008027.1 XM_017008028.1 XM_017008030.1

SNP Detail

SNP ID Chromosome Location
rs781094029 Chr.4: 75913880-75913880 on Build GRCh38
rs988949426 Chr.4: 75913846-75913846 on Build GRCh38
rs750077889 Chr.4: 75913925-75913925 on Build GRCh38
rs372396263 Chr.4: 75914064-75914064 on Build GRCh38
rs548638282 Chr.4: 75913914-75913914 on Build GRCh38
rs958023718 Chr.4: 75914163-75914163 on Build GRCh38
rs922988258 Chr.4: 75913966-75913966 on Build GRCh38
rs532903688 Chr.4: 75913868-75913868 on Build GRCh38
rs908914501 Chr.4: 75913986-75913986 on Build GRCh38
rs371645724 Chr.4: 75914138-75914138 on Build GRCh38
rs111717782 Chr.4: 75914031-75914031 on Build GRCh38
rs1032754368 Chr.4: 75914161-75914161 on Build GRCh38
rs1004775145 Chr.4: 75914133-75914133 on Build GRCh38
rs887702058 Chr.4: 75914089-75914089 on Build GRCh38
rs975924976 Chr.4: 75913956-75913956 on Build GRCh38
rs1031936993 Chr.4: 75914225-75914225 on Build GRCh38
rs531407421 Chr.4: 75914210-75914210 on Build GRCh38
rs1013679169 Chr.4: 75914088-75914088 on Build GRCh38
rs950159430 Chr.4: 75913910-75913910 on Build GRCh38
rs374582678 Chr.4: 75914217-75914217 on Build GRCh38
rs892745226 Chr.4: 75914023-75914023 on Build GRCh38
rs950513609 Chr.4: 75914262-75914262 on Build GRCh38
rs754868417 Chr.4: 75913820-75913820 on Build GRCh38
rs957634575 Chr.4: 75914279-75914279 on Build GRCh38
rs552771896 Chr.4: 75914121-75914121 on Build GRCh38
rs534537910 Chr.4: 75913974-75913974 on Build GRCh38
rs1513890 Chr.4: 75914211-75914211 on Build GRCh38
rs901659114 Chr.4: 75914153-75914153 on Build GRCh38
rs999649841 Chr.4: 75914154-75914154 on Build GRCh38
rs936219863 Chr.4: 75913839-75913839 on Build GRCh38
rs1513891 Chr.4: 75914040-75914040 on Build GRCh38
rs910472579 Chr.4: 75913901-75913901 on Build GRCh38
rs58103426 Chr.4: 75914157-75914157 on Build GRCh38
rs538011561 Chr.4: 75914042-75914042 on Build GRCh38
rs1006902201 Chr.4: 75914169-75914169 on Build GRCh38
rs11539367 Chr.4: 75913865-75913865 on Build GRCh38
rs1046216783 Chr.4: 75914003-75914003 on Build GRCh38
rs181287132 Chr.4: 75913953-75913953 on Build GRCh38
rs1050460 Chr.4: 75914215-75914215 on Build GRCh38
rs17001192 Chr.4: 75913815-75913815 on Build GRCh38
rs913376107 Chr.4: 75913867-75913867 on Build GRCh38
rs1047838122 Chr.4: 75913982-75913982 on Build GRCh38
rs905896949 Chr.4: 75914019-75914019 on Build GRCh38
rs1032372836 Chr.4: 75913842-75913842 on Build GRCh38
rs997306388 Chr.4: 75914155-75914155 on Build GRCh38
rs562969358 Chr.4: 75913848-75913848 on Build GRCh38
rs1014784002 Chr.4: 75914134-75914134 on Build GRCh38
rs567049210 Chr.4: 75913845-75913845 on Build GRCh38
rs964505304 Chr.4: 75913918-75913918 on Build GRCh38
rs545799486 Chr.4: 75913885-75913886 on Build GRCh38
rs1022423478 Chr.4: 75914074-75914074 on Build GRCh38
rs868120140 Chr.4: 75913937-75913937 on Build GRCh38
rs990104766 Chr.4: 75913890-75913890 on Build GRCh38
rs150612684 Chr.4: 75914224-75914224 on Build GRCh38
rs754438895 Chr.4: 75914218-75914218 on Build GRCh38
rs566865173 Chr.4: 75913930-75913930 on Build GRCh38
rs1046578581 Chr.4: 75914126-75914126 on Build GRCh38
rs530197819 Chr.4: 75913863-75913863 on Build GRCh38

View Full Product Details