Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 77714040-77714585 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CNOT6L
Entrez Gene ID
246175
Gene Name
CCR4-NOT transcription complex subunit 6 like
Gene Aliases
CCR4b
UniGene
Hs.592519
Gene Chromosome Location
Chr.4: 77713387-77820238 on Build GRCh38
Transcripts
NM_001286790.1 NM_144571.2 XM_011531806.2 XM_011531807.2 XM_011531808.2 XM_011531809.1 XM_011531810.1 XM_017007956.1 XM_017007957.1

SNP Detail

SNP ID Chromosome Location
rs977876750 Chr.4: 77714247-77714247 on Build GRCh38
rs1047164796 Chr.4: 77714342-77714343 on Build GRCh38
rs1296473 Chr.4: 77714228-77714228 on Build GRCh38
rs898591000 Chr.4: 77714088-77714088 on Build GRCh38
rs1013635244 Chr.4: 77714264-77714264 on Build GRCh38
rs886395518 Chr.4: 77714437-77714460 on Build GRCh38
rs773716379 Chr.4: 77714440-77714462 on Build GRCh38
rs551085629 Chr.4: 77714109-77714109 on Build GRCh38
rs376165838 Chr.4: 77714136-77714137 on Build GRCh38
rs543184488 Chr.4: 77714420-77714420 on Build GRCh38
rs967730503 Chr.4: 77714533-77714533 on Build GRCh38
rs900090827 Chr.4: 77714446-77714446 on Build GRCh38
rs898540198 Chr.4: 77714334-77714337 on Build GRCh38
rs113193969 Chr.4: 77714250-77714250 on Build GRCh38
rs916503350 Chr.4: 77714287-77714287 on Build GRCh38
rs1299928 Chr.4: 77714219-77714219 on Build GRCh38
rs1012276589 Chr.4: 77714489-77714489 on Build GRCh38
rs932086706 Chr.4: 77714550-77714550 on Build GRCh38
rs974438207 Chr.4: 77714212-77714212 on Build GRCh38
rs967784290 Chr.4: 77714244-77714244 on Build GRCh38
rs1018543834 Chr.4: 77714145-77714145 on Build GRCh38
rs187078942 Chr.4: 77714084-77714084 on Build GRCh38
rs750550501 Chr.4: 77714441-77714463 on Build GRCh38
rs531109513 Chr.4: 77714366-77714366 on Build GRCh38
rs572795670 Chr.4: 77714348-77714349 on Build GRCh38
rs878887559 Chr.4: 77714439-77714460 on Build GRCh38
rs531025897 Chr.4: 77714453-77714475 on Build GRCh38
rs982636622 Chr.4: 77714075-77714075 on Build GRCh38
rs907900142 Chr.4: 77714093-77714093 on Build GRCh38
rs771069800 Chr.4: 77714441-77714462 on Build GRCh38
rs573588142 Chr.4: 77714289-77714289 on Build GRCh38
rs574121857 Chr.4: 77714195-77714195 on Build GRCh38
rs1021967609 Chr.4: 77714520-77714520 on Build GRCh38
rs976529318 Chr.4: 77714395-77714395 on Build GRCh38
rs982666028 Chr.4: 77714376-77714376 on Build GRCh38
rs901959215 Chr.4: 77714185-77714185 on Build GRCh38
rs113140311 Chr.4: 77714429-77714429 on Build GRCh38
rs7654331 Chr.4: 77714115-77714115 on Build GRCh38
rs571319083 Chr.4: 77714229-77714229 on Build GRCh38
rs952527312 Chr.4: 77714338-77714338 on Build GRCh38
rs1008416989 Chr.4: 77714431-77714432 on Build GRCh38
rs760125968 Chr.4: 77714354-77714354 on Build GRCh38
rs1040470143 Chr.4: 77714111-77714111 on Build GRCh38
rs777531092 Chr.4: 77714439-77714462 on Build GRCh38
rs904009091 Chr.4: 77714226-77714226 on Build GRCh38
rs201499481 Chr.4: 77714439-77714461 on Build GRCh38
rs1050606745 Chr.4: 77714209-77714209 on Build GRCh38
rs869262556 Chr.4: 77714438-77714461 on Build GRCh38
rs191958468 Chr.4: 77714263-77714263 on Build GRCh38
rs965316732 Chr.4: 77714390-77714390 on Build GRCh38
rs1008884011 Chr.4: 77714138-77714139 on Build GRCh38
rs1022019568 Chr.4: 77714223-77714223 on Build GRCh38
rs1298043 Chr.4: 77714255-77714255 on Build GRCh38
rs908433937 Chr.4: 77714386-77714386 on Build GRCh38
rs1025763810 Chr.4: 77714124-77714124 on Build GRCh38
rs923277559 Chr.4: 77714483-77714483 on Build GRCh38
rs528911352 Chr.4: 77714432-77714432 on Build GRCh38
rs749521449 Chr.4: 77714314-77714314 on Build GRCh38
rs561660011 Chr.4: 77714360-77714361 on Build GRCh38
rs771664358 Chr.4: 77714125-77714125 on Build GRCh38
rs565357387 Chr.4: 77714311-77714311 on Build GRCh38
rs545646665 Chr.4: 77714303-77714303 on Build GRCh38

View Full Product Details