Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 82422884-82423394 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HNRNPDL
Entrez Gene ID
9987
Gene Name
heterogeneous nuclear ribonucleoprotein D like
Gene Aliases
HNRNP, HNRPDL, JKTBP, JKTBP2, LGMD1G, laAUF1
UniGene
Hs.527105
Gene Chromosome Location
Chr.4: 82422564-82430225 on Build GRCh38
Transcripts
NM_001207000.1 NM_031372.3 NR_003249.2

SNP Detail

SNP ID Chromosome Location
rs908309367 Chr.4: 82423025-82423025 on Build GRCh38
rs551276009 Chr.4: 82423269-82423269 on Build GRCh38
rs11932284 Chr.4: 82422986-82422986 on Build GRCh38
rs996745275 Chr.4: 82423152-82423152 on Build GRCh38
rs1802543 Chr.4: 82423353-82423353 on Build GRCh38
rs140163431 Chr.4: 82422972-82422972 on Build GRCh38
rs534861091 Chr.4: 82422992-82422992 on Build GRCh38
rs141564694 Chr.4: 82423318-82423318 on Build GRCh38
rs546580080 Chr.4: 82423356-82423356 on Build GRCh38
rs889277823 Chr.4: 82422923-82422923 on Build GRCh38
rs543959126 Chr.4: 82423029-82423029 on Build GRCh38
rs939176129 Chr.4: 82422999-82422999 on Build GRCh38
rs1029607881 Chr.4: 82422918-82422918 on Build GRCh38
rs1021485809 Chr.4: 82423163-82423163 on Build GRCh38
rs1802544 Chr.4: 82423369-82423369 on Build GRCh38
rs968406175 Chr.4: 82422984-82422984 on Build GRCh38
rs929463341 Chr.4: 82423246-82423246 on Build GRCh38
rs150917590 Chr.4: 82423319-82423319 on Build GRCh38
rs202203776 Chr.4: 82423066-82423066 on Build GRCh38
rs550145830 Chr.4: 82422952-82422952 on Build GRCh38
rs567950798 Chr.4: 82423245-82423245 on Build GRCh38
rs1018992828 Chr.4: 82422960-82422960 on Build GRCh38
rs564773986 Chr.4: 82423281-82423281 on Build GRCh38
rs1050700673 Chr.4: 82423158-82423158 on Build GRCh38
rs769725600 Chr.4: 82423212-82423212 on Build GRCh38
rs1047859520 Chr.4: 82423274-82423274 on Build GRCh38
rs900542616 Chr.4: 82423315-82423315 on Build GRCh38
rs563791539 Chr.4: 82423039-82423039 on Build GRCh38
rs1008978700 Chr.4: 82422925-82422925 on Build GRCh38
rs1047882184 Chr.4: 82423100-82423100 on Build GRCh38
rs1801067 Chr.4: 82423288-82423288 on Build GRCh38
rs755186869 Chr.4: 82423183-82423183 on Build GRCh38
rs780549942 Chr.4: 82423187-82423187 on Build GRCh38
rs915554790 Chr.4: 82423200-82423200 on Build GRCh38
rs137931240 Chr.4: 82423049-82423068 on Build GRCh38
rs920785357 Chr.4: 82423055-82423055 on Build GRCh38
rs1039513970 Chr.4: 82423296-82423296 on Build GRCh38
rs879517039 Chr.4: 82422915-82422915 on Build GRCh38
rs920845422 Chr.4: 82423236-82423236 on Build GRCh38
rs767932349 Chr.4: 82423310-82423311 on Build GRCh38
rs930927857 Chr.4: 82423080-82423080 on Build GRCh38
rs146129186 Chr.4: 82422970-82422970 on Build GRCh38
rs543017954 Chr.4: 82423145-82423145 on Build GRCh38
rs560443083 Chr.4: 82422941-82422941 on Build GRCh38
rs889331573 Chr.4: 82423160-82423160 on Build GRCh38
rs911978157 Chr.4: 82423276-82423277 on Build GRCh38
rs998652082 Chr.4: 82423340-82423340 on Build GRCh38
rs566242493 Chr.4: 82423366-82423366 on Build GRCh38
rs758811072 Chr.4: 82423156-82423156 on Build GRCh38
rs780259027 Chr.4: 82423172-82423172 on Build GRCh38
rs530501276 Chr.4: 82423311-82423311 on Build GRCh38
rs779520094 Chr.4: 82423226-82423229 on Build GRCh38
rs188817622 Chr.4: 82423211-82423211 on Build GRCh38
rs574046440 Chr.4: 82423134-82423134 on Build GRCh38
rs956766615 Chr.4: 82422968-82422968 on Build GRCh38
rs959780872 Chr.4: 82423196-82423197 on Build GRCh38
rs904408523 Chr.4: 82423322-82423322 on Build GRCh38
rs193014392 Chr.4: 82422969-82422969 on Build GRCh38
rs369680059 Chr.4: 82423215-82423215 on Build GRCh38
rs368344815 Chr.4: 82423069-82423069 on Build GRCh38
rs995086560 Chr.4: 82422913-82422913 on Build GRCh38
rs760328821 Chr.4: 82423323-82423325 on Build GRCh38
rs998640633 Chr.4: 82423174-82423174 on Build GRCh38
rs924374803 Chr.4: 82422997-82422997 on Build GRCh38
rs869100438 Chr.4: 82423048-82423068 on Build GRCh38
rs536792596 Chr.4: 82422947-82422947 on Build GRCh38
rs772489105 Chr.4: 82423170-82423173 on Build GRCh38
rs578132115 Chr.4: 82423010-82423010 on Build GRCh38
rs760055182 Chr.4: 82423190-82423193 on Build GRCh38
rs899767404 Chr.4: 82423151-82423151 on Build GRCh38
rs1009322415 Chr.4: 82423143-82423143 on Build GRCh38
rs200246259 Chr.4: 82423067-82423067 on Build GRCh38
rs77885237 Chr.4: 82423198-82423198 on Build GRCh38
rs751817353 Chr.4: 82423176-82423176 on Build GRCh38
rs929453256 Chr.4: 82423075-82423075 on Build GRCh38
rs898150559 Chr.4: 82423044-82423044 on Build GRCh38
rs991964916 Chr.4: 82423013-82423013 on Build GRCh38
rs367896765 Chr.4: 82422993-82422993 on Build GRCh38

View Full Product Details