Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 28388872-28389376 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZSCAN12
Entrez Gene ID
9753
Gene Name
zinc finger and SCAN domain containing 12
Gene Aliases
ZFP96, ZNF29K1, ZNF305, ZNF96, dJ29K1.2
UniGene
Hs.134816
Gene Chromosome Location
Chr.6: 28378821-28399768 on Build GRCh38
Transcripts
NM_001163391.1 NR_028077.1 NR_136509.1 NR_136510.1 NR_136511.1 NR_136512.1 NR_136513.1 XM_011515014.2 XM_011515015.2 XM_011515017.2 XM_017011528.1

SNP Detail

SNP ID Chromosome Location
rs761563651 Chr.6: 28389139-28389139 on Build GRCh38
rs889266300 Chr.6: 28388898-28388898 on Build GRCh38
rs966182463 Chr.6: 28389085-28389085 on Build GRCh38
rs1035690805 Chr.6: 28389081-28389081 on Build GRCh38
rs749972422 Chr.6: 28389335-28389335 on Build GRCh38
rs907286676 Chr.6: 28388949-28388949 on Build GRCh38
rs924515632 Chr.6: 28389307-28389307 on Build GRCh38
rs962616305 Chr.6: 28389046-28389046 on Build GRCh38
rs916020297 Chr.6: 28389191-28389191 on Build GRCh38
rs892329055 Chr.6: 28389015-28389015 on Build GRCh38
rs1016489969 Chr.6: 28389003-28389003 on Build GRCh38
rs1009421532 Chr.6: 28389041-28389041 on Build GRCh38
rs960061271 Chr.6: 28389086-28389086 on Build GRCh38
rs1006821443 Chr.6: 28388944-28388944 on Build GRCh38
rs1056693930 Chr.6: 28388952-28388952 on Build GRCh38
rs950882865 Chr.6: 28389223-28389223 on Build GRCh38
rs948715104 Chr.6: 28388912-28388912 on Build GRCh38
rs775709670 Chr.6: 28389009-28389009 on Build GRCh38
rs1017314522 Chr.6: 28389062-28389062 on Build GRCh38
rs1007187085 Chr.6: 28389057-28389057 on Build GRCh38
rs972051558 Chr.6: 28389054-28389054 on Build GRCh38
rs146587637 Chr.6: 28389252-28389252 on Build GRCh38
rs192420796 Chr.6: 28389237-28389237 on Build GRCh38

View Full Product Details