Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 28534550-28535016 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GPX5
Entrez Gene ID
2880
Gene Name
glutathione peroxidase 5
Gene Aliases
HEL-S-75p
UniGene
Hs.248129
Gene Chromosome Location
Chr.6: 28526012-28534951 on Build GRCh38
Transcripts
NM_001509.2 NM_003996.3

SNP Detail

SNP ID Chromosome Location
rs887875377 Chr.6: 28534640-28534640 on Build GRCh38
rs763827016 Chr.6: 28534925-28534925 on Build GRCh38
rs1046534206 Chr.6: 28534626-28534626 on Build GRCh38
rs1027594505 Chr.6: 28534835-28534835 on Build GRCh38
rs567898558 Chr.6: 28534990-28534990 on Build GRCh38
rs537517475 Chr.6: 28534968-28534968 on Build GRCh38
rs451774 Chr.6: 28534773-28534773 on Build GRCh38
rs552108652 Chr.6: 28534878-28534878 on Build GRCh38
rs929278750 Chr.6: 28534951-28534951 on Build GRCh38
rs765735176 Chr.6: 28534823-28534823 on Build GRCh38
rs568873549 Chr.6: 28534950-28534950 on Build GRCh38
rs771545001 Chr.6: 28534689-28534689 on Build GRCh38
rs532012150 Chr.6: 28534874-28534874 on Build GRCh38
rs951788298 Chr.6: 28534839-28534839 on Build GRCh38
rs114169464 Chr.6: 28534781-28534781 on Build GRCh38
rs1055290766 Chr.6: 28534926-28534926 on Build GRCh38
rs906713271 Chr.6: 28534636-28534636 on Build GRCh38
rs116070091 Chr.6: 28534737-28534737 on Build GRCh38
rs28382613 Chr.6: 28534795-28534795 on Build GRCh38
rs910752661 Chr.6: 28534880-28534880 on Build GRCh38
rs938230645 Chr.6: 28534638-28534638 on Build GRCh38
rs545532101 Chr.6: 28534756-28534756 on Build GRCh38
rs996089947 Chr.6: 28534819-28534819 on Build GRCh38
rs919301908 Chr.6: 28534584-28534584 on Build GRCh38
rs1055344414 Chr.6: 28534655-28534655 on Build GRCh38
rs893790834 Chr.6: 28534685-28534685 on Build GRCh38
rs9380078 Chr.6: 28534840-28534840 on Build GRCh38
rs928128497 Chr.6: 28534888-28534888 on Build GRCh38
rs566504938 Chr.6: 28534864-28534864 on Build GRCh38
rs983586894 Chr.6: 28534582-28534582 on Build GRCh38
rs561441227 Chr.6: 28534803-28534803 on Build GRCh38
rs28382614 Chr.6: 28534980-28534980 on Build GRCh38
rs1049068998 Chr.6: 28534962-28534962 on Build GRCh38
rs28382612 Chr.6: 28534599-28534599 on Build GRCh38
rs938206021 Chr.6: 28534911-28534911 on Build GRCh38
rs865965252 Chr.6: 28534635-28534635 on Build GRCh38
rs1044913347 Chr.6: 28534693-28534693 on Build GRCh38
rs1040518922 Chr.6: 28534982-28534982 on Build GRCh38
rs1018758320 Chr.6: 28534788-28534788 on Build GRCh38
rs553365961 Chr.6: 28534725-28534725 on Build GRCh38
rs1042796013 Chr.6: 28534757-28534757 on Build GRCh38
rs1042354128 Chr.6: 28534940-28534940 on Build GRCh38
rs540113725 Chr.6: 28534711-28534711 on Build GRCh38
rs1005317693 Chr.6: 28534673-28534673 on Build GRCh38
rs574412536 Chr.6: 28534622-28534622 on Build GRCh38
rs915465916 Chr.6: 28534929-28534929 on Build GRCh38
rs541472197 Chr.6: 28534860-28534860 on Build GRCh38
rs946978720 Chr.6: 28534939-28534939 on Build GRCh38
rs530448042 Chr.6: 28534859-28534859 on Build GRCh38
rs546199182 Chr.6: 28534877-28534877 on Build GRCh38
rs948840656 Chr.6: 28534985-28534985 on Build GRCh38
rs752945175 Chr.6: 28534849-28534849 on Build GRCh38
rs554306870 Chr.6: 28534592-28534592 on Build GRCh38

View Full Product Details