Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 29307408-29307918 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
OR14J1
Entrez Gene ID
442191
Gene Name
olfactory receptor family 14 subfamily J member 1
Gene Aliases
OR5U1, OR6-25, bA150A6.4, hs6M1-28
UniGene
Hs.632004
Gene Chromosome Location
Chr.6: 29306690-29307655 on Build GRCh38
Transcripts
NM_030946.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1286798 c.768G>T p.E256D Chr.6: 29307457-29307457 on Build GRCh38
4561179 c.873G>A p.R291R Chr.6: 29307562-29307562 on Build GRCh38
3250405 c.925C>T p.P309S Chr.6: 29307614-29307614 on Build GRCh38
3861245 c.948A>G p.K316K Chr.6: 29307637-29307637 on Build GRCh38
3250401 c.797C>T p.S266L Chr.6: 29307486-29307486 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs142403297 Chr.6: 29307598-29307598 on Build GRCh38
rs748835504 Chr.6: 29307460-29307460 on Build GRCh38
rs756135978 Chr.6: 29307537-29307537 on Build GRCh38
rs769847834 Chr.6: 29307680-29307680 on Build GRCh38
rs778352534 Chr.6: 29307653-29307653 on Build GRCh38
rs151244120 Chr.6: 29307639-29307639 on Build GRCh38
rs776530256 Chr.6: 29307572-29307572 on Build GRCh38
rs747730629 Chr.6: 29307655-29307655 on Build GRCh38
rs9257696 Chr.6: 29307730-29307730 on Build GRCh38
rs548059514 Chr.6: 29307508-29307508 on Build GRCh38
rs149080595 Chr.6: 29307824-29307824 on Build GRCh38
rs774540146 Chr.6: 29307471-29307471 on Build GRCh38
rs182078656 Chr.6: 29307867-29307867 on Build GRCh38
rs780011406 Chr.6: 29307618-29307618 on Build GRCh38
rs764442309 Chr.6: 29307687-29307687 on Build GRCh38
rs753963089 Chr.6: 29307505-29307505 on Build GRCh38
rs765524812 Chr.6: 29307509-29307509 on Build GRCh38
rs768091172 Chr.6: 29307559-29307559 on Build GRCh38
rs959944025 Chr.6: 29307546-29307546 on Build GRCh38
rs536363434 Chr.6: 29307544-29307544 on Build GRCh38
rs779663166 Chr.6: 29307540-29307540 on Build GRCh38
rs767773206 Chr.6: 29307610-29307610 on Build GRCh38
rs960300165 Chr.6: 29307746-29307746 on Build GRCh38
rs752914713 Chr.6: 29307513-29307513 on Build GRCh38
rs761109650 Chr.6: 29307567-29307567 on Build GRCh38
rs771322172 Chr.6: 29307665-29307665 on Build GRCh38
rs745734240 Chr.6: 29307531-29307531 on Build GRCh38
rs781203455 Chr.6: 29307527-29307527 on Build GRCh38
rs17404424 Chr.6: 29307521-29307521 on Build GRCh38
rs762205231 Chr.6: 29307480-29307480 on Build GRCh38
rs756075788 Chr.6: 29307614-29307614 on Build GRCh38
rs1019951901 Chr.6: 29307790-29307790 on Build GRCh38
rs766145245 Chr.6: 29307703-29307703 on Build GRCh38
rs978466959 Chr.6: 29307882-29307882 on Build GRCh38
rs142365974 Chr.6: 29307603-29307603 on Build GRCh38
rs61735224 Chr.6: 29307451-29307451 on Build GRCh38
rs752962130 Chr.6: 29307596-29307596 on Build GRCh38
rs988493892 Chr.6: 29307548-29307548 on Build GRCh38
rs532095657 Chr.6: 29307432-29307432 on Build GRCh38
rs144651776 Chr.6: 29307483-29307483 on Build GRCh38
rs772419726 Chr.6: 29307562-29307562 on Build GRCh38
rs374669047 Chr.6: 29307663-29307663 on Build GRCh38
rs9257695 Chr.6: 29307722-29307722 on Build GRCh38
rs905991447 Chr.6: 29307718-29307718 on Build GRCh38
rs139773321 Chr.6: 29307486-29307486 on Build GRCh38
rs538290613 Chr.6: 29307817-29307817 on Build GRCh38
rs1045629012 Chr.6: 29307879-29307879 on Build GRCh38
rs538397769 Chr.6: 29307621-29307621 on Build GRCh38
rs574945870 Chr.6: 29307804-29307804 on Build GRCh38
rs186022893 Chr.6: 29307885-29307885 on Build GRCh38
rs948194645 Chr.6: 29307470-29307470 on Build GRCh38
rs199859435 Chr.6: 29307696-29307696 on Build GRCh38
rs912924954 Chr.6: 29307552-29307552 on Build GRCh38
rs753807383 Chr.6: 29307706-29307706 on Build GRCh38
rs371650943 Chr.6: 29307642-29307642 on Build GRCh38
rs769426724 Chr.6: 29307450-29307450 on Build GRCh38
rs149908924 Chr.6: 29307560-29307560 on Build GRCh38
rs143752928 Chr.6: 29307850-29307850 on Build GRCh38
rs778060025 Chr.6: 29307661-29307661 on Build GRCh38
rs750268499 Chr.6: 29307611-29307611 on Build GRCh38
rs766889482 Chr.6: 29307569-29307569 on Build GRCh38
rs760523725 Chr.6: 29307699-29307699 on Build GRCh38
rs763220744 Chr.6: 29307685-29307685 on Build GRCh38
rs1016374277 Chr.6: 29307458-29307466 on Build GRCh38
rs777186078 Chr.6: 29307675-29307675 on Build GRCh38
rs773567399 Chr.6: 29307563-29307563 on Build GRCh38
rs759807793 Chr.6: 29307560-29307561 on Build GRCh38
rs554877975 Chr.6: 29307799-29307799 on Build GRCh38
rs538609796 Chr.6: 29307684-29307684 on Build GRCh38
rs1010519846 Chr.6: 29307785-29307785 on Build GRCh38
rs765295589 Chr.6: 29307581-29307581 on Build GRCh38
rs985255833 Chr.6: 29307604-29307604 on Build GRCh38
rs1474859 Chr.6: 29307808-29307808 on Build GRCh38
rs891481677 Chr.6: 29307549-29307549 on Build GRCh38
rs749231922 Chr.6: 29307627-29307627 on Build GRCh38
rs760032007 Chr.6: 29307676-29307676 on Build GRCh38
rs768377065 Chr.6: 29307461-29307461 on Build GRCh38
rs531580647 Chr.6: 29307487-29307487 on Build GRCh38
rs1037830113 Chr.6: 29307712-29307712 on Build GRCh38
rs147619517 Chr.6: 29307561-29307561 on Build GRCh38
rs766375996 Chr.6: 29307504-29307504 on Build GRCh38
rs771722883 Chr.6: 29307660-29307660 on Build GRCh38
rs942553604 Chr.6: 29307566-29307566 on Build GRCh38
rs761236221 Chr.6: 29307503-29307503 on Build GRCh38
rs567871531 Chr.6: 29307579-29307579 on Build GRCh38
rs765149395 Chr.6: 29307726-29307726 on Build GRCh38

View Full Product Details