Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 30158319-30158828 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TRIM10
Entrez Gene ID
10107
Gene Name
tripartite motif containing 10
Gene Aliases
HERF1, RFB30, RNF9
UniGene
Hs.709483
Gene Chromosome Location
Chr.6: 30151939-30163216 on Build GRCh38
Transcripts
NM_006778.3 NM_052828.2 XM_011514221.1 XM_011514222.2 XM_011514223.2 XM_011514225.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3251099 c.567C>T p.F189F Chr.6: 30158588-30158588 on Build GRCh38
7199921 c.529C>G p.Q177E Chr.6: 30158626-30158626 on Build GRCh38
5870019 c.682delG p.E228fs*24 Chr.6: 30158473-30158473 on Build GRCh38
5692985 c.592G>T p.E198* Chr.6: 30158563-30158563 on Build GRCh38
3410898 c.692G>A p.R231Q Chr.6: 30158463-30158463 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1023830974 Chr.6: 30158605-30158605 on Build GRCh38
rs949450579 Chr.6: 30158437-30158437 on Build GRCh38
rs767722466 Chr.6: 30158432-30158432 on Build GRCh38
rs116497639 Chr.6: 30158760-30158760 on Build GRCh38
rs111803166 Chr.6: 30158448-30158448 on Build GRCh38
rs897848556 Chr.6: 30158777-30158777 on Build GRCh38
rs746604908 Chr.6: 30158500-30158500 on Build GRCh38
rs538896069 Chr.6: 30158558-30158558 on Build GRCh38
rs1054042653 Chr.6: 30158791-30158791 on Build GRCh38
rs374424680 Chr.6: 30158372-30158372 on Build GRCh38
rs866632574 Chr.6: 30158434-30158434 on Build GRCh38
rs745554277 Chr.6: 30158473-30158473 on Build GRCh38
rs374299419 Chr.6: 30158635-30158635 on Build GRCh38
rs575887974 Chr.6: 30158719-30158719 on Build GRCh38
rs749526932 Chr.6: 30158491-30158491 on Build GRCh38
rs765008695 Chr.6: 30158555-30158555 on Build GRCh38
rs760743692 Chr.6: 30158665-30158665 on Build GRCh38
rs377070529 Chr.6: 30158375-30158375 on Build GRCh38
rs762183688 Chr.6: 30158651-30158651 on Build GRCh38
rs760939998 Chr.6: 30158548-30158548 on Build GRCh38
rs765727213 Chr.6: 30158410-30158410 on Build GRCh38
rs560494648 Chr.6: 30158355-30158355 on Build GRCh38
rs777714494 Chr.6: 30158576-30158576 on Build GRCh38
rs746467216 Chr.6: 30158466-30158466 on Build GRCh38
rs979051126 Chr.6: 30158394-30158394 on Build GRCh38
rs766650120 Chr.6: 30158666-30158666 on Build GRCh38
rs772212519 Chr.6: 30158655-30158655 on Build GRCh38
rs763921171 Chr.6: 30158645-30158645 on Build GRCh38
rs751729897 Chr.6: 30158428-30158428 on Build GRCh38
rs138874445 Chr.6: 30158610-30158610 on Build GRCh38
rs114405037 Chr.6: 30158647-30158647 on Build GRCh38
rs548134704 Chr.6: 30158490-30158490 on Build GRCh38
rs776108850 Chr.6: 30158420-30158420 on Build GRCh38
rs61737427 Chr.6: 30158526-30158526 on Build GRCh38
rs757289413 Chr.6: 30158431-30158431 on Build GRCh38
rs1030633584 Chr.6: 30158636-30158636 on Build GRCh38
rs770568149 Chr.6: 30158639-30158639 on Build GRCh38
rs138056642 Chr.6: 30158503-30158503 on Build GRCh38
rs902340625 Chr.6: 30158456-30158456 on Build GRCh38
rs979102026 Chr.6: 30158627-30158627 on Build GRCh38
rs538429585 Chr.6: 30158732-30158732 on Build GRCh38
rs750667956 Chr.6: 30158441-30158441 on Build GRCh38
rs761905357 Chr.6: 30158536-30158536 on Build GRCh38
rs1036354906 Chr.6: 30158478-30158478 on Build GRCh38
rs763603651 Chr.6: 30158421-30158421 on Build GRCh38
rs746633780 Chr.6: 30158397-30158397 on Build GRCh38
rs773214911 Chr.6: 30158661-30158661 on Build GRCh38
rs148067817 Chr.6: 30158759-30158759 on Build GRCh38
rs868721021 Chr.6: 30158443-30158443 on Build GRCh38
rs761800101 Chr.6: 30158758-30158758 on Build GRCh38
rs754730380 Chr.6: 30158447-30158447 on Build GRCh38
rs995829243 Chr.6: 30158484-30158484 on Build GRCh38
rs113447735 Chr.6: 30158609-30158609 on Build GRCh38
rs764068515 Chr.6: 30158423-30158423 on Build GRCh38
rs931829655 Chr.6: 30158721-30158721 on Build GRCh38
rs751423482 Chr.6: 30158588-30158588 on Build GRCh38
rs769504238 Chr.6: 30158639-30158639 on Build GRCh38
rs181857093 Chr.6: 30158764-30158764 on Build GRCh38
rs559448550 Chr.6: 30158416-30158422 on Build GRCh38
rs568128993 Chr.6: 30158499-30158499 on Build GRCh38
rs758138844 Chr.6: 30158560-30158560 on Build GRCh38
rs767528469 Chr.6: 30158546-30158546 on Build GRCh38
rs1040249610 Chr.6: 30158425-30158425 on Build GRCh38
rs531527319 Chr.6: 30158452-30158452 on Build GRCh38
rs763550880 Chr.6: 30158507-30158507 on Build GRCh38
rs769895582 Chr.6: 30158463-30158463 on Build GRCh38
rs923197434 Chr.6: 30158718-30158718 on Build GRCh38
rs770546848 Chr.6: 30158399-30158399 on Build GRCh38
rs779270305 Chr.6: 30158427-30158429 on Build GRCh38
rs757557280 Chr.6: 30158351-30158351 on Build GRCh38
rs61735038 Chr.6: 30158400-30158400 on Build GRCh38
rs942048269 Chr.6: 30158474-30158474 on Build GRCh38
rs936505796 Chr.6: 30158781-30158781 on Build GRCh38
rs780856379 Chr.6: 30158470-30158470 on Build GRCh38
rs755329558 Chr.6: 30158421-30158422 on Build GRCh38
rs774960542 Chr.6: 30158649-30158649 on Build GRCh38
rs150423865 Chr.6: 30158709-30158709 on Build GRCh38
rs985978837 Chr.6: 30158722-30158722 on Build GRCh38
rs780802203 Chr.6: 30158615-30158615 on Build GRCh38
rs773315163 Chr.6: 30158486-30158486 on Build GRCh38
rs749046875 Chr.6: 30158370-30158370 on Build GRCh38
rs138647866 Chr.6: 30158385-30158385 on Build GRCh38
rs1013073467 Chr.6: 30158563-30158563 on Build GRCh38
rs777572853 Chr.6: 30158464-30158464 on Build GRCh38
rs61737424 Chr.6: 30158508-30158508 on Build GRCh38
rs779847481 Chr.6: 30158644-30158644 on Build GRCh38
rs750615347 Chr.6: 30158547-30158547 on Build GRCh38

View Full Product Details