Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 986886-987375 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C7orf50
Entrez Gene ID
84310
Gene Name
chromosome 7 open reading frame 50
Gene Aliases
YCR016W
UniGene
Hs.653258
Gene Chromosome Location
Chr.7: 978485-1138291 on Build GRCh38
Transcripts
XM_011515580.2 XM_011515581.2 XM_011515582.2 XM_011515583.2 XM_011515584.2 XM_017012719.1 XM_017012720.1 XM_017012721.1 NM_017781.2 XM_011515440.2 XM_011515441.2

Gene Symbol
CYP2W1
Entrez Gene ID
54905
Gene Name
cytochrome P450 family 2 subfamily W member 1
Gene Aliases
-
UniGene
Hs.272795
Gene Chromosome Location
Chr.7: 983157-989640 on Build GRCh38
Transcripts
XM_011515580.2 XM_011515581.2 XM_011515582.2 XM_011515583.2 XM_011515584.2 XM_017012719.1 XM_017012720.1 XM_017012721.1 NM_017781.2 XM_011515440.2 XM_011515441.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4675973 c.962G>A p.R321Q Chr.7: 987350-987350 on Build GRCh38
5497874 c.911C>T p.T304M Chr.7: 987198-987198 on Build GRCh38
7281216 c.952G>A p.V318M Chr.7: 987239-987239 on Build GRCh38
598353 c.982C>T p.R328C Chr.7: 987370-987370 on Build GRCh38
4632409 c.894G>A p.T298T Chr.7: 987181-987181 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs548719233 Chr.7: 987145-987145 on Build GRCh38
rs890960571 Chr.7: 987243-987243 on Build GRCh38
rs746735643 Chr.7: 987349-987349 on Build GRCh38
rs1017730393 Chr.7: 987022-987022 on Build GRCh38
rs747016601 Chr.7: 987118-987118 on Build GRCh38
rs556515220 Chr.7: 987143-987143 on Build GRCh38
rs776149252 Chr.7: 987293-987293 on Build GRCh38
rs888026417 Chr.7: 987187-987187 on Build GRCh38
rs376749499 Chr.7: 987176-987176 on Build GRCh38
rs879457768 Chr.7: 987331-987333 on Build GRCh38
rs562495627 Chr.7: 987269-987269 on Build GRCh38
rs371388116 Chr.7: 987268-987268 on Build GRCh38
rs770573282 Chr.7: 987291-987291 on Build GRCh38
rs758835138 Chr.7: 987163-987163 on Build GRCh38
rs759025327 Chr.7: 987299-987299 on Build GRCh38
rs753155574 Chr.7: 987239-987239 on Build GRCh38
rs999211491 Chr.7: 986955-986955 on Build GRCh38
rs139443941 Chr.7: 986910-986910 on Build GRCh38
rs777719372 Chr.7: 987348-987348 on Build GRCh38
rs752013339 Chr.7: 987256-987256 on Build GRCh38
rs748510204 Chr.7: 987097-987097 on Build GRCh38
rs138410337 Chr.7: 987180-987180 on Build GRCh38
rs751928696 Chr.7: 987325-987325 on Build GRCh38
rs113795030 Chr.7: 987262-987262 on Build GRCh38
rs79364415 Chr.7: 987132-987132 on Build GRCh38
rs572801095 Chr.7: 987230-987230 on Build GRCh38
rs749247516 Chr.7: 987123-987123 on Build GRCh38
rs764550796 Chr.7: 987255-987255 on Build GRCh38
rs527453749 Chr.7: 987114-987114 on Build GRCh38
rs995611991 Chr.7: 987069-987069 on Build GRCh38
rs924312571 Chr.7: 986920-986920 on Build GRCh38
rs768589576 Chr.7: 987194-987194 on Build GRCh38
rs917877689 Chr.7: 987227-987227 on Build GRCh38
rs921942671 Chr.7: 987304-987304 on Build GRCh38
rs535590454 Chr.7: 987125-987125 on Build GRCh38
rs554900839 Chr.7: 987258-987258 on Build GRCh38
rs58781963 Chr.7: 987199-987199 on Build GRCh38
rs528996615 Chr.7: 986963-986963 on Build GRCh38
rs781292506 Chr.7: 987259-987259 on Build GRCh38
rs376141354 Chr.7: 987235-987235 on Build GRCh38
rs955178909 Chr.7: 987062-987062 on Build GRCh38
rs370647090 Chr.7: 987338-987338 on Build GRCh38
rs772856571 Chr.7: 987139-987139 on Build GRCh38
rs887794682 Chr.7: 987012-987012 on Build GRCh38
rs766335746 Chr.7: 987335-987335 on Build GRCh38
rs758281340 Chr.7: 987346-987346 on Build GRCh38
rs769170780 Chr.7: 987300-987300 on Build GRCh38
rs760375957 Chr.7: 987142-987142 on Build GRCh38
rs745500143 Chr.7: 987352-987352 on Build GRCh38
rs745988107 Chr.7: 987193-987193 on Build GRCh38
rs755777655 Chr.7: 986954-986954 on Build GRCh38
rs774085257 Chr.7: 987065-987065 on Build GRCh38
rs975791793 Chr.7: 986931-986931 on Build GRCh38
rs748033735 Chr.7: 987070-987070 on Build GRCh38
rs369508693 Chr.7: 987121-987121 on Build GRCh38
rs777521268 Chr.7: 987286-987286 on Build GRCh38
rs763355596 Chr.7: 987313-987313 on Build GRCh38
rs997064387 Chr.7: 987270-987270 on Build GRCh38
rs766805858 Chr.7: 987083-987083 on Build GRCh38
rs975594070 Chr.7: 987175-987175 on Build GRCh38
rs371343381 Chr.7: 987072-987072 on Build GRCh38
rs531160646 Chr.7: 987329-987329 on Build GRCh38
rs948899375 Chr.7: 986965-986965 on Build GRCh38
rs551440848 Chr.7: 987198-987198 on Build GRCh38
rs1045152203 Chr.7: 987041-987041 on Build GRCh38
rs778081695 Chr.7: 987170-987170 on Build GRCh38
rs746827801 Chr.7: 987287-987287 on Build GRCh38
rs562371552 Chr.7: 986988-986988 on Build GRCh38
rs372963929 Chr.7: 987211-987211 on Build GRCh38
rs935630964 Chr.7: 986926-986926 on Build GRCh38
rs900579328 Chr.7: 987030-987030 on Build GRCh38
rs774174678 Chr.7: 987127-987127 on Build GRCh38
rs1006597686 Chr.7: 987016-987016 on Build GRCh38
rs767712332 Chr.7: 987328-987328 on Build GRCh38
rs1039425012 Chr.7: 987212-987212 on Build GRCh38
rs542593170 Chr.7: 986978-986978 on Build GRCh38
rs987766289 Chr.7: 987314-987314 on Build GRCh38
rs571210763 Chr.7: 987108-987108 on Build GRCh38
rs761535713 Chr.7: 987128-987128 on Build GRCh38
rs763609907 Chr.7: 987240-987240 on Build GRCh38
rs950953455 Chr.7: 987271-987271 on Build GRCh38
rs61420495 Chr.7: 987263-987263 on Build GRCh38
rs376302659 Chr.7: 987303-987303 on Build GRCh38
rs755501387 Chr.7: 987094-987094 on Build GRCh38
rs748072164 Chr.7: 987265-987265 on Build GRCh38
rs112286741 Chr.7: 987290-987290 on Build GRCh38
rs749887300 Chr.7: 987079-987079 on Build GRCh38
rs377155029 Chr.7: 987082-987082 on Build GRCh38
rs935210499 Chr.7: 986983-986983 on Build GRCh38
rs562466858 Chr.7: 986977-986977 on Build GRCh38
rs965542629 Chr.7: 986914-986914 on Build GRCh38
rs760287935 Chr.7: 987228-987228 on Build GRCh38
rs750476691 Chr.7: 987260-987260 on Build GRCh38
rs370828451 Chr.7: 987099-987099 on Build GRCh38
rs757209232 Chr.7: 987245-987262 on Build GRCh38
rs762300475 Chr.7: 987327-987327 on Build GRCh38
rs117425670 Chr.7: 987011-987011 on Build GRCh38
rs757013139 Chr.7: 987350-987350 on Build GRCh38
rs753062234 Chr.7: 987158-987158 on Build GRCh38
rs765681396 Chr.7: 987154-987154 on Build GRCh38
rs536373592 Chr.7: 987138-987138 on Build GRCh38
rs772035855 Chr.7: 987266-987266 on Build GRCh38
rs896305453 Chr.7: 986998-986998 on Build GRCh38
rs781305530 Chr.7: 987186-987186 on Build GRCh38
rs547288712 Chr.7: 987119-987119 on Build GRCh38
rs775760760 Chr.7: 987273-987273 on Build GRCh38
rs773055759 Chr.7: 987222-987222 on Build GRCh38
rs574569786 Chr.7: 987096-987096 on Build GRCh38
rs968936198 Chr.7: 987295-987295 on Build GRCh38
rs186092597 Chr.7: 987234-987234 on Build GRCh38
rs551219950 Chr.7: 987049-987049 on Build GRCh38
rs768522467 Chr.7: 987040-987040 on Build GRCh38
rs770999220 Chr.7: 987110-987110 on Build GRCh38
rs373924223 Chr.7: 987144-987144 on Build GRCh38
rs988047184 Chr.7: 987215-987215 on Build GRCh38
rs763526504 Chr.7: 987310-987310 on Build GRCh38
rs754355918 Chr.7: 987085-987085 on Build GRCh38

View Full Product Details