Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 5064767-5065274 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RBAK
Entrez Gene ID
57786
Gene Name
RB associated KRAB zinc finger
Gene Aliases
ZNF769
UniGene
Hs.396178
Gene Chromosome Location
Chr.7: 5045821-5069488 on Build GRCh38
Transcripts
NM_001204456.1 NM_021163.3 NM_001204513.1

Gene Symbol
RBAK-RBAKDN
Entrez Gene ID
100533952
Gene Name
RBAK-RBAKDN readthrough
Gene Aliases
RBAK-LOC389458
UniGene
Hs.396178
Gene Chromosome Location
Chr.7: 5045821-5073223 on Build GRCh38
Transcripts
NM_001204456.1 NM_021163.3 NM_001204513.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
746554 c.1648G>A p.E550K Chr.7: 5065104-5065104 on Build GRCh38
6478923 c.1543G>C p.E515Q Chr.7: 5064999-5064999 on Build GRCh38
7242557 c.1421G>C p.R474T Chr.7: 5064877-5064877 on Build GRCh38
5602189 c.1635A>C p.G545G Chr.7: 5065091-5065091 on Build GRCh38
1090682 c.1552A>G p.K518E Chr.7: 5065008-5065008 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759441266 Chr.7: 5065220-5065224 on Build GRCh38
rs760043040 Chr.7: 5065104-5065104 on Build GRCh38
rs199539533 Chr.7: 5065042-5065042 on Build GRCh38
rs752330581 Chr.7: 5064985-5064985 on Build GRCh38
rs773458820 Chr.7: 5065033-5065033 on Build GRCh38
rs778475217 Chr.7: 5064855-5064855 on Build GRCh38
rs748319662 Chr.7: 5065007-5065007 on Build GRCh38
rs755562530 Chr.7: 5064832-5064832 on Build GRCh38
rs373062673 Chr.7: 5064958-5064958 on Build GRCh38
rs763245866 Chr.7: 5064803-5064803 on Build GRCh38
rs773271464 Chr.7: 5064869-5064869 on Build GRCh38
rs746906376 Chr.7: 5064939-5064939 on Build GRCh38
rs779541981 Chr.7: 5064838-5064838 on Build GRCh38
rs755976667 Chr.7: 5065160-5065160 on Build GRCh38
rs539259619 Chr.7: 5064921-5064921 on Build GRCh38
rs565727836 Chr.7: 5064917-5064917 on Build GRCh38
rs751977303 Chr.7: 5064813-5064813 on Build GRCh38
rs752912140 Chr.7: 5064799-5064799 on Build GRCh38
rs776519644 Chr.7: 5065218-5065218 on Build GRCh38
rs201713068 Chr.7: 5065144-5065144 on Build GRCh38
rs773587795 Chr.7: 5064960-5064960 on Build GRCh38
rs771262238 Chr.7: 5065097-5065097 on Build GRCh38
rs11771682 Chr.7: 5065202-5065202 on Build GRCh38
rs777136789 Chr.7: 5065035-5065035 on Build GRCh38
rs761587290 Chr.7: 5065130-5065130 on Build GRCh38
rs756026179 Chr.7: 5065055-5065055 on Build GRCh38
rs781530374 Chr.7: 5065185-5065185 on Build GRCh38
rs770780647 Chr.7: 5065213-5065214 on Build GRCh38
rs750861797 Chr.7: 5064902-5064902 on Build GRCh38
rs779846594 Chr.7: 5065166-5065166 on Build GRCh38
rs746796795 Chr.7: 5065091-5065092 on Build GRCh38
rs193921102 Chr.7: 5065058-5065058 on Build GRCh38
rs1055563893 Chr.7: 5065028-5065028 on Build GRCh38
rs770260621 Chr.7: 5065198-5065198 on Build GRCh38
rs781771231 Chr.7: 5065117-5065117 on Build GRCh38
rs764329000 Chr.7: 5064804-5064804 on Build GRCh38
rs867631797 Chr.7: 5065071-5065071 on Build GRCh38
rs771827540 Chr.7: 5064857-5064857 on Build GRCh38
rs757445798 Chr.7: 5065182-5065182 on Build GRCh38
rs747242155 Chr.7: 5065092-5065092 on Build GRCh38
rs779122946 Chr.7: 5065025-5065025 on Build GRCh38
rs148394535 Chr.7: 5065049-5065049 on Build GRCh38
rs868618303 Chr.7: 5065188-5065188 on Build GRCh38
rs147190676 Chr.7: 5064872-5064872 on Build GRCh38
rs377198438 Chr.7: 5064999-5064999 on Build GRCh38
rs1039140345 Chr.7: 5064893-5064893 on Build GRCh38
rs769932705 Chr.7: 5064882-5064882 on Build GRCh38
rs866986149 Chr.7: 5065017-5065017 on Build GRCh38
rs549061122 Chr.7: 5064953-5064953 on Build GRCh38
rs770182530 Chr.7: 5065109-5065109 on Build GRCh38
rs762235727 Chr.7: 5064896-5064896 on Build GRCh38
rs747810750 Chr.7: 5064931-5064931 on Build GRCh38
rs749357079 Chr.7: 5064948-5064948 on Build GRCh38
rs535079682 Chr.7: 5064851-5064851 on Build GRCh38
rs768216955 Chr.7: 5065229-5065229 on Build GRCh38
rs780088006 Chr.7: 5065056-5065056 on Build GRCh38
rs903213369 Chr.7: 5065063-5065063 on Build GRCh38
rs764356759 Chr.7: 5064887-5064887 on Build GRCh38
rs767042267 Chr.7: 5065238-5065238 on Build GRCh38
rs774018944 Chr.7: 5065230-5065230 on Build GRCh38
rs753827279 Chr.7: 5065167-5065167 on Build GRCh38
rs761944320 Chr.7: 5064955-5064955 on Build GRCh38
rs757611790 Chr.7: 5064822-5064822 on Build GRCh38
rs745853477 Chr.7: 5065003-5065003 on Build GRCh38
rs145473784 Chr.7: 5064962-5064962 on Build GRCh38
rs868199292 Chr.7: 5064991-5064991 on Build GRCh38
rs369502461 Chr.7: 5064946-5064946 on Build GRCh38
rs777955109 Chr.7: 5065087-5065087 on Build GRCh38
rs772203103 Chr.7: 5065079-5065079 on Build GRCh38
rs772979673 Chr.7: 5065242-5065242 on Build GRCh38
rs768712393 Chr.7: 5065004-5065004 on Build GRCh38
rs374417308 Chr.7: 5064845-5064845 on Build GRCh38
rs778890045 Chr.7: 5065006-5065006 on Build GRCh38
rs781186316 Chr.7: 5065002-5065002 on Build GRCh38
rs761432734 Chr.7: 5065236-5065236 on Build GRCh38
rs370383120 Chr.7: 5065218-5065218 on Build GRCh38
rs772258734 Chr.7: 5065020-5065020 on Build GRCh38
rs758159090 Chr.7: 5064988-5064988 on Build GRCh38
rs754495200 Chr.7: 5064973-5064973 on Build GRCh38
rs781643476 Chr.7: 5064823-5064823 on Build GRCh38
rs774795748 Chr.7: 5065240-5065240 on Build GRCh38
rs777718869 Chr.7: 5064937-5064937 on Build GRCh38
rs143678050 Chr.7: 5064856-5064856 on Build GRCh38
rs140646162 Chr.7: 5064883-5064883 on Build GRCh38
rs776894660 Chr.7: 5065098-5065098 on Build GRCh38
rs754366722 Chr.7: 5064969-5064969 on Build GRCh38
rs774478324 Chr.7: 5064889-5064889 on Build GRCh38
rs149739845 Chr.7: 5064954-5064954 on Build GRCh38
rs374791225 Chr.7: 5065054-5065054 on Build GRCh38
rs776129102 Chr.7: 5065201-5065201 on Build GRCh38
rs750161506 Chr.7: 5065153-5065153 on Build GRCh38
rs756869935 Chr.7: 5065211-5065211 on Build GRCh38
rs766836301 Chr.7: 5064966-5064966 on Build GRCh38
rs764768527 Chr.7: 5065037-5065037 on Build GRCh38
rs764670112 Chr.7: 5064984-5064984 on Build GRCh38
rs762536253 Chr.7: 5065246-5065261 on Build GRCh38
rs941049273 Chr.7: 5064795-5064795 on Build GRCh38
rs763101971 Chr.7: 5065178-5065178 on Build GRCh38
rs543771074 Chr.7: 5065077-5065077 on Build GRCh38
rs990654053 Chr.7: 5065005-5065005 on Build GRCh38
rs746192169 Chr.7: 5065187-5065187 on Build GRCh38
rs759968367 Chr.7: 5065036-5065036 on Build GRCh38
rs765413482 Chr.7: 5064824-5064824 on Build GRCh38
rs1034937947 Chr.7: 5064995-5064995 on Build GRCh38
rs758212171 Chr.7: 5064936-5064936 on Build GRCh38
rs762548190 Chr.7: 5065111-5065111 on Build GRCh38
rs377673260 Chr.7: 5064944-5064944 on Build GRCh38
rs746779347 Chr.7: 5064996-5064996 on Build GRCh38
rs761019553 Chr.7: 5065034-5065034 on Build GRCh38
rs555675840 Chr.7: 5065180-5065180 on Build GRCh38
rs767879426 Chr.7: 5064900-5064900 on Build GRCh38
rs142569406 Chr.7: 5065135-5065135 on Build GRCh38
rs758412354 Chr.7: 5065174-5065174 on Build GRCh38
rs752174757 Chr.7: 5065040-5065040 on Build GRCh38
rs1048483348 Chr.7: 5065127-5065127 on Build GRCh38
rs763212100 Chr.7: 5064825-5064825 on Build GRCh38
rs780864363 Chr.7: 5065050-5065050 on Build GRCh38
rs562837535 Chr.7: 5064910-5064910 on Build GRCh38
rs918268185 Chr.7: 5065083-5065083 on Build GRCh38
rs753362715 Chr.7: 5064916-5064916 on Build GRCh38
rs769800716 Chr.7: 5065228-5065236 on Build GRCh38
rs751325675 Chr.7: 5065044-5065044 on Build GRCh38
rs751201147 Chr.7: 5065120-5065120 on Build GRCh38

View Full Product Details