Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 5290888-5291395 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC29A4
Entrez Gene ID
222962
Gene Name
solute carrier family 29 member 4
Gene Aliases
ENT4, PMAT
UniGene
Hs.4302
Gene Chromosome Location
Chr.7: 5274311-5304073 on Build GRCh38
Transcripts
NM_001040661.1 NM_001300847.1 NM_153247.2 XM_005249658.4 XM_006715667.3 XM_006715668.2 XM_011515200.2 XM_011515201.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6407166 c.394C>A p.L132M Chr.7: 5291216-5291216 on Build GRCh38
5006134 c.315G>A p.V105V Chr.7: 5291137-5291137 on Build GRCh38
3881728 c.411C>T p.T137T Chr.7: 5291233-5291233 on Build GRCh38
3265991 c.392C>A p.T131N Chr.7: 5291214-5291214 on Build GRCh38
7169156 c.366G>A p.L122L Chr.7: 5291188-5291188 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759271014 Chr.7: 5291283-5291283 on Build GRCh38
rs781585479 Chr.7: 5291109-5291109 on Build GRCh38
rs148426706 Chr.7: 5291234-5291234 on Build GRCh38
rs767110362 Chr.7: 5291137-5291137 on Build GRCh38
rs1011431175 Chr.7: 5290955-5290955 on Build GRCh38
rs371054725 Chr.7: 5291082-5291082 on Build GRCh38
rs761275194 Chr.7: 5291206-5291206 on Build GRCh38
rs971029407 Chr.7: 5291317-5291317 on Build GRCh38
rs536440464 Chr.7: 5291306-5291306 on Build GRCh38
rs772658007 Chr.7: 5291055-5291055 on Build GRCh38
rs952833635 Chr.7: 5291188-5291188 on Build GRCh38
rs964565289 Chr.7: 5291037-5291037 on Build GRCh38
rs370434277 Chr.7: 5291245-5291245 on Build GRCh38
rs1041592553 Chr.7: 5291115-5291115 on Build GRCh38
rs779817725 Chr.7: 5291093-5291093 on Build GRCh38
rs17855675 Chr.7: 5291194-5291194 on Build GRCh38
rs1019321592 Chr.7: 5291036-5291036 on Build GRCh38
rs899482646 Chr.7: 5291293-5291293 on Build GRCh38
rs776093640 Chr.7: 5290913-5290913 on Build GRCh38
rs759306260 Chr.7: 5291107-5291107 on Build GRCh38
rs748632646 Chr.7: 5291263-5291263 on Build GRCh38
rs764760265 Chr.7: 5291035-5291035 on Build GRCh38
rs369769905 Chr.7: 5291241-5291241 on Build GRCh38
rs563430306 Chr.7: 5291087-5291087 on Build GRCh38
rs145785340 Chr.7: 5291233-5291233 on Build GRCh38
rs749623687 Chr.7: 5291195-5291195 on Build GRCh38
rs765983478 Chr.7: 5291084-5291084 on Build GRCh38
rs975639916 Chr.7: 5291050-5291050 on Build GRCh38
rs557234442 Chr.7: 5291281-5291281 on Build GRCh38
rs1019935397 Chr.7: 5290979-5290979 on Build GRCh38
rs959610877 Chr.7: 5291369-5291369 on Build GRCh38
rs765611746 Chr.7: 5290911-5290911 on Build GRCh38
rs765100275 Chr.7: 5291288-5291288 on Build GRCh38
rs183490341 Chr.7: 5290978-5290978 on Build GRCh38
rs751123970 Chr.7: 5291081-5291081 on Build GRCh38
rs538286344 Chr.7: 5291177-5291177 on Build GRCh38
rs538251295 Chr.7: 5291057-5291057 on Build GRCh38
rs778728743 Chr.7: 5291158-5291158 on Build GRCh38
rs907034992 Chr.7: 5290925-5290925 on Build GRCh38
rs754877063 Chr.7: 5291155-5291155 on Build GRCh38
rs757442875 Chr.7: 5291169-5291169 on Build GRCh38
rs766013204 Chr.7: 5291217-5291217 on Build GRCh38
rs776429681 Chr.7: 5291219-5291219 on Build GRCh38
rs755332633 Chr.7: 5291257-5291257 on Build GRCh38
rs1034590021 Chr.7: 5290939-5290939 on Build GRCh38
rs746515095 Chr.7: 5291273-5291273 on Build GRCh38
rs760287143 Chr.7: 5291145-5291145 on Build GRCh38
rs893095613 Chr.7: 5290949-5290949 on Build GRCh38
rs549204893 Chr.7: 5291132-5291132 on Build GRCh38
rs375525665 Chr.7: 5291190-5291190 on Build GRCh38
rs375751095 Chr.7: 5291089-5291089 on Build GRCh38
rs759271432 Chr.7: 5291223-5291223 on Build GRCh38
rs371767928 Chr.7: 5291183-5291183 on Build GRCh38
rs771541135 Chr.7: 5291208-5291208 on Build GRCh38
rs756289585 Chr.7: 5291248-5291248 on Build GRCh38
rs752684921 Chr.7: 5291153-5291153 on Build GRCh38
rs367878795 Chr.7: 5291110-5291110 on Build GRCh38
rs766584252 Chr.7: 5291371-5291372 on Build GRCh38
rs373343179 Chr.7: 5291269-5291269 on Build GRCh38
rs533237982 Chr.7: 5291059-5291059 on Build GRCh38
rs192901389 Chr.7: 5291294-5291294 on Build GRCh38
rs775763107 Chr.7: 5291118-5291118 on Build GRCh38
rs763500311 Chr.7: 5291125-5291125 on Build GRCh38
rs772030743 Chr.7: 5290963-5290963 on Build GRCh38
rs769105333 Chr.7: 5291127-5291127 on Build GRCh38
rs761330090 Chr.7: 5291135-5291135 on Build GRCh38
rs774419676 Chr.7: 5291261-5291280 on Build GRCh38
rs575443353 Chr.7: 5290935-5290935 on Build GRCh38
rs368196054 Chr.7: 5291094-5291094 on Build GRCh38
rs146784971 Chr.7: 5291372-5291376 on Build GRCh38
rs763407024 Chr.7: 5290915-5290915 on Build GRCh38
rs754920521 Chr.7: 5291099-5291099 on Build GRCh38
rs746063078 Chr.7: 5291178-5291178 on Build GRCh38
rs781460370 Chr.7: 5291171-5291171 on Build GRCh38
rs938092292 Chr.7: 5291101-5291101 on Build GRCh38
rs769153457 Chr.7: 5291200-5291200 on Build GRCh38
rs926659397 Chr.7: 5291352-5291352 on Build GRCh38
rs371724080 Chr.7: 5291100-5291100 on Build GRCh38
rs112618708 Chr.7: 5291244-5291244 on Build GRCh38
rs770162183 Chr.7: 5291113-5291113 on Build GRCh38
rs573636652 Chr.7: 5291334-5291334 on Build GRCh38
rs538906579 Chr.7: 5291256-5291256 on Build GRCh38
rs571328947 Chr.7: 5291253-5291253 on Build GRCh38
rs970963559 Chr.7: 5291218-5291218 on Build GRCh38
rs555181078 Chr.7: 5291309-5291309 on Build GRCh38
rs1046455089 Chr.7: 5290919-5290919 on Build GRCh38
rs112877955 Chr.7: 5291371-5291371 on Build GRCh38
rs540656952 Chr.7: 5291003-5291003 on Build GRCh38
rs567644235 Chr.7: 5291182-5291182 on Build GRCh38
rs982760666 Chr.7: 5291335-5291335 on Build GRCh38
rs375977271 Chr.7: 5291108-5291108 on Build GRCh38
rs753634614 Chr.7: 5291154-5291154 on Build GRCh38
rs779292858 Chr.7: 5291262-5291262 on Build GRCh38
rs760363818 Chr.7: 5291216-5291216 on Build GRCh38
rs947180705 Chr.7: 5291242-5291242 on Build GRCh38
rs772625602 Chr.7: 5291268-5291268 on Build GRCh38
rs113532570 Chr.7: 5290986-5290986 on Build GRCh38
rs149798710 Chr.7: 5291144-5291144 on Build GRCh38
rs774864187 Chr.7: 5291203-5291203 on Build GRCh38
rs751639276 Chr.7: 5291159-5291159 on Build GRCh38
rs1004418960 Chr.7: 5291314-5291314 on Build GRCh38
rs772760058 Chr.7: 5291212-5291212 on Build GRCh38
rs986531326 Chr.7: 5291008-5291008 on Build GRCh38
rs759990625 Chr.7: 5290909-5290909 on Build GRCh38
rs543066910 Chr.7: 5290936-5290936 on Build GRCh38
rs780157919 Chr.7: 5291251-5291251 on Build GRCh38
rs776177851 Chr.7: 5291282-5291282 on Build GRCh38
rs368499379 Chr.7: 5291079-5291079 on Build GRCh38

View Full Product Details