Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 75983652-75984156 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
POR
Entrez Gene ID
5447
Gene Name
cytochrome p450 oxidoreductase
Gene Aliases
CPR, CYPOR, P450R
UniGene
Hs.354056
Gene Chromosome Location
Chr.7: 75915102-75986855 on Build GRCh38
Transcripts
NM_000941.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1242686 c.965A>G p.H322R Chr.7: 75983755-75983755 on Build GRCh38
6579763 c.1044C>T p.V348V Chr.7: 75983834-75983834 on Build GRCh38
1242687 c.662A>G p.H221R Chr.7: 75983755-75983755 on Build GRCh38
3882430 c.1018A>G p.I340V Chr.7: 75983808-75983808 on Build GRCh38
307533 c.1019T>C p.I340T Chr.7: 75983809-75983809 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs782200489 Chr.7: 75983826-75983826 on Build GRCh38
rs782650331 Chr.7: 75983716-75983716 on Build GRCh38
rs373626024 Chr.7: 75983900-75983900 on Build GRCh38
rs556003823 Chr.7: 75983674-75983674 on Build GRCh38
rs782213803 Chr.7: 75983738-75983738 on Build GRCh38
rs368514691 Chr.7: 75983898-75983898 on Build GRCh38
rs782440366 Chr.7: 75983792-75983792 on Build GRCh38
rs781942398 Chr.7: 75983769-75983769 on Build GRCh38
rs372380641 Chr.7: 75983777-75983777 on Build GRCh38
rs782186218 Chr.7: 75983708-75983709 on Build GRCh38
rs72557940 Chr.7: 75983714-75983714 on Build GRCh38
rs538355181 Chr.7: 75983991-75983991 on Build GRCh38
rs41301406 Chr.7: 75984003-75984003 on Build GRCh38
rs782722775 Chr.7: 75983702-75983702 on Build GRCh38
rs72557942 Chr.7: 75983819-75983819 on Build GRCh38
rs782377179 Chr.7: 75983898-75983899 on Build GRCh38
rs545133270 Chr.7: 75983894-75983894 on Build GRCh38
rs373128937 Chr.7: 75983832-75983832 on Build GRCh38
rs952447160 Chr.7: 75983718-75983718 on Build GRCh38
rs899780848 Chr.7: 75983923-75983923 on Build GRCh38
rs187216317 Chr.7: 75983708-75983708 on Build GRCh38
rs567100500 Chr.7: 75984018-75984018 on Build GRCh38
rs782029570 Chr.7: 75983867-75983867 on Build GRCh38
rs562843415 Chr.7: 75983877-75983877 on Build GRCh38
rs376804667 Chr.7: 75983717-75983717 on Build GRCh38
rs781905621 Chr.7: 75983696-75983696 on Build GRCh38
rs1040891152 Chr.7: 75984062-75984062 on Build GRCh38
rs782425291 Chr.7: 75983818-75983818 on Build GRCh38
rs781796073 Chr.7: 75983706-75983706 on Build GRCh38
rs982488324 Chr.7: 75983749-75983749 on Build GRCh38
rs112270565 Chr.7: 75983756-75983756 on Build GRCh38
rs982337248 Chr.7: 75983983-75983983 on Build GRCh38
rs201568454 Chr.7: 75983828-75983828 on Build GRCh38
rs562241770 Chr.7: 75983790-75983790 on Build GRCh38
rs782448070 Chr.7: 75983744-75983744 on Build GRCh38
rs782535519 Chr.7: 75983701-75983701 on Build GRCh38
rs782160753 Chr.7: 75983763-75983763 on Build GRCh38
rs41301412 Chr.7: 75984067-75984067 on Build GRCh38
rs782419727 Chr.7: 75983766-75983766 on Build GRCh38
rs201960141 Chr.7: 75983803-75983803 on Build GRCh38
rs782312165 Chr.7: 75983721-75983721 on Build GRCh38
rs782396207 Chr.7: 75983742-75983742 on Build GRCh38
rs782143653 Chr.7: 75983873-75983873 on Build GRCh38
rs368889644 Chr.7: 75983905-75983905 on Build GRCh38
rs996731724 Chr.7: 75983943-75983943 on Build GRCh38
rs782424131 Chr.7: 75983820-75983820 on Build GRCh38
rs540924885 Chr.7: 75983757-75983757 on Build GRCh38
rs781947561 Chr.7: 75983907-75983907 on Build GRCh38
rs926937573 Chr.7: 75983794-75983794 on Build GRCh38
rs41301409 Chr.7: 75984051-75984051 on Build GRCh38
rs782133385 Chr.7: 75983689-75983689 on Build GRCh38
rs912019417 Chr.7: 75984050-75984050 on Build GRCh38
rs782270201 Chr.7: 75983815-75983815 on Build GRCh38
rs945483933 Chr.7: 75983875-75983875 on Build GRCh38
rs945408727 Chr.7: 75984035-75984035 on Build GRCh38
rs41301400 Chr.7: 75983957-75983957 on Build GRCh38
rs782612942 Chr.7: 75983732-75983732 on Build GRCh38
rs782647655 Chr.7: 75983810-75983810 on Build GRCh38
rs369776567 Chr.7: 75983831-75983831 on Build GRCh38
rs782658941 Chr.7: 75983822-75983822 on Build GRCh38
rs72557941 Chr.7: 75983774-75983774 on Build GRCh38
rs932573082 Chr.7: 75984105-75984105 on Build GRCh38
rs989669475 Chr.7: 75984011-75984011 on Build GRCh38
rs41301403 Chr.7: 75983980-75983980 on Build GRCh38
rs1015371002 Chr.7: 75983970-75983970 on Build GRCh38
rs782024141 Chr.7: 75983843-75983843 on Build GRCh38
rs782369494 Chr.7: 75983843-75983843 on Build GRCh38
rs527493996 Chr.7: 75983952-75983952 on Build GRCh38
rs782493618 Chr.7: 75983707-75983707 on Build GRCh38
rs72557944 Chr.7: 75983956-75983956 on Build GRCh38
rs782236213 Chr.7: 75983711-75983711 on Build GRCh38
rs782754173 Chr.7: 75983695-75983695 on Build GRCh38
rs72557943 Chr.7: 75983899-75983899 on Build GRCh38
rs574408042 Chr.7: 75984092-75984092 on Build GRCh38
rs781868952 Chr.7: 75983891-75983891 on Build GRCh38
rs750172302 Chr.7: 75984036-75984036 on Build GRCh38
rs533482743 Chr.7: 75983884-75983884 on Build GRCh38
rs41301397 Chr.7: 75983885-75983885 on Build GRCh38
rs781946056 Chr.7: 75983979-75983979 on Build GRCh38
rs781888274 Chr.7: 75983901-75983901 on Build GRCh38
rs549385228 Chr.7: 75983958-75983958 on Build GRCh38
rs376830846 Chr.7: 75983789-75983789 on Build GRCh38
rs782720450 Chr.7: 75983876-75983876 on Build GRCh38
rs1034230747 Chr.7: 75983988-75983988 on Build GRCh38
rs886062441 Chr.7: 75983724-75983724 on Build GRCh38
rs978479622 Chr.7: 75984045-75984045 on Build GRCh38
rs781831773 Chr.7: 75983778-75983778 on Build GRCh38
rs781974546 Chr.7: 75983829-75983829 on Build GRCh38

View Full Product Details