Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 83958258-83958769 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SEMA3A
Entrez Gene ID
10371
Gene Name
semaphorin 3A
Gene Aliases
COLL1, HH16, Hsema-I, Hsema-III, SEMA1, SEMAD, SEMAIII, SEMAL, SemD, coll-1
UniGene
Hs.252451
Gene Chromosome Location
Chr.7: 83956846-84492724 on Build GRCh38
Transcripts
NM_006080.2 XM_005250110.3 XM_005250111.4 XM_006715839.3 XM_011515734.2 XM_017011673.1

SNP Detail

SNP ID Chromosome Location
rs932163066 Chr.7: 83958320-83958320 on Build GRCh38
rs969231064 Chr.7: 83958304-83958304 on Build GRCh38
rs1019920467 Chr.7: 83958589-83958589 on Build GRCh38
rs554005403 Chr.7: 83958452-83958452 on Build GRCh38
rs7801026 Chr.7: 83958569-83958569 on Build GRCh38
rs572587883 Chr.7: 83958470-83958470 on Build GRCh38
rs905693499 Chr.7: 83958528-83958528 on Build GRCh38
rs987643386 Chr.7: 83958701-83958701 on Build GRCh38
rs550423031 Chr.7: 83958332-83958332 on Build GRCh38
rs1027340618 Chr.7: 83958520-83958520 on Build GRCh38
rs913446735 Chr.7: 83958296-83958296 on Build GRCh38
rs1039908196 Chr.7: 83958592-83958592 on Build GRCh38
rs1026300598 Chr.7: 83958644-83958644 on Build GRCh38
rs932159094 Chr.7: 83958637-83958637 on Build GRCh38
rs1051844620 Chr.7: 83958422-83958422 on Build GRCh38
rs1016558682 Chr.7: 83958679-83958679 on Build GRCh38
rs983912974 Chr.7: 83958291-83958291 on Build GRCh38
rs754906092 Chr.7: 83958642-83958644 on Build GRCh38
rs1035368767 Chr.7: 83958569-83958570 on Build GRCh38
rs866907894 Chr.7: 83958504-83958504 on Build GRCh38
rs7802100 Chr.7: 83958599-83958599 on Build GRCh38
rs988447664 Chr.7: 83958488-83958488 on Build GRCh38
rs980573897 Chr.7: 83958305-83958305 on Build GRCh38
rs566160327 Chr.7: 83958342-83958342 on Build GRCh38
rs548258924 Chr.7: 83958647-83958647 on Build GRCh38
rs890576000 Chr.7: 83958425-83958425 on Build GRCh38
rs939292566 Chr.7: 83958454-83958454 on Build GRCh38
rs940635046 Chr.7: 83958446-83958446 on Build GRCh38
rs913261058 Chr.7: 83958529-83958529 on Build GRCh38
rs560037260 Chr.7: 83958648-83958648 on Build GRCh38
rs897036420 Chr.7: 83958489-83958489 on Build GRCh38
rs969125369 Chr.7: 83958721-83958721 on Build GRCh38
rs995424935 Chr.7: 83958513-83958513 on Build GRCh38
rs922108962 Chr.7: 83958315-83958315 on Build GRCh38
rs1005626225 Chr.7: 83958674-83958674 on Build GRCh38
rs959691946 Chr.7: 83958284-83958284 on Build GRCh38
rs979308918 Chr.7: 83958315-83958315 on Build GRCh38
rs768053032 Chr.7: 83958587-83958587 on Build GRCh38
rs149682686 Chr.7: 83958580-83958580 on Build GRCh38
rs927773734 Chr.7: 83958362-83958362 on Build GRCh38
rs1017085237 Chr.7: 83958292-83958292 on Build GRCh38
rs947680452 Chr.7: 83958313-83958313 on Build GRCh38
rs968816852 Chr.7: 83958599-83958599 on Build GRCh38
rs1036384401 Chr.7: 83958471-83958471 on Build GRCh38
rs980467364 Chr.7: 83958742-83958742 on Build GRCh38
rs868603550 Chr.7: 83958565-83958565 on Build GRCh38
rs529691230 Chr.7: 83958630-83958630 on Build GRCh38
rs183962282 Chr.7: 83958519-83958519 on Build GRCh38
rs922214915 Chr.7: 83958619-83958619 on Build GRCh38
rs576816480 Chr.7: 83958576-83958576 on Build GRCh38

View Full Product Details