Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 87361310-87361813 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CROT
Entrez Gene ID
54677
Gene Name
carnitine O-octanoyltransferase
Gene Aliases
COT
UniGene
Hs.125039
Gene Chromosome Location
Chr.7: 87345635-87399796 on Build GRCh38
Transcripts
NM_001143935.1 NM_021151.3 XM_011516337.2 XM_017012370.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6635694 c.311C>T p.A104V Chr.7: 87361460-87361460 on Build GRCh38
1132926 c.403T>A p.Y135N Chr.7: 87361552-87361552 on Build GRCh38
485664 c.485T>C p.L162P Chr.7: 87361790-87361790 on Build GRCh38
3750052 c.280C>T p.R94C Chr.7: 87361429-87361429 on Build GRCh38
747764 c.385C>T p.L129F Chr.7: 87361534-87361534 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1050735675 Chr.7: 87361390-87361390 on Build GRCh38
rs758831385 Chr.7: 87361512-87361512 on Build GRCh38
rs755204543 Chr.7: 87361694-87361694 on Build GRCh38
rs149695092 Chr.7: 87361557-87361557 on Build GRCh38
rs754399073 Chr.7: 87361502-87361502 on Build GRCh38
rs549782479 Chr.7: 87361411-87361411 on Build GRCh38
rs772718483 Chr.7: 87361679-87361679 on Build GRCh38
rs146422634 Chr.7: 87361505-87361505 on Build GRCh38
rs751267864 Chr.7: 87361389-87361389 on Build GRCh38
rs867264477 Chr.7: 87361414-87361414 on Build GRCh38
rs184470229 Chr.7: 87361732-87361732 on Build GRCh38
rs766069576 Chr.7: 87361384-87361384 on Build GRCh38
rs770275831 Chr.7: 87361580-87361581 on Build GRCh38
rs755153166 Chr.7: 87361543-87361543 on Build GRCh38
rs576028938 Chr.7: 87361693-87361693 on Build GRCh38
rs771827440 Chr.7: 87361583-87361583 on Build GRCh38
rs751918418 Chr.7: 87361782-87361782 on Build GRCh38
rs767024587 Chr.7: 87361396-87361396 on Build GRCh38
rs3827653 Chr.7: 87361430-87361430 on Build GRCh38
rs763518234 Chr.7: 87361760-87361760 on Build GRCh38
rs963239444 Chr.7: 87361550-87361550 on Build GRCh38
rs747153492 Chr.7: 87361424-87361424 on Build GRCh38
rs980787485 Chr.7: 87361537-87361537 on Build GRCh38
rs545287241 Chr.7: 87361338-87361338 on Build GRCh38
rs554709981 Chr.7: 87361347-87361347 on Build GRCh38
rs561811108 Chr.7: 87361413-87361413 on Build GRCh38
rs763680381 Chr.7: 87361615-87361615 on Build GRCh38
rs200737876 Chr.7: 87361680-87361680 on Build GRCh38
rs6971676 Chr.7: 87361718-87361718 on Build GRCh38
rs761920131 Chr.7: 87361728-87361729 on Build GRCh38
rs145487734 Chr.7: 87361753-87361753 on Build GRCh38
rs773279246 Chr.7: 87361378-87361378 on Build GRCh38
rs775921558 Chr.7: 87361582-87361583 on Build GRCh38
rs140945935 Chr.7: 87361511-87361511 on Build GRCh38
rs148986377 Chr.7: 87361783-87361783 on Build GRCh38
rs760105748 Chr.7: 87361576-87361576 on Build GRCh38
rs552010365 Chr.7: 87361547-87361547 on Build GRCh38
rs764041945 Chr.7: 87361685-87361685 on Build GRCh38
rs772364247 Chr.7: 87361418-87361418 on Build GRCh38
rs768128333 Chr.7: 87361742-87361742 on Build GRCh38
rs577793067 Chr.7: 87361335-87361335 on Build GRCh38
rs773956752 Chr.7: 87361460-87361460 on Build GRCh38
rs763098385 Chr.7: 87361677-87361678 on Build GRCh38
rs535721145 Chr.7: 87361546-87361546 on Build GRCh38
rs771186096 Chr.7: 87361341-87361341 on Build GRCh38
rs766822686 Chr.7: 87361688-87361688 on Build GRCh38
rs752937841 Chr.7: 87361785-87361785 on Build GRCh38
rs775665102 Chr.7: 87361357-87361357 on Build GRCh38
rs747216846 Chr.7: 87361521-87361521 on Build GRCh38
rs748240367 Chr.7: 87361697-87361697 on Build GRCh38
rs188617695 Chr.7: 87361416-87361416 on Build GRCh38
rs751865318 Chr.7: 87361692-87361692 on Build GRCh38
rs776344240 Chr.7: 87361746-87361746 on Build GRCh38
rs758887503 Chr.7: 87361685-87361685 on Build GRCh38
rs754277085 Chr.7: 87361402-87361402 on Build GRCh38
rs370194673 Chr.7: 87361695-87361695 on Build GRCh38
rs199970637 Chr.7: 87361398-87361398 on Build GRCh38
rs750929666 Chr.7: 87361682-87361682 on Build GRCh38
rs745717997 Chr.7: 87361570-87361570 on Build GRCh38
rs111311863 Chr.7: 87361688-87361688 on Build GRCh38
rs373955214 Chr.7: 87361561-87361561 on Build GRCh38
rs142152283 Chr.7: 87361393-87361393 on Build GRCh38
rs773253678 Chr.7: 87361429-87361429 on Build GRCh38
rs756184084 Chr.7: 87361786-87361786 on Build GRCh38
rs767687967 Chr.7: 87361784-87361784 on Build GRCh38
rs775107161 Chr.7: 87361493-87361493 on Build GRCh38
rs1003965495 Chr.7: 87361599-87361599 on Build GRCh38
rs767220583 Chr.7: 87361471-87361471 on Build GRCh38
rs779243437 Chr.7: 87361345-87361345 on Build GRCh38
rs779775074 Chr.7: 87361734-87361734 on Build GRCh38
rs776951599 Chr.7: 87361497-87361497 on Build GRCh38
rs1052361957 Chr.7: 87361633-87361633 on Build GRCh38
rs35136455 Chr.7: 87361722-87361722 on Build GRCh38
rs112064599 Chr.7: 87361769-87361769 on Build GRCh38
rs778305374 Chr.7: 87361400-87361400 on Build GRCh38
rs563441866 Chr.7: 87361349-87361349 on Build GRCh38
rs775162744 Chr.7: 87361584-87361584 on Build GRCh38
rs933755169 Chr.7: 87361571-87361571 on Build GRCh38
rs780481683 Chr.7: 87361518-87361518 on Build GRCh38
rs771346008 Chr.7: 87361363-87361364 on Build GRCh38
rs181190933 Chr.7: 87361708-87361708 on Build GRCh38
rs760279726 Chr.7: 87361603-87361603 on Build GRCh38
rs1050704534 Chr.7: 87361638-87361638 on Build GRCh38
rs144004176 Chr.7: 87361503-87361503 on Build GRCh38
rs768848006 Chr.7: 87361427-87361427 on Build GRCh38
rs150802563 Chr.7: 87361431-87361431 on Build GRCh38
rs149947132 Chr.7: 87361678-87361678 on Build GRCh38
rs760228295 Chr.7: 87361498-87361498 on Build GRCh38
rs76309506 Chr.7: 87361507-87361507 on Build GRCh38
rs765732337 Chr.7: 87361681-87361681 on Build GRCh38
rs780285542 Chr.7: 87361422-87361422 on Build GRCh38
rs371152487 Chr.7: 87361366-87361366 on Build GRCh38
rs746676213 Chr.7: 87361737-87361737 on Build GRCh38
rs770793890 Chr.7: 87361444-87361444 on Build GRCh38
rs761288409 Chr.7: 87361747-87361747 on Build GRCh38
rs769788031 Chr.7: 87361372-87361372 on Build GRCh38
rs766875656 Chr.7: 87361762-87361762 on Build GRCh38
rs139396146 Chr.7: 87361461-87361461 on Build GRCh38
rs769325453 Chr.7: 87361750-87361750 on Build GRCh38
rs956109864 Chr.7: 87361473-87361473 on Build GRCh38
rs371217644 Chr.7: 87361675-87361675 on Build GRCh38
rs751830361 Chr.7: 87361715-87361716 on Build GRCh38
rs778761286 Chr.7: 87361713-87361713 on Build GRCh38
rs762867956 Chr.7: 87361382-87361382 on Build GRCh38

View Full Product Details