Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 128028978-128029485 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LRRC4
Entrez Gene ID
64101
Gene Name
leucine rich repeat containing 4
Gene Aliases
NAG14, NGL-2
UniGene
Hs.655003
Gene Chromosome Location
Chr.7: 128027071-128032273 on Build GRCh38
Transcripts
NM_022143.4 XM_011516461.2 XM_017012505.1 NM_014390.2

Gene Symbol
SND1
Entrez Gene ID
27044
Gene Name
staphylococcal nuclease and tudor domain containing 1
Gene Aliases
TDRD11, p100
UniGene
Hs.122523
Gene Chromosome Location
Chr.7: 127652119-128092607 on Build GRCh38
Transcripts
NM_022143.4 XM_011516461.2 XM_017012505.1 NM_014390.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2768735 c.1623C>T p.A541A Chr.7: 128029018-128029018 on Build GRCh38
261832 c.1216G>A p.D406N Chr.7: 128029425-128029425 on Build GRCh38
4587100 c.1492A>C p.T498P Chr.7: 128029149-128029149 on Build GRCh38
3831817 c.1210C>G p.L404V Chr.7: 128029431-128029431 on Build GRCh38
1085190 c.1183G>A p.A395T Chr.7: 128029458-128029458 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779529922 Chr.7: 128029417-128029417 on Build GRCh38
rs184416499 Chr.7: 128029347-128029347 on Build GRCh38
rs757471408 Chr.7: 128029206-128029206 on Build GRCh38
rs921880113 Chr.7: 128029355-128029355 on Build GRCh38
rs764553288 Chr.7: 128029299-128029299 on Build GRCh38
rs371222195 Chr.7: 128029165-128029165 on Build GRCh38
rs531914482 Chr.7: 128029205-128029205 on Build GRCh38
rs73721283 Chr.7: 128029006-128029006 on Build GRCh38
rs746290650 Chr.7: 128029149-128029149 on Build GRCh38
rs374792277 Chr.7: 128029153-128029153 on Build GRCh38
rs369606572 Chr.7: 128029458-128029458 on Build GRCh38
rs968496713 Chr.7: 128029275-128029275 on Build GRCh38
rs200457381 Chr.7: 128029069-128029069 on Build GRCh38
rs760145180 Chr.7: 128029011-128029011 on Build GRCh38
rs774982699 Chr.7: 128029334-128029334 on Build GRCh38
rs750645543 Chr.7: 128029386-128029386 on Build GRCh38
rs763445033 Chr.7: 128029162-128029162 on Build GRCh38
rs767304897 Chr.7: 128029082-128029082 on Build GRCh38
rs982383976 Chr.7: 128029442-128029442 on Build GRCh38
rs751774318 Chr.7: 128029202-128029202 on Build GRCh38
rs770638484 Chr.7: 128029235-128029235 on Build GRCh38
rs758579040 Chr.7: 128029131-128029131 on Build GRCh38
rs766036491 Chr.7: 128029102-128029102 on Build GRCh38
rs778062327 Chr.7: 128029425-128029425 on Build GRCh38
rs547391147 Chr.7: 128029422-128029422 on Build GRCh38
rs779369673 Chr.7: 128029316-128029316 on Build GRCh38
rs368708135 Chr.7: 128029451-128029451 on Build GRCh38
rs143703816 Chr.7: 128029333-128029333 on Build GRCh38
rs113169990 Chr.7: 128029018-128029018 on Build GRCh38
rs762940928 Chr.7: 128029272-128029272 on Build GRCh38
rs929508147 Chr.7: 128029110-128029110 on Build GRCh38
rs944590490 Chr.7: 128029114-128029114 on Build GRCh38
rs141815231 Chr.7: 128029450-128029450 on Build GRCh38
rs760896251 Chr.7: 128029456-128029456 on Build GRCh38
rs752557447 Chr.7: 128029124-128029124 on Build GRCh38
rs560143316 Chr.7: 128029083-128029083 on Build GRCh38
rs761641479 Chr.7: 128029356-128029356 on Build GRCh38
rs767269069 Chr.7: 128029381-128029381 on Build GRCh38
rs572177898 Chr.7: 128029120-128029120 on Build GRCh38
rs745454908 Chr.7: 128029426-128029426 on Build GRCh38
rs760464481 Chr.7: 128029177-128029177 on Build GRCh38
rs1003774576 Chr.7: 128029231-128029231 on Build GRCh38
rs375494923 Chr.7: 128029186-128029186 on Build GRCh38
rs767816304 Chr.7: 128029287-128029287 on Build GRCh38
rs368886768 Chr.7: 128029154-128029154 on Build GRCh38
rs774253630 Chr.7: 128029447-128029447 on Build GRCh38
rs951063528 Chr.7: 128029432-128029432 on Build GRCh38
rs770981043 Chr.7: 128029137-128029137 on Build GRCh38
rs915342400 Chr.7: 128029013-128029013 on Build GRCh38
rs754152622 Chr.7: 128029463-128029463 on Build GRCh38
rs770164159 Chr.7: 128029215-128029215 on Build GRCh38
rs759055705 Chr.7: 128029244-128029244 on Build GRCh38
rs745984603 Chr.7: 128029209-128029209 on Build GRCh38
rs532590910 Chr.7: 128029423-128029423 on Build GRCh38
rs761501656 Chr.7: 128029052-128029052 on Build GRCh38
rs765804071 Chr.7: 128029182-128029182 on Build GRCh38
rs146699127 Chr.7: 128029033-128029033 on Build GRCh38
rs753982427 Chr.7: 128029406-128029406 on Build GRCh38
rs757374250 Chr.7: 128029285-128029285 on Build GRCh38
rs1042651847 Chr.7: 128029076-128029076 on Build GRCh38
rs781397665 Chr.7: 128029146-128029146 on Build GRCh38
rs780056809 Chr.7: 128029306-128029306 on Build GRCh38
rs201166040 Chr.7: 128029346-128029346 on Build GRCh38
rs1022378418 Chr.7: 128029267-128029267 on Build GRCh38
rs139266176 Chr.7: 128029187-128029187 on Build GRCh38
rs776324571 Chr.7: 128029332-128029332 on Build GRCh38
rs769342110 Chr.7: 128029429-128029429 on Build GRCh38
rs758164381 Chr.7: 128029194-128029194 on Build GRCh38
rs758876125 Chr.7: 128029118-128029118 on Build GRCh38
rs752616161 Chr.7: 128029192-128029192 on Build GRCh38
rs781078670 Chr.7: 128029208-128029208 on Build GRCh38
rs756197644 Chr.7: 128029304-128029304 on Build GRCh38
rs1004638703 Chr.7: 128029189-128029189 on Build GRCh38
rs58695944 Chr.7: 128029003-128029003 on Build GRCh38
rs755548826 Chr.7: 128029315-128029315 on Build GRCh38
rs763808571 Chr.7: 128029281-128029281 on Build GRCh38
rs755500186 Chr.7: 128029416-128029416 on Build GRCh38
rs900153637 Chr.7: 128029122-128029122 on Build GRCh38
rs749810617 Chr.7: 128029313-128029313 on Build GRCh38
rs772976701 Chr.7: 128029170-128029170 on Build GRCh38
rs561511542 Chr.7: 128029190-128029190 on Build GRCh38
rs371687712 Chr.7: 128029109-128029109 on Build GRCh38
rs777598811 Chr.7: 128029197-128029197 on Build GRCh38
rs751367087 Chr.7: 128029282-128029282 on Build GRCh38
rs369764530 Chr.7: 128029225-128029225 on Build GRCh38
rs959348549 Chr.7: 128029238-128029238 on Build GRCh38
rs148125780 Chr.7: 128029446-128029446 on Build GRCh38
rs149413214 Chr.7: 128029324-128029324 on Build GRCh38
rs749917007 Chr.7: 128029097-128029097 on Build GRCh38
rs780582855 Chr.7: 128029219-128029219 on Build GRCh38
rs369406229 Chr.7: 128029431-128029431 on Build GRCh38
rs766500767 Chr.7: 128029402-128029402 on Build GRCh38
rs762817576 Chr.7: 128029339-128029339 on Build GRCh38
rs774095958 Chr.7: 128029351-128029351 on Build GRCh38
rs755655980 Chr.7: 128029098-128029098 on Build GRCh38
rs774924951 Chr.7: 128029229-128029229 on Build GRCh38
rs764702274 Chr.7: 128029249-128029249 on Build GRCh38
rs995820688 Chr.7: 128029126-128029126 on Build GRCh38
rs772889415 Chr.7: 128029453-128029453 on Build GRCh38
rs747276886 Chr.7: 128029136-128029136 on Build GRCh38
rs775963012 Chr.7: 128029243-128029243 on Build GRCh38
rs564369603 Chr.7: 128029232-128029232 on Build GRCh38
rs748553631 Chr.7: 128029322-128029322 on Build GRCh38
rs752948982 Chr.7: 128029099-128029099 on Build GRCh38
rs752614906 Chr.7: 128029268-128029268 on Build GRCh38
rs532669056 Chr.7: 128029392-128029392 on Build GRCh38

View Full Product Details