Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 128839940-128840434 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FLNC
Entrez Gene ID
2318
Gene Name
filamin C
Gene Aliases
ABP-280, ABP280A, ABPA, ABPL, FLN2, MFM5, MPD4
UniGene
Hs.58414
Gene Chromosome Location
Chr.7: 128830429-128859274 on Build GRCh38
Transcripts
NM_001127487.1 NM_001458.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1197411 c.1542G>T p.K514N Chr.7: 128840153-128840153 on Build GRCh38
3633071 c.1435G>C p.A479P Chr.7: 128840046-128840046 on Build GRCh38
452304 c.1489T>C p.F497L Chr.7: 128840100-128840100 on Build GRCh38
6665094 c.1536G>A p.T512T Chr.7: 128840147-128840147 on Build GRCh38
3633073 c.1542G>A p.K514K Chr.7: 128840153-128840153 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs767087470 Chr.7: 128839998-128839998 on Build GRCh38
rs1012536919 Chr.7: 128840065-128840065 on Build GRCh38
rs753700851 Chr.7: 128840017-128840017 on Build GRCh38
rs182501358 Chr.7: 128840007-128840007 on Build GRCh38
rs118056738 Chr.7: 128840085-128840085 on Build GRCh38
rs747289343 Chr.7: 128840029-128840029 on Build GRCh38
rs983662260 Chr.7: 128840307-128840307 on Build GRCh38
rs770120714 Chr.7: 128839985-128839985 on Build GRCh38
rs371509764 Chr.7: 128840013-128840013 on Build GRCh38
rs763330423 Chr.7: 128840159-128840162 on Build GRCh38
rs746359389 Chr.7: 128840037-128840037 on Build GRCh38
rs111806457 Chr.7: 128840162-128840162 on Build GRCh38
rs533419161 Chr.7: 128840028-128840028 on Build GRCh38
rs763782008 Chr.7: 128840153-128840153 on Build GRCh38
rs907635458 Chr.7: 128840325-128840325 on Build GRCh38
rs758715550 Chr.7: 128840192-128840192 on Build GRCh38
rs560530105 Chr.7: 128840041-128840041 on Build GRCh38
rs1044007387 Chr.7: 128840374-128840374 on Build GRCh38
rs375881193 Chr.7: 128840152-128840152 on Build GRCh38
rs943315242 Chr.7: 128839967-128839967 on Build GRCh38
rs368101036 Chr.7: 128840129-128840129 on Build GRCh38
rs761448496 Chr.7: 128839997-128839997 on Build GRCh38
rs947661715 Chr.7: 128840373-128840373 on Build GRCh38
rs765490580 Chr.7: 128840020-128840020 on Build GRCh38
rs781127889 Chr.7: 128840103-128840103 on Build GRCh38
rs780953548 Chr.7: 128840201-128840201 on Build GRCh38
rs1046320257 Chr.7: 128840080-128840080 on Build GRCh38
rs772697482 Chr.7: 128840046-128840046 on Build GRCh38
rs770264114 Chr.7: 128840082-128840082 on Build GRCh38
rs751708045 Chr.7: 128840086-128840086 on Build GRCh38
rs1008303104 Chr.7: 128840405-128840405 on Build GRCh38
rs760050314 Chr.7: 128840004-128840004 on Build GRCh38
rs745556227 Chr.7: 128840209-128840209 on Build GRCh38
rs376040467 Chr.7: 128839996-128839996 on Build GRCh38
rs751435394 Chr.7: 128840154-128840154 on Build GRCh38
rs750435647 Chr.7: 128840106-128840106 on Build GRCh38
rs201810745 Chr.7: 128840045-128840045 on Build GRCh38
rs749593461 Chr.7: 128840044-128840044 on Build GRCh38
rs1047165548 Chr.7: 128840308-128840308 on Build GRCh38
rs957391561 Chr.7: 128840118-128840118 on Build GRCh38
rs754318650 Chr.7: 128840182-128840182 on Build GRCh38
rs752476855 Chr.7: 128840022-128840022 on Build GRCh38
rs373574362 Chr.7: 128840269-128840269 on Build GRCh38
rs756141780 Chr.7: 128840178-128840178 on Build GRCh38
rs538058257 Chr.7: 128840357-128840357 on Build GRCh38
rs757254472 Chr.7: 128840030-128840030 on Build GRCh38
rs772928460 Chr.7: 128840000-128840000 on Build GRCh38
rs780486915 Chr.7: 128840116-128840116 on Build GRCh38
rs190302391 Chr.7: 128840413-128840413 on Build GRCh38
rs185491512 Chr.7: 128840374-128840374 on Build GRCh38
rs774649424 Chr.7: 128840387-128840387 on Build GRCh38
rs533740546 Chr.7: 128840331-128840331 on Build GRCh38
rs765212618 Chr.7: 128840081-128840081 on Build GRCh38
rs200935123 Chr.7: 128840124-128840124 on Build GRCh38
rs749823564 Chr.7: 128840117-128840117 on Build GRCh38
rs544782496 Chr.7: 128839968-128839968 on Build GRCh38
rs760412891 Chr.7: 128840048-128840048 on Build GRCh38
rs574342062 Chr.7: 128840346-128840346 on Build GRCh38
rs765122668 Chr.7: 128840018-128840018 on Build GRCh38
rs748491057 Chr.7: 128840191-128840191 on Build GRCh38
rs111324750 Chr.7: 128840161-128840161 on Build GRCh38
rs758281857 Chr.7: 128840024-128840024 on Build GRCh38
rs181134489 Chr.7: 128840175-128840175 on Build GRCh38
rs575670745 Chr.7: 128839993-128839993 on Build GRCh38
rs757555168 Chr.7: 128840087-128840087 on Build GRCh38
rs939122802 Chr.7: 128840344-128840344 on Build GRCh38
rs759069062 Chr.7: 128840137-128840137 on Build GRCh38
rs768868050 Chr.7: 128840123-128840123 on Build GRCh38
rs780144889 Chr.7: 128840181-128840181 on Build GRCh38
rs916288568 Chr.7: 128840372-128840372 on Build GRCh38
rs770337434 Chr.7: 128840056-128840056 on Build GRCh38
rs578068207 Chr.7: 128840395-128840395 on Build GRCh38
rs189525930 Chr.7: 128840130-128840130 on Build GRCh38
rs551122435 Chr.7: 128840157-128840157 on Build GRCh38
rs746426370 Chr.7: 128839973-128839973 on Build GRCh38
rs753110010 Chr.7: 128840122-128840123 on Build GRCh38
rs794727967 Chr.7: 128840113-128840113 on Build GRCh38
rs185847889 Chr.7: 128840125-128840125 on Build GRCh38
rs767720224 Chr.7: 128840164-128840164 on Build GRCh38
rs769461137 Chr.7: 128840138-128840138 on Build GRCh38
rs1053055471 Chr.7: 128840391-128840391 on Build GRCh38
rs759126217 Chr.7: 128840072-128840072 on Build GRCh38
rs919664093 Chr.7: 128840187-128840187 on Build GRCh38
rs148068774 Chr.7: 128840334-128840334 on Build GRCh38
rs2291563 Chr.7: 128840069-128840069 on Build GRCh38
rs143610360 Chr.7: 128840036-128840036 on Build GRCh38
rs929689748 Chr.7: 128840217-128840217 on Build GRCh38
rs775538827 Chr.7: 128840146-128840146 on Build GRCh38
rs756196607 Chr.7: 128840114-128840114 on Build GRCh38
rs780542423 Chr.7: 128840043-128840043 on Build GRCh38
rs755578117 Chr.7: 128840183-128840183 on Build GRCh38
rs955475416 Chr.7: 128840139-128840139 on Build GRCh38

View Full Product Details