Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 129388964-129389460 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AHCYL2
Entrez Gene ID
23382
Gene Name
adenosylhomocysteinase like 2
Gene Aliases
ADOHCYASE3, IRBIT2
UniGene
Hs.600789
Gene Chromosome Location
Chr.7: 129225014-129430211 on Build GRCh38
Transcripts
NM_001130720.2 NM_001130722.2 NM_001130723.2 NM_015328.3 XM_005250237.1 XM_011515987.2 XM_011515988.2 XM_017011904.1 XM_017011905.1 XM_017011906.1 XM_017011907.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1448020 c.534G>T p.K178N Chr.7: 129389114-129389114 on Build GRCh38
6530257 c.274G>C p.E92Q Chr.7: 129389163-129389163 on Build GRCh38
127279 c.549C>T p.S183S Chr.7: 129389129-129389129 on Build GRCh38
4553616 c.287G>A p.R96K Chr.7: 129389176-129389176 on Build GRCh38
7261533 c.278A>G p.D93G Chr.7: 129389101-129389101 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs745870129 Chr.7: 129389034-129389035 on Build GRCh38
rs770162016 Chr.7: 129389099-129389099 on Build GRCh38
rs764427748 Chr.7: 129389130-129389130 on Build GRCh38
rs898369460 Chr.7: 129389404-129389404 on Build GRCh38
rs766793750 Chr.7: 129389201-129389201 on Build GRCh38
rs746614999 Chr.7: 129389231-129389231 on Build GRCh38
rs941551093 Chr.7: 129389300-129389300 on Build GRCh38
rs895707625 Chr.7: 129389403-129389403 on Build GRCh38
rs376412412 Chr.7: 129389029-129389029 on Build GRCh38
rs1022825206 Chr.7: 129389056-129389056 on Build GRCh38
rs762018637 Chr.7: 129389147-129389147 on Build GRCh38
rs752407857 Chr.7: 129389014-129389014 on Build GRCh38
rs1050814463 Chr.7: 129389318-129389318 on Build GRCh38
rs750447678 Chr.7: 129389173-129389173 on Build GRCh38
rs769933305 Chr.7: 129389045-129389045 on Build GRCh38
rs778706060 Chr.7: 129389223-129389225 on Build GRCh38
rs777434202 Chr.7: 129389222-129389222 on Build GRCh38
rs898922548 Chr.7: 129389352-129389352 on Build GRCh38
rs62480637 Chr.7: 129389405-129389405 on Build GRCh38
rs398086162 Chr.7: 129389026-129389026 on Build GRCh38
rs776455331 Chr.7: 129389102-129389102 on Build GRCh38
rs911419276 Chr.7: 129389274-129389274 on Build GRCh38
rs148295579 Chr.7: 129389122-129389122 on Build GRCh38
rs35404531 Chr.7: 129389017-129389017 on Build GRCh38
rs199536323 Chr.7: 129389027-129389027 on Build GRCh38
rs10954231 Chr.7: 129389252-129389252 on Build GRCh38
rs750905488 Chr.7: 129389033-129389033 on Build GRCh38
rs902676415 Chr.7: 129389283-129389283 on Build GRCh38
rs529051619 Chr.7: 129389286-129389286 on Build GRCh38
rs780517347 Chr.7: 129389049-129389049 on Build GRCh38
rs541972541 Chr.7: 129389017-129389018 on Build GRCh38
rs561639214 Chr.7: 129389269-129389269 on Build GRCh38
rs564152454 Chr.7: 129389057-129389057 on Build GRCh38
rs377149539 Chr.7: 129389379-129389379 on Build GRCh38
rs572851652 Chr.7: 129389251-129389251 on Build GRCh38
rs751140597 Chr.7: 129389267-129389267 on Build GRCh38
rs780791057 Chr.7: 129389235-129389235 on Build GRCh38
rs988816413 Chr.7: 129389151-129389151 on Build GRCh38
rs757506306 Chr.7: 129389016-129389017 on Build GRCh38
rs1037385396 Chr.7: 129389312-129389312 on Build GRCh38
rs770388020 Chr.7: 129389093-129389093 on Build GRCh38
rs61371803 Chr.7: 129389402-129389403 on Build GRCh38
rs928970869 Chr.7: 129389359-129389359 on Build GRCh38
rs762314576 Chr.7: 129389226-129389226 on Build GRCh38
rs1025481592 Chr.7: 129389071-129389071 on Build GRCh38
rs1047423689 Chr.7: 129389385-129389385 on Build GRCh38
rs200782522 Chr.7: 129389015-129389015 on Build GRCh38
rs372217666 Chr.7: 129389030-129389030 on Build GRCh38
rs746525069 Chr.7: 129389092-129389092 on Build GRCh38
rs754288760 Chr.7: 129389206-129389206 on Build GRCh38
rs759563174 Chr.7: 129389106-129389106 on Build GRCh38
rs889560152 Chr.7: 129389346-129389349 on Build GRCh38
rs1014495217 Chr.7: 129389034-129389034 on Build GRCh38
rs755266867 Chr.7: 129389211-129389211 on Build GRCh38
rs762826250 Chr.7: 129389128-129389128 on Build GRCh38
rs797005355 Chr.7: 129389027-129389027 on Build GRCh38
rs181166021 Chr.7: 129389377-129389377 on Build GRCh38
rs756457852 Chr.7: 129389036-129389036 on Build GRCh38
rs767589420 Chr.7: 129389165-129389165 on Build GRCh38
rs761633540 Chr.7: 129389291-129389291 on Build GRCh38
rs80057305 Chr.7: 129389402-129389402 on Build GRCh38
rs201615414 Chr.7: 129389114-129389114 on Build GRCh38
rs769583889 Chr.7: 129389109-129389109 on Build GRCh38
rs369142297 Chr.7: 129389140-129389140 on Build GRCh38
rs796955184 Chr.7: 129389017-129389018 on Build GRCh38
rs376155056 Chr.7: 129389032-129389032 on Build GRCh38
rs1016609408 Chr.7: 129389403-129389405 on Build GRCh38
rs777347868 Chr.7: 129389063-129389063 on Build GRCh38
rs11389356 Chr.7: 129389384-129389385 on Build GRCh38
rs373123443 Chr.7: 129389250-129389250 on Build GRCh38
rs756686551 Chr.7: 129389193-129389193 on Build GRCh38
rs200426402 Chr.7: 129389016-129389016 on Build GRCh38

View Full Product Details