Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 139035557-139036095 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZC3HAV1L
Entrez Gene ID
92092
Gene Name
zinc finger CCCH-type containing, antiviral 1 like
Gene Aliases
C7orf39
UniGene
Hs.512833
Gene Chromosome Location
Chr.7: 139021498-139036472 on Build GRCh38
Transcripts
NM_080660.3 XM_006716176.3 XM_011516688.2 XM_011516689.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6370722 c.203C>T p.A68V Chr.7: 139035815-139035815 on Build GRCh38
7274220 c.21C>T p.C7C Chr.7: 139035997-139035997 on Build GRCh38
4765962 c.352_353insC p.N118fs*13 Chr.7: 139035665-139035666 on Build GRCh38
6536410 c.301G>C p.A101P Chr.7: 139035717-139035717 on Build GRCh38
6497158 c.318C>T p.H106H Chr.7: 139035700-139035700 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs983427360 Chr.7: 139035609-139035609 on Build GRCh38
rs1030921206 Chr.7: 139036008-139036008 on Build GRCh38
rs778258375 Chr.7: 139036043-139036043 on Build GRCh38
rs760708929 Chr.7: 139035700-139035700 on Build GRCh38
rs989887284 Chr.7: 139036054-139036054 on Build GRCh38
rs1037900580 Chr.7: 139035853-139035853 on Build GRCh38
rs774164394 Chr.7: 139035969-139035969 on Build GRCh38
rs1039429642 Chr.7: 139035797-139035797 on Build GRCh38
rs770932085 Chr.7: 139035909-139035909 on Build GRCh38
rs560015943 Chr.7: 139035907-139035907 on Build GRCh38
rs748901612 Chr.7: 139035855-139035855 on Build GRCh38
rs541598734 Chr.7: 139035577-139035577 on Build GRCh38
rs763843904 Chr.7: 139035950-139035950 on Build GRCh38
rs763946851 Chr.7: 139035747-139035747 on Build GRCh38
rs746341322 Chr.7: 139035804-139035812 on Build GRCh38
rs865896552 Chr.7: 139035818-139035818 on Build GRCh38
rs776856943 Chr.7: 139035728-139035728 on Build GRCh38
rs921648854 Chr.7: 139035799-139035799 on Build GRCh38
rs757238947 Chr.7: 139035779-139035779 on Build GRCh38
rs921895117 Chr.7: 139036041-139036041 on Build GRCh38
rs982037447 Chr.7: 139035662-139035662 on Build GRCh38
rs914634136 Chr.7: 139035775-139035775 on Build GRCh38
rs563923758 Chr.7: 139035817-139035817 on Build GRCh38
rs776785925 Chr.7: 139035911-139035911 on Build GRCh38
rs780728506 Chr.7: 139035766-139035767 on Build GRCh38
rs956646748 Chr.7: 139036010-139036010 on Build GRCh38
rs376177836 Chr.7: 139035862-139035862 on Build GRCh38
rs753487086 Chr.7: 139036014-139036014 on Build GRCh38
rs892787268 Chr.7: 139035831-139035831 on Build GRCh38
rs921432014 Chr.7: 139036037-139036037 on Build GRCh38
rs779584170 Chr.7: 139035851-139035851 on Build GRCh38
rs748538197 Chr.7: 139036032-139036032 on Build GRCh38
rs754631700 Chr.7: 139036015-139036015 on Build GRCh38
rs532882453 Chr.7: 139035838-139035838 on Build GRCh38
rs766613463 Chr.7: 139035727-139035727 on Build GRCh38
rs755686766 Chr.7: 139036009-139036009 on Build GRCh38
rs772602776 Chr.7: 139035904-139035904 on Build GRCh38
rs750325837 Chr.7: 139036003-139036003 on Build GRCh38
rs189417805 Chr.7: 139036035-139036035 on Build GRCh38
rs974872234 Chr.7: 139036039-139036039 on Build GRCh38
rs759791547 Chr.7: 139035924-139035924 on Build GRCh38
rs886728556 Chr.7: 139035847-139035847 on Build GRCh38
rs951442310 Chr.7: 139036020-139036020 on Build GRCh38
rs747512198 Chr.7: 139036045-139036045 on Build GRCh38
rs528718214 Chr.7: 139035990-139035990 on Build GRCh38
rs774721975 Chr.7: 139035637-139035637 on Build GRCh38
rs779318940 Chr.7: 139035879-139035879 on Build GRCh38
rs951446391 Chr.7: 139035601-139035601 on Build GRCh38
rs561465664 Chr.7: 139035578-139035578 on Build GRCh38
rs748697958 Chr.7: 139035889-139035889 on Build GRCh38
rs765905538 Chr.7: 139036012-139036012 on Build GRCh38
rs945753819 Chr.7: 139035835-139035835 on Build GRCh38
rs548765851 Chr.7: 139036026-139036026 on Build GRCh38
rs575209710 Chr.7: 139035590-139035590 on Build GRCh38
rs756645233 Chr.7: 139036003-139036004 on Build GRCh38
rs995541133 Chr.7: 139035983-139035983 on Build GRCh38
rs989369622 Chr.7: 139035743-139035743 on Build GRCh38
rs537101211 Chr.7: 139036051-139036051 on Build GRCh38
rs894351533 Chr.7: 139035943-139035943 on Build GRCh38
rs1027098395 Chr.7: 139035986-139035986 on Build GRCh38
rs762661502 Chr.7: 139035740-139035740 on Build GRCh38
rs935154773 Chr.7: 139035729-139035729 on Build GRCh38
rs1003422063 Chr.7: 139035992-139035992 on Build GRCh38
rs749982553 Chr.7: 139035841-139035841 on Build GRCh38
rs748680843 Chr.7: 139035640-139035640 on Build GRCh38
rs751509427 Chr.7: 139035753-139035753 on Build GRCh38
rs965665416 Chr.7: 139036034-139036034 on Build GRCh38
rs772193104 Chr.7: 139035676-139035676 on Build GRCh38
rs769004458 Chr.7: 139035610-139035610 on Build GRCh38
rs773249595 Chr.7: 139035694-139035694 on Build GRCh38
rs755633887 Chr.7: 139035843-139035843 on Build GRCh38
rs944180258 Chr.7: 139035693-139035693 on Build GRCh38
rs773663345 Chr.7: 139035905-139035905 on Build GRCh38
rs544199512 Chr.7: 139035697-139035697 on Build GRCh38
rs975591622 Chr.7: 139035798-139035798 on Build GRCh38
rs761658876 Chr.7: 139035976-139035976 on Build GRCh38
rs947524914 Chr.7: 139035784-139035784 on Build GRCh38

View Full Product Details