Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 41930143-41930637 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KAT6A
Entrez Gene ID
7994
Gene Name
lysine acetyltransferase 6A
Gene Aliases
MOZ, MRD32, MYST-3, MYST3, RUNXBP2, ZC2HC6A, ZNF220
UniGene
Hs.491577
Gene Chromosome Location
Chr.8: 41929479-42052026 on Build GRCh38
Transcripts
NM_006766.4 XM_011544656.2 XM_011544657.2 XM_011544658.2 XM_011544659.2 XM_017013863.1 XM_017013864.1

SNP Detail

SNP ID Chromosome Location
rs1018707038 Chr.8: 41930616-41930616 on Build GRCh38
rs958099107 Chr.8: 41930252-41930252 on Build GRCh38
rs748358221 Chr.8: 41930187-41930188 on Build GRCh38
rs929670815 Chr.8: 41930340-41930340 on Build GRCh38
rs1039117557 Chr.8: 41930475-41930475 on Build GRCh38
rs577438796 Chr.8: 41930485-41930485 on Build GRCh38
rs1014144803 Chr.8: 41930429-41930429 on Build GRCh38
rs202132528 Chr.8: 41930441-41930441 on Build GRCh38
rs933498593 Chr.8: 41930497-41930498 on Build GRCh38
rs747571452 Chr.8: 41930387-41930387 on Build GRCh38
rs922781515 Chr.8: 41930295-41930295 on Build GRCh38
rs970762850 Chr.8: 41930174-41930174 on Build GRCh38
rs1045501535 Chr.8: 41930441-41930441 on Build GRCh38
rs992434878 Chr.8: 41930369-41930370 on Build GRCh38
rs570830969 Chr.8: 41930407-41930407 on Build GRCh38
rs964548589 Chr.8: 41930284-41930284 on Build GRCh38
rs962363000 Chr.8: 41930297-41930297 on Build GRCh38
rs911504317 Chr.8: 41930278-41930278 on Build GRCh38
rs958262492 Chr.8: 41930578-41930578 on Build GRCh38
rs976886509 Chr.8: 41930289-41930289 on Build GRCh38
rs925243922 Chr.8: 41930427-41930427 on Build GRCh38
rs75790682 Chr.8: 41930444-41930444 on Build GRCh38
rs3832594 Chr.8: 41930191-41930195 on Build GRCh38
rs66535265 Chr.8: 41930188-41930192 on Build GRCh38
rs1030926822 Chr.8: 41930217-41930217 on Build GRCh38
rs886727961 Chr.8: 41930236-41930236 on Build GRCh38
rs764285935 Chr.8: 41930190-41930194 on Build GRCh38
rs1030571018 Chr.8: 41930232-41930232 on Build GRCh38
rs1031126886 Chr.8: 41930551-41930551 on Build GRCh38
rs555291451 Chr.8: 41930393-41930393 on Build GRCh38
rs973765305 Chr.8: 41930334-41930334 on Build GRCh38
rs557239406 Chr.8: 41930440-41930440 on Build GRCh38
rs999559061 Chr.8: 41930542-41930542 on Build GRCh38
rs537661413 Chr.8: 41930410-41930410 on Build GRCh38
rs909530367 Chr.8: 41930380-41930380 on Build GRCh38
rs879009726 Chr.8: 41930368-41930368 on Build GRCh38
rs903032574 Chr.8: 41930443-41930443 on Build GRCh38
rs1003833920 Chr.8: 41930258-41930258 on Build GRCh38
rs990137994 Chr.8: 41930257-41930257 on Build GRCh38
rs190681430 Chr.8: 41930580-41930580 on Build GRCh38
rs894420546 Chr.8: 41930409-41930409 on Build GRCh38
rs187050871 Chr.8: 41930495-41930495 on Build GRCh38
rs900643647 Chr.8: 41930168-41930168 on Build GRCh38
rs1046853697 Chr.8: 41930346-41930346 on Build GRCh38
rs568615187 Chr.8: 41930408-41930408 on Build GRCh38
rs1033408077 Chr.8: 41930342-41930342 on Build GRCh38
rs999568960 Chr.8: 41930183-41930183 on Build GRCh38

View Full Product Details