Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 42730219-42730737 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHRNB3
Entrez Gene ID
1142
Gene Name
cholinergic receptor nicotinic beta 3 subunit
Gene Aliases
-
UniGene
Hs.654576
Gene Chromosome Location
Chr.8: 42697419-42737407 on Build GRCh38
Transcripts
NM_000749.3 XM_011544390.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4971566 c.295G>T p.G99W Chr.8: 42730639-42730639 on Build GRCh38
4400489 c.308T>G p.I103S Chr.8: 42730652-42730652 on Build GRCh38
3649342 c.316C>T p.P106S Chr.8: 42730660-42730660 on Build GRCh38
3649341 c.305C>T p.S102F Chr.8: 42730649-42730649 on Build GRCh38
422119 c.349C>A p.L117I Chr.8: 42730693-42730693 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780587598 Chr.8: 42730465-42730465 on Build GRCh38
rs199573864 Chr.8: 42730540-42730540 on Build GRCh38
rs149613797 Chr.8: 42730692-42730692 on Build GRCh38
rs770493197 Chr.8: 42730634-42730634 on Build GRCh38
rs752690899 Chr.8: 42730704-42730704 on Build GRCh38
rs758424582 Chr.8: 42730610-42730610 on Build GRCh38
rs771681832 Chr.8: 42730651-42730651 on Build GRCh38
rs978709818 Chr.8: 42730409-42730409 on Build GRCh38
rs1014389548 Chr.8: 42730606-42730606 on Build GRCh38
rs914261767 Chr.8: 42730518-42730518 on Build GRCh38
rs907766850 Chr.8: 42730492-42730492 on Build GRCh38
rs749046596 Chr.8: 42730696-42730697 on Build GRCh38
rs747998864 Chr.8: 42730321-42730321 on Build GRCh38
rs377712071 Chr.8: 42730657-42730657 on Build GRCh38
rs778740784 Chr.8: 42730702-42730705 on Build GRCh38
rs55817013 Chr.8: 42730551-42730551 on Build GRCh38
rs998387478 Chr.8: 42730605-42730605 on Build GRCh38
rs1029784461 Chr.8: 42730367-42730367 on Build GRCh38
rs894094051 Chr.8: 42730295-42730295 on Build GRCh38
rs561206798 Chr.8: 42730359-42730359 on Build GRCh38
rs1055586672 Chr.8: 42730566-42730566 on Build GRCh38
rs755744915 Chr.8: 42730600-42730606 on Build GRCh38
rs750504248 Chr.8: 42730609-42730609 on Build GRCh38
rs140126510 Chr.8: 42730672-42730672 on Build GRCh38
rs769575283 Chr.8: 42730322-42730322 on Build GRCh38
rs761168131 Chr.8: 42730571-42730571 on Build GRCh38
rs549338666 Chr.8: 42730386-42730386 on Build GRCh38
rs76413629 Chr.8: 42730474-42730474 on Build GRCh38
rs1037351122 Chr.8: 42730520-42730520 on Build GRCh38
rs971290719 Chr.8: 42730431-42730431 on Build GRCh38
rs369390365 Chr.8: 42730375-42730375 on Build GRCh38
rs900004412 Chr.8: 42730363-42730363 on Build GRCh38
rs899994444 Chr.8: 42730592-42730592 on Build GRCh38
rs75170626 Chr.8: 42730699-42730699 on Build GRCh38
rs749052989 Chr.8: 42730625-42730625 on Build GRCh38
rs1037384185 Chr.8: 42730293-42730293 on Build GRCh38
rs1004519836 Chr.8: 42730273-42730273 on Build GRCh38
rs1012779025 Chr.8: 42730664-42730664 on Build GRCh38
rs1001306825 Chr.8: 42730351-42730351 on Build GRCh38
rs374032164 Chr.8: 42730678-42730678 on Build GRCh38
rs551966783 Chr.8: 42730470-42730470 on Build GRCh38
rs563257921 Chr.8: 42730419-42730419 on Build GRCh38
rs759050497 Chr.8: 42730553-42730553 on Build GRCh38
rs762852497 Chr.8: 42730440-42730440 on Build GRCh38
rs115881166 Chr.8: 42730463-42730463 on Build GRCh38
rs748058161 Chr.8: 42730704-42730705 on Build GRCh38
rs934302627 Chr.8: 42730473-42730473 on Build GRCh38
rs779588048 Chr.8: 42730686-42730692 on Build GRCh38
rs368226322 Chr.8: 42730562-42730562 on Build GRCh38
rs764606295 Chr.8: 42730658-42730658 on Build GRCh38
rs374984781 Chr.8: 42730618-42730618 on Build GRCh38
rs541367437 Chr.8: 42730319-42730319 on Build GRCh38
rs545919747 Chr.8: 42730400-42730400 on Build GRCh38
rs200121784 Chr.8: 42730560-42730560 on Build GRCh38
rs764520045 Chr.8: 42730572-42730572 on Build GRCh38
rs1014918305 Chr.8: 42730381-42730381 on Build GRCh38
rs774015181 Chr.8: 42730639-42730639 on Build GRCh38
rs768336422 Chr.8: 42730699-42730700 on Build GRCh38
rs776178196 Chr.8: 42730655-42730655 on Build GRCh38
rs753202547 Chr.8: 42730561-42730561 on Build GRCh38
rs915695667 Chr.8: 42730427-42730427 on Build GRCh38
rs748047952 Chr.8: 42730616-42730616 on Build GRCh38
rs781157626 Chr.8: 42730615-42730615 on Build GRCh38
rs1004197248 Chr.8: 42730382-42730382 on Build GRCh38
rs998667934 Chr.8: 42730366-42730366 on Build GRCh38
rs1052671545 Chr.8: 42730270-42730270 on Build GRCh38
rs757437380 Chr.8: 42730589-42730589 on Build GRCh38
rs962578669 Chr.8: 42730385-42730385 on Build GRCh38
rs940605585 Chr.8: 42730499-42730499 on Build GRCh38
rs762201079 Chr.8: 42730673-42730673 on Build GRCh38
rs778857137 Chr.8: 42730267-42730271 on Build GRCh38
rs758795959 Chr.8: 42730453-42730453 on Build GRCh38
rs370274047 Chr.8: 42730578-42730578 on Build GRCh38
rs964084322 Chr.8: 42730421-42730421 on Build GRCh38
rs770530288 Chr.8: 42730547-42730547 on Build GRCh38
rs758514542 Chr.8: 42730691-42730691 on Build GRCh38
rs779128053 Chr.8: 42730597-42730597 on Build GRCh38
rs1016903316 Chr.8: 42730383-42730383 on Build GRCh38

View Full Product Details