Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 10730066-10730558 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LOC102723313
Entrez Gene ID
102723313
Gene Name
uncharacterized LOC102723313
Gene Aliases
-
UniGene
Hs.583894
Gene Chromosome Location
Chr.8: 10728358-10771953 on Build GRCh38
Transcripts
NM_031439.3

Gene Symbol
SOX7
Entrez Gene ID
83595
Gene Name
SRY-box 7
Gene Aliases
-
UniGene
Hs.709543
Gene Chromosome Location
Chr.8: 10723768-10730574 on Build GRCh38
Transcripts
NM_031439.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6297641 c.53C>A p.A18D Chr.8: 10730381-10730381 on Build GRCh38
5030642 c.96_102delCCCCCGG p.P35fs*13 Chr.8: 10730332-10730338 on Build GRCh38
6303036 c.100C>T p.R34W Chr.8: 10730334-10730334 on Build GRCh38
1453959 c.177G>A p.E59E Chr.8: 10730257-10730257 on Build GRCh38
453809 c.135C>T p.I45I Chr.8: 10730299-10730299 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs752164129 Chr.8: 10730330-10730330 on Build GRCh38
rs541034707 Chr.8: 10730161-10730161 on Build GRCh38
rs991656737 Chr.8: 10730215-10730215 on Build GRCh38
rs781631864 Chr.8: 10730178-10730178 on Build GRCh38
rs760745676 Chr.8: 10730466-10730466 on Build GRCh38
rs755062601 Chr.8: 10730353-10730353 on Build GRCh38
rs780192420 Chr.8: 10730277-10730277 on Build GRCh38
rs776186767 Chr.8: 10730390-10730390 on Build GRCh38
rs957447009 Chr.8: 10730400-10730400 on Build GRCh38
rs756470317 Chr.8: 10730429-10730429 on Build GRCh38
rs527769142 Chr.8: 10730144-10730144 on Build GRCh38
rs746935283 Chr.8: 10730459-10730459 on Build GRCh38
rs757740144 Chr.8: 10730259-10730259 on Build GRCh38
rs549199093 Chr.8: 10730254-10730254 on Build GRCh38
rs756913483 Chr.8: 10730149-10730149 on Build GRCh38
rs746866287 Chr.8: 10730383-10730383 on Build GRCh38
rs377053507 Chr.8: 10730181-10730181 on Build GRCh38
rs751243613 Chr.8: 10730146-10730146 on Build GRCh38
rs1029303675 Chr.8: 10730532-10730532 on Build GRCh38
rs572208241 Chr.8: 10730499-10730499 on Build GRCh38
rs557472510 Chr.8: 10730322-10730322 on Build GRCh38
rs773476734 Chr.8: 10730189-10730190 on Build GRCh38
rs1000177021 Chr.8: 10730449-10730449 on Build GRCh38
rs545288397 Chr.8: 10730311-10730311 on Build GRCh38
rs959235417 Chr.8: 10730204-10730204 on Build GRCh38
rs774195594 Chr.8: 10730293-10730293 on Build GRCh38
rs775102680 Chr.8: 10730407-10730407 on Build GRCh38
rs529132388 Chr.8: 10730240-10730240 on Build GRCh38
rs866120475 Chr.8: 10730228-10730228 on Build GRCh38
rs547345150 Chr.8: 10730301-10730301 on Build GRCh38
rs765890102 Chr.8: 10730323-10730323 on Build GRCh38
rs376106254 Chr.8: 10730245-10730245 on Build GRCh38
rs945186942 Chr.8: 10730537-10730537 on Build GRCh38
rs890535209 Chr.8: 10730147-10730147 on Build GRCh38
rs899532312 Chr.8: 10730529-10730529 on Build GRCh38
rs754029549 Chr.8: 10730438-10730438 on Build GRCh38
rs186242121 Chr.8: 10730444-10730444 on Build GRCh38
rs369872856 Chr.8: 10730260-10730260 on Build GRCh38
rs768027133 Chr.8: 10730331-10730331 on Build GRCh38
rs779407943 Chr.8: 10730162-10730162 on Build GRCh38
rs112398503 Chr.8: 10730483-10730483 on Build GRCh38
rs890933278 Chr.8: 10730409-10730409 on Build GRCh38
rs746117020 Chr.8: 10730268-10730268 on Build GRCh38
rs766743289 Chr.8: 10730430-10730430 on Build GRCh38
rs886903457 Chr.8: 10730491-10730491 on Build GRCh38
rs373581511 Chr.8: 10730166-10730166 on Build GRCh38
rs778967436 Chr.8: 10730441-10730441 on Build GRCh38
rs778914334 Chr.8: 10730369-10730369 on Build GRCh38
rs767210512 Chr.8: 10730156-10730156 on Build GRCh38
rs761716324 Chr.8: 10730299-10730299 on Build GRCh38
rs770684932 Chr.8: 10730461-10730461 on Build GRCh38
rs763977905 Chr.8: 10730145-10730145 on Build GRCh38
rs772234520 Chr.8: 10730156-10730162 on Build GRCh38
rs542057637 Chr.8: 10730465-10730465 on Build GRCh38
rs1038217636 Chr.8: 10730134-10730135 on Build GRCh38
rs113170791 Chr.8: 10730128-10730128 on Build GRCh38
rs773006628 Chr.8: 10730374-10730374 on Build GRCh38
rs759035753 Chr.8: 10730325-10730325 on Build GRCh38
rs760271237 Chr.8: 10730230-10730230 on Build GRCh38
rs924678937 Chr.8: 10730142-10730142 on Build GRCh38
rs201119602 Chr.8: 10730231-10730231 on Build GRCh38
rs1024561478 Chr.8: 10730467-10730467 on Build GRCh38
rs1045375787 Chr.8: 10730167-10730167 on Build GRCh38
rs899783546 Chr.8: 10730142-10730148 on Build GRCh38
rs571195317 Chr.8: 10730392-10730392 on Build GRCh38
rs370694180 Chr.8: 10730477-10730477 on Build GRCh38
rs771929190 Chr.8: 10730373-10730373 on Build GRCh38
rs1013561811 Chr.8: 10730466-10730466 on Build GRCh38
rs534625650 Chr.8: 10730141-10730142 on Build GRCh38
rs372332206 Chr.8: 10730089-10730089 on Build GRCh38
rs746060890 Chr.8: 10730179-10730179 on Build GRCh38
rs941023676 Chr.8: 10730345-10730345 on Build GRCh38
rs750957212 Chr.8: 10730425-10730425 on Build GRCh38
rs768192694 Chr.8: 10730424-10730424 on Build GRCh38
rs1054481291 Chr.8: 10730150-10730150 on Build GRCh38
rs772949381 Chr.8: 10730314-10730314 on Build GRCh38
rs963673705 Chr.8: 10730275-10730275 on Build GRCh38
rs565823930 Chr.8: 10730272-10730272 on Build GRCh38
rs1008959720 Chr.8: 10730148-10730148 on Build GRCh38
rs768416438 Chr.8: 10730188-10730188 on Build GRCh38
rs199995288 Chr.8: 10730371-10730371 on Build GRCh38
rs1020603748 Chr.8: 10730326-10730326 on Build GRCh38
rs148916739 Chr.8: 10730290-10730290 on Build GRCh38
rs761659259 Chr.8: 10730213-10730213 on Build GRCh38
rs770050593 Chr.8: 10730180-10730180 on Build GRCh38
rs978789581 Chr.8: 10730181-10730182 on Build GRCh38
rs759209484 Chr.8: 10730394-10730394 on Build GRCh38
rs550013139 Chr.8: 10730226-10730226 on Build GRCh38
rs971503144 Chr.8: 10730470-10730470 on Build GRCh38
rs750757046 Chr.8: 10730335-10730335 on Build GRCh38
rs749382095 Chr.8: 10730283-10730283 on Build GRCh38
rs539760565 Chr.8: 10730435-10730435 on Build GRCh38
rs1051416200 Chr.8: 10730503-10730503 on Build GRCh38
rs771409658 Chr.8: 10730177-10730177 on Build GRCh38
rs934152161 Chr.8: 10730115-10730115 on Build GRCh38
rs770757055 Chr.8: 10730389-10730389 on Build GRCh38
rs1009260960 Chr.8: 10730095-10730095 on Build GRCh38
rs1005234295 Chr.8: 10730250-10730250 on Build GRCh38
rs899730499 Chr.8: 10730399-10730399 on Build GRCh38
rs1005292984 Chr.8: 10730498-10730498 on Build GRCh38
rs924751611 Chr.8: 10730334-10730334 on Build GRCh38
rs28438834 Chr.8: 10730453-10730453 on Build GRCh38
rs753223058 Chr.8: 10730234-10730234 on Build GRCh38
rs764777065 Chr.8: 10730329-10730329 on Build GRCh38
rs774327784 Chr.8: 10730191-10730191 on Build GRCh38
rs753516105 Chr.8: 10730324-10730324 on Build GRCh38
rs976036860 Chr.8: 10730085-10730085 on Build GRCh38
rs996904565 Chr.8: 10730530-10730530 on Build GRCh38
rs747279855 Chr.8: 10730174-10730174 on Build GRCh38
rs541998011 Chr.8: 10730108-10730108 on Build GRCh38
rs764679856 Chr.8: 10730396-10730396 on Build GRCh38
rs561893365 Chr.8: 10730124-10730124 on Build GRCh38
rs778205959 Chr.8: 10730173-10730173 on Build GRCh38
rs1003767401 Chr.8: 10730221-10730221 on Build GRCh38
rs762949676 Chr.8: 10730185-10730185 on Build GRCh38
rs749491728 Chr.8: 10730346-10730346 on Build GRCh38
rs758793273 Chr.8: 10730171-10730171 on Build GRCh38
rs932590022 Chr.8: 10730401-10730401 on Build GRCh38
rs762421135 Chr.8: 10730417-10730417 on Build GRCh38
rs756484143 Chr.8: 10730336-10730336 on Build GRCh38
rs780204034 Chr.8: 10730338-10730338 on Build GRCh38
rs778203278 Chr.8: 10730247-10730247 on Build GRCh38
rs555497960 Chr.8: 10730495-10730495 on Build GRCh38

View Full Product Details