Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 51345641-51346165 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PXDNL
Entrez Gene ID
137902
Gene Name
peroxidasin like
Gene Aliases
PMR1, PRM1, VPO2
UniGene
Hs.444882
Gene Chromosome Location
Chr.8: 51319575-51811031 on Build GRCh38
Transcripts
NM_144651.4 XM_005251168.3 XM_006716420.3 XM_011517456.2 XM_011517457.2 XM_011517458.2 XM_017013041.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
256168 c.3935C>T p.T1312M Chr.8: 51345914-51345914 on Build GRCh38
6760637 c.3966A>C p.Q1322H Chr.8: 51345883-51345883 on Build GRCh38
5670710 c.3929C>A p.A1310E Chr.8: 51345920-51345920 on Build GRCh38
272273 c.3936G>A p.T1312T Chr.8: 51345913-51345913 on Build GRCh38
2718321 c.3917G>T p.G1306V Chr.8: 51345932-51345932 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs771428185 Chr.8: 51345959-51345959 on Build GRCh38
rs141672452 Chr.8: 51346004-51346004 on Build GRCh38
rs1020067754 Chr.8: 51345827-51345827 on Build GRCh38
rs1006438080 Chr.8: 51346132-51346132 on Build GRCh38
rs868440408 Chr.8: 51346005-51346005 on Build GRCh38
rs747031210 Chr.8: 51345849-51345849 on Build GRCh38
rs1035811881 Chr.8: 51345724-51345724 on Build GRCh38
rs747842639 Chr.8: 51345815-51345815 on Build GRCh38
rs531194688 Chr.8: 51346091-51346091 on Build GRCh38
rs757055142 Chr.8: 51345906-51345906 on Build GRCh38
rs753615336 Chr.8: 51345905-51345905 on Build GRCh38
rs887917810 Chr.8: 51346126-51346126 on Build GRCh38
rs529156941 Chr.8: 51346136-51346136 on Build GRCh38
rs754306345 Chr.8: 51345790-51345791 on Build GRCh38
rs1005894440 Chr.8: 51346103-51346103 on Build GRCh38
rs768707574 Chr.8: 51345925-51345925 on Build GRCh38
rs754557711 Chr.8: 51345802-51345802 on Build GRCh38
rs538897133 Chr.8: 51345932-51345932 on Build GRCh38
rs757038555 Chr.8: 51345989-51345989 on Build GRCh38
rs774290354 Chr.8: 51345840-51345840 on Build GRCh38
rs758992372 Chr.8: 51345792-51345792 on Build GRCh38
rs780135369 Chr.8: 51345914-51345914 on Build GRCh38
rs11774588 Chr.8: 51345869-51345869 on Build GRCh38
rs1002635565 Chr.8: 51346080-51346080 on Build GRCh38
rs545553376 Chr.8: 51346076-51346076 on Build GRCh38
rs529091530 Chr.8: 51345837-51345837 on Build GRCh38
rs755487260 Chr.8: 51345828-51345828 on Build GRCh38
rs745869671 Chr.8: 51345923-51345923 on Build GRCh38
rs1050438660 Chr.8: 51345690-51345690 on Build GRCh38
rs147301930 Chr.8: 51345934-51345934 on Build GRCh38
rs773666642 Chr.8: 51345864-51345864 on Build GRCh38
rs763258771 Chr.8: 51345867-51345867 on Build GRCh38
rs765496785 Chr.8: 51345863-51345863 on Build GRCh38
rs575897545 Chr.8: 51346089-51346089 on Build GRCh38
rs770017416 Chr.8: 51345943-51345943 on Build GRCh38
rs763352489 Chr.8: 51345950-51345950 on Build GRCh38
rs780935381 Chr.8: 51345808-51345808 on Build GRCh38
rs753433885 Chr.8: 51345985-51345985 on Build GRCh38
rs376461949 Chr.8: 51345891-51345891 on Build GRCh38
rs11775272 Chr.8: 51345698-51345698 on Build GRCh38
rs867976190 Chr.8: 51345844-51345844 on Build GRCh38
rs370583510 Chr.8: 51346094-51346094 on Build GRCh38
rs570733328 Chr.8: 51345945-51345945 on Build GRCh38
rs1001245020 Chr.8: 51345725-51345725 on Build GRCh38
rs768724102 Chr.8: 51345850-51345850 on Build GRCh38
rs773247152 Chr.8: 51346138-51346138 on Build GRCh38
rs566976870 Chr.8: 51345870-51345870 on Build GRCh38
rs760138222 Chr.8: 51345979-51345979 on Build GRCh38
rs1054396781 Chr.8: 51345772-51345772 on Build GRCh38
rs755859869 Chr.8: 51345828-51345828 on Build GRCh38
rs778325354 Chr.8: 51345888-51345889 on Build GRCh38
rs923414167 Chr.8: 51345940-51345940 on Build GRCh38
rs368104169 Chr.8: 51345793-51345793 on Build GRCh38
rs769831692 Chr.8: 51345689-51345689 on Build GRCh38
rs776712193 Chr.8: 51345853-51345853 on Build GRCh38
rs561962735 Chr.8: 51346133-51346133 on Build GRCh38
rs115947796 Chr.8: 51345990-51345990 on Build GRCh38
rs536218034 Chr.8: 51345817-51345817 on Build GRCh38
rs938307676 Chr.8: 51346047-51346047 on Build GRCh38
rs758846002 Chr.8: 51345909-51345909 on Build GRCh38
rs763739878 Chr.8: 51345980-51345980 on Build GRCh38
rs751074382 Chr.8: 51345801-51345801 on Build GRCh38
rs894249284 Chr.8: 51346086-51346086 on Build GRCh38
rs971161181 Chr.8: 51345858-51345858 on Build GRCh38
rs571823463 Chr.8: 51346023-51346023 on Build GRCh38
rs933417944 Chr.8: 51345948-51345948 on Build GRCh38
rs1012867710 Chr.8: 51345771-51345771 on Build GRCh38
rs769011665 Chr.8: 51346065-51346065 on Build GRCh38
rs10091690 Chr.8: 51345976-51345976 on Build GRCh38
rs778752839 Chr.8: 51345913-51345913 on Build GRCh38
rs139851606 Chr.8: 51345961-51345961 on Build GRCh38
rs984842326 Chr.8: 51345892-51345892 on Build GRCh38
rs1010078593 Chr.8: 51345805-51345805 on Build GRCh38
rs149855662 Chr.8: 51345972-51345972 on Build GRCh38
rs543741883 Chr.8: 51346092-51346092 on Build GRCh38
rs193274927 Chr.8: 51345833-51345833 on Build GRCh38
rs777402487 Chr.8: 51345902-51345902 on Build GRCh38
rs145857441 Chr.8: 51346088-51346088 on Build GRCh38
rs767204951 Chr.8: 51345873-51345873 on Build GRCh38
rs972294035 Chr.8: 51346137-51346137 on Build GRCh38
rs755848253 Chr.8: 51345894-51345894 on Build GRCh38
rs531657507 Chr.8: 51345730-51345730 on Build GRCh38
rs1010568215 Chr.8: 51346017-51346017 on Build GRCh38
rs184214274 Chr.8: 51345975-51345975 on Build GRCh38
rs376784692 Chr.8: 51345791-51345791 on Build GRCh38
rs188278379 Chr.8: 51346018-51346018 on Build GRCh38
rs747649171 Chr.8: 51345942-51345943 on Build GRCh38
rs762151954 Chr.8: 51345859-51345859 on Build GRCh38

View Full Product Details