Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 54047770-54048280 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LYPLA1
Entrez Gene ID
10434
Gene Name
lysophospholipase I
Gene Aliases
APT-1, APT1, LPL-I, LPL1, hAPT1
UniGene
Hs.744046
Gene Chromosome Location
Chr.8: 54046367-54102017 on Build GRCh38
Transcripts
NM_001279356.1 NM_001279357.1 NM_001279358.1 NM_001279359.1 NM_001279360.1 NM_006330.3 XM_005251127.4 XM_017012956.1 XM_017012957.1 XM_017012958.1 XM_017012959.1 XM_017012960.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6588047 c.688G>A p.D230N Chr.8: 54048070-54048070 on Build GRCh38
4755567 c.658C>T p.Q220* Chr.8: 54048100-54048100 on Build GRCh38
5874806 c.652G>C p.V218L Chr.8: 54048106-54048106 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs748309283 Chr.8: 54048017-54048019 on Build GRCh38
rs754313139 Chr.8: 54048079-54048079 on Build GRCh38
rs547596792 Chr.8: 54048101-54048101 on Build GRCh38
rs1009914594 Chr.8: 54048144-54048144 on Build GRCh38
rs562054386 Chr.8: 54048050-54048050 on Build GRCh38
rs891500501 Chr.8: 54048135-54048135 on Build GRCh38
rs913598333 Chr.8: 54048112-54048112 on Build GRCh38
rs950135858 Chr.8: 54048233-54048235 on Build GRCh38
rs199878046 Chr.8: 54048040-54048040 on Build GRCh38
rs758092929 Chr.8: 54048102-54048102 on Build GRCh38
rs764541154 Chr.8: 54048075-54048075 on Build GRCh38
rs188908999 Chr.8: 54048156-54048156 on Build GRCh38
rs945246099 Chr.8: 54047833-54047833 on Build GRCh38
rs894568218 Chr.8: 54048219-54048219 on Build GRCh38
rs369501704 Chr.8: 54048148-54048148 on Build GRCh38
rs569675062 Chr.8: 54048187-54048187 on Build GRCh38
rs1044604 Chr.8: 54047939-54047939 on Build GRCh38
rs779722996 Chr.8: 54048115-54048115 on Build GRCh38
rs879223752 Chr.8: 54048236-54048236 on Build GRCh38
rs776587983 Chr.8: 54048014-54048014 on Build GRCh38
rs772266554 Chr.8: 54048127-54048129 on Build GRCh38
rs368393610 Chr.8: 54048107-54048107 on Build GRCh38
rs772224293 Chr.8: 54048149-54048149 on Build GRCh38
rs1056314979 Chr.8: 54047892-54047892 on Build GRCh38
rs1043651474 Chr.8: 54047841-54047841 on Build GRCh38
rs533367365 Chr.8: 54047961-54047961 on Build GRCh38
rs937822946 Chr.8: 54047868-54047868 on Build GRCh38
rs940915079 Chr.8: 54048220-54048220 on Build GRCh38
rs1046220129 Chr.8: 54048250-54048250 on Build GRCh38
rs570841619 Chr.8: 54048127-54048127 on Build GRCh38
rs976060374 Chr.8: 54048246-54048246 on Build GRCh38
rs773485589 Chr.8: 54048134-54048138 on Build GRCh38
rs192519511 Chr.8: 54047978-54047978 on Build GRCh38
rs745645842 Chr.8: 54048192-54048192 on Build GRCh38
rs545933796 Chr.8: 54047940-54047940 on Build GRCh38
rs894524975 Chr.8: 54047925-54047925 on Build GRCh38
rs1034553736 Chr.8: 54048179-54048179 on Build GRCh38
rs1054924879 Chr.8: 54048193-54048193 on Build GRCh38
rs535498375 Chr.8: 54048188-54048188 on Build GRCh38
rs372447255 Chr.8: 54048121-54048121 on Build GRCh38
rs908128723 Chr.8: 54048212-54048212 on Build GRCh38
rs975600729 Chr.8: 54048025-54048025 on Build GRCh38
rs776974209 Chr.8: 54048159-54048159 on Build GRCh38
rs771080578 Chr.8: 54048033-54048033 on Build GRCh38
rs150305251 Chr.8: 54048083-54048083 on Build GRCh38
rs769438282 Chr.8: 54048225-54048225 on Build GRCh38
rs375134683 Chr.8: 54048133-54048133 on Build GRCh38
rs765206207 Chr.8: 54047977-54047977 on Build GRCh38
rs763152302 Chr.8: 54048019-54048019 on Build GRCh38
rs775768892 Chr.8: 54048064-54048064 on Build GRCh38
rs765758869 Chr.8: 54047797-54047797 on Build GRCh38
rs756764292 Chr.8: 54048082-54048082 on Build GRCh38
rs555784304 Chr.8: 54048253-54048253 on Build GRCh38
rs762363003 Chr.8: 54048168-54048168 on Build GRCh38
rs866954760 Chr.8: 54047819-54047819 on Build GRCh38
rs1014671901 Chr.8: 54047932-54047932 on Build GRCh38
rs962889633 Chr.8: 54047996-54047996 on Build GRCh38
rs761071028 Chr.8: 54048073-54048073 on Build GRCh38
rs903711503 Chr.8: 54047862-54047862 on Build GRCh38
rs184876816 Chr.8: 54048146-54048146 on Build GRCh38
rs760960813 Chr.8: 54048157-54048157 on Build GRCh38

View Full Product Details