Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 54100829-54101344 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LYPLA1
Entrez Gene ID
10434
Gene Name
lysophospholipase I
Gene Aliases
APT-1, APT1, LPL-I, LPL1, hAPT1
UniGene
Hs.744046
Gene Chromosome Location
Chr.8: 54046367-54102017 on Build GRCh38
Transcripts
NM_001279356.1 NM_001279357.1 NM_001279358.1 NM_001279359.1 NM_001279360.1 NM_006330.3 XM_005251127.4 XM_017012956.1 XM_017012957.1 XM_017012958.1 XM_017012959.1 XM_017012960.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5787157 c.80T>G p.L27R Chr.8: 54100929-54100929 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs756705739 Chr.8: 54100936-54100936 on Build GRCh38
rs912295717 Chr.8: 54101261-54101261 on Build GRCh38
rs532305601 Chr.8: 54100864-54100864 on Build GRCh38
rs978173866 Chr.8: 54101312-54101312 on Build GRCh38
rs974667188 Chr.8: 54100896-54100896 on Build GRCh38
rs967098339 Chr.8: 54101122-54101122 on Build GRCh38
rs368673068 Chr.8: 54101155-54101156 on Build GRCh38
rs77029186 Chr.8: 54101131-54101131 on Build GRCh38
rs375108228 Chr.8: 54101262-54101262 on Build GRCh38
rs1003268958 Chr.8: 54101163-54101163 on Build GRCh38
rs761230161 Chr.8: 54100915-54100915 on Build GRCh38
rs779952366 Chr.8: 54100960-54100960 on Build GRCh38
rs568562214 Chr.8: 54101149-54101149 on Build GRCh38
rs902446766 Chr.8: 54101084-54101084 on Build GRCh38
rs1018210418 Chr.8: 54100996-54100996 on Build GRCh38
rs112018444 Chr.8: 54100943-54100943 on Build GRCh38
rs909340946 Chr.8: 54100931-54100931 on Build GRCh38
rs965132652 Chr.8: 54101303-54101303 on Build GRCh38
rs777926565 Chr.8: 54100975-54100976 on Build GRCh38
rs981753059 Chr.8: 54100862-54100862 on Build GRCh38
rs958376307 Chr.8: 54101153-54101153 on Build GRCh38
rs577593234 Chr.8: 54101314-54101314 on Build GRCh38
rs551930867 Chr.8: 54101007-54101007 on Build GRCh38
rs775713811 Chr.8: 54100898-54100898 on Build GRCh38
rs892920645 Chr.8: 54101031-54101031 on Build GRCh38
rs764564243 Chr.8: 54100928-54100928 on Build GRCh38
rs751286748 Chr.8: 54100971-54100971 on Build GRCh38
rs146719663 Chr.8: 54100908-54100908 on Build GRCh38
rs759155731 Chr.8: 54101146-54101146 on Build GRCh38
rs1053558775 Chr.8: 54101115-54101115 on Build GRCh38
rs755002247 Chr.8: 54100981-54100981 on Build GRCh38
rs757979477 Chr.8: 54100956-54100956 on Build GRCh38
rs961593407 Chr.8: 54100888-54100888 on Build GRCh38
rs533906797 Chr.8: 54101182-54101182 on Build GRCh38
rs936572501 Chr.8: 54101114-54101114 on Build GRCh38
rs1035303622 Chr.8: 54101146-54101146 on Build GRCh38
rs1051749947 Chr.8: 54100967-54100967 on Build GRCh38
rs370147043 Chr.8: 54100876-54100876 on Build GRCh38
rs991729563 Chr.8: 54100875-54100875 on Build GRCh38
rs889150709 Chr.8: 54100982-54100982 on Build GRCh38
rs748217664 Chr.8: 54100901-54100904 on Build GRCh38
rs933432665 Chr.8: 54100966-54100966 on Build GRCh38
rs995731160 Chr.8: 54101206-54101206 on Build GRCh38
rs909475884 Chr.8: 54101183-54101183 on Build GRCh38
rs772774508 Chr.8: 54101212-54101213 on Build GRCh38
rs943688937 Chr.8: 54101036-54101036 on Build GRCh38
rs537938505 Chr.8: 54101065-54101065 on Build GRCh38
rs747266730 Chr.8: 54100978-54100979 on Build GRCh38
rs912263529 Chr.8: 54101002-54101002 on Build GRCh38
rs972257769 Chr.8: 54101306-54101306 on Build GRCh38
rs750024013 Chr.8: 54100945-54100945 on Build GRCh38
rs1015574113 Chr.8: 54101164-54101164 on Build GRCh38
rs753376011 Chr.8: 54100934-54100934 on Build GRCh38
rs894943477 Chr.8: 54101118-54101119 on Build GRCh38
rs985234725 Chr.8: 54101252-54101252 on Build GRCh38
rs183462124 Chr.8: 54101042-54101042 on Build GRCh38
rs758510422 Chr.8: 54100944-54100948 on Build GRCh38
rs1036823089 Chr.8: 54100948-54100948 on Build GRCh38
rs1006784379 Chr.8: 54100991-54100991 on Build GRCh38
rs954184721 Chr.8: 54101220-54101220 on Build GRCh38
rs35405582 Chr.8: 54101145-54101146 on Build GRCh38
rs768889395 Chr.8: 54100942-54100942 on Build GRCh38

View Full Product Details