Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 56959183-56959707 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IMPAD1
Entrez Gene ID
54928
Gene Name
inositol monophosphatase domain containing 1
Gene Aliases
GPAPP, IMP 3, IMP-3, IMPA3
UniGene
Hs.438689
Gene Chromosome Location
Chr.8: 56957929-56993874 on Build GRCh38
Transcripts
NM_017813.4

SNP Detail

SNP ID Chromosome Location
rs1044557329 Chr.8: 56959617-56959617 on Build GRCh38
rs996618256 Chr.8: 56959575-56959575 on Build GRCh38
rs954897875 Chr.8: 56959331-56959331 on Build GRCh38
rs778597453 Chr.8: 56959344-56959344 on Build GRCh38
rs552306119 Chr.8: 56959448-56959448 on Build GRCh38
rs1396116 Chr.8: 56959584-56959584 on Build GRCh38
rs576225144 Chr.8: 56959229-56959229 on Build GRCh38
rs1032742408 Chr.8: 56959257-56959257 on Build GRCh38
rs892814404 Chr.8: 56959594-56959594 on Build GRCh38
rs756774533 Chr.8: 56959534-56959537 on Build GRCh38
rs573606888 Chr.8: 56959358-56959358 on Build GRCh38
rs949902553 Chr.8: 56959487-56959487 on Build GRCh38
rs1046901858 Chr.8: 56959510-56959510 on Build GRCh38
rs907339742 Chr.8: 56959633-56959633 on Build GRCh38
rs912968611 Chr.8: 56959354-56959354 on Build GRCh38
rs927729322 Chr.8: 56959509-56959509 on Build GRCh38
rs998548366 Chr.8: 56959213-56959213 on Build GRCh38
rs117323870 Chr.8: 56959328-56959328 on Build GRCh38
rs1026550136 Chr.8: 56959676-56959676 on Build GRCh38
rs561898077 Chr.8: 56959297-56959297 on Build GRCh38
rs988066533 Chr.8: 56959348-56959348 on Build GRCh38
rs557759212 Chr.8: 56959529-56959529 on Build GRCh38
rs1396117 Chr.8: 56959363-56959363 on Build GRCh38
rs184464893 Chr.8: 56959314-56959314 on Build GRCh38
rs974808326 Chr.8: 56959460-56959460 on Build GRCh38
rs1020383261 Chr.8: 56959643-56959643 on Build GRCh38
rs369907391 Chr.8: 56959301-56959301 on Build GRCh38
rs902818471 Chr.8: 56959212-56959212 on Build GRCh38
rs143068938 Chr.8: 56959668-56959670 on Build GRCh38
rs1004984449 Chr.8: 56959280-56959280 on Build GRCh38
rs955157097 Chr.8: 56959627-56959627 on Build GRCh38
rs934310775 Chr.8: 56959558-56959558 on Build GRCh38
rs921081409 Chr.8: 56959462-56959462 on Build GRCh38
rs549676621 Chr.8: 56959292-56959292 on Build GRCh38
rs950734919 Chr.8: 56959387-56959387 on Build GRCh38
rs974988569 Chr.8: 56959308-56959308 on Build GRCh38
rs778291867 Chr.8: 56959449-56959449 on Build GRCh38
rs984782330 Chr.8: 56959401-56959401 on Build GRCh38
rs534618768 Chr.8: 56959427-56959427 on Build GRCh38
rs543846823 Chr.8: 56959478-56959478 on Build GRCh38
rs568359173 Chr.8: 56959592-56959592 on Build GRCh38
rs548002917 Chr.8: 56959542-56959542 on Build GRCh38
rs899156227 Chr.8: 56959543-56959544 on Build GRCh38
rs886959541 Chr.8: 56959521-56959521 on Build GRCh38
rs1003482729 Chr.8: 56959661-56959661 on Build GRCh38
rs937039816 Chr.8: 56959377-56959377 on Build GRCh38
rs1012632566 Chr.8: 56959595-56959595 on Build GRCh38
rs1026997378 Chr.8: 56959376-56959376 on Build GRCh38

View Full Product Details