Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 60280661-60281153 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CA8
Entrez Gene ID
767
Gene Name
carbonic anhydrase 8
Gene Aliases
CA-RP, CA-VIII, CALS, CAMRQ3, CARP
UniGene
Hs.654388
Gene Chromosome Location
Chr.8: 60185420-60281423 on Build GRCh38
Transcripts
NM_001321837.1 NM_001321838.1 NM_001321839.1 NM_004056.5 NR_135821.1 XM_011517587.2 XM_011517588.2 XM_017013818.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4928051 c.24A>T p.E8D Chr.8: 60281124-60281124 on Build GRCh38
6113629 c.83A>T p.E28V Chr.8: 60281065-60281065 on Build GRCh38
4889996 c.76G>T p.G26C Chr.8: 60281072-60281072 on Build GRCh38
4561802 c.89G>A p.G30D Chr.8: 60281059-60281059 on Build GRCh38
1199147 c.2T>C p.M1T Chr.8: 60281146-60281146 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs570574480 Chr.8: 60280780-60280780 on Build GRCh38
rs952430236 Chr.8: 60280870-60280870 on Build GRCh38
rs970893645 Chr.8: 60280744-60280744 on Build GRCh38
rs775106662 Chr.8: 60281036-60281036 on Build GRCh38
rs541122984 Chr.8: 60280930-60280930 on Build GRCh38
rs760270516 Chr.8: 60281118-60281118 on Build GRCh38
rs753467239 Chr.8: 60281051-60281051 on Build GRCh38
rs1024004845 Chr.8: 60280883-60280883 on Build GRCh38
rs1026815869 Chr.8: 60281054-60281054 on Build GRCh38
rs961710660 Chr.8: 60280861-60280862 on Build GRCh38
rs528318434 Chr.8: 60281106-60281106 on Build GRCh38
rs749485766 Chr.8: 60281095-60281095 on Build GRCh38
rs1033840531 Chr.8: 60280908-60280908 on Build GRCh38
rs116144151 Chr.8: 60280919-60280919 on Build GRCh38
rs945386763 Chr.8: 60280782-60280782 on Build GRCh38
rs913388989 Chr.8: 60280744-60280744 on Build GRCh38
rs10086200 Chr.8: 60281029-60281029 on Build GRCh38
rs111928693 Chr.8: 60280824-60280824 on Build GRCh38
rs1049545217 Chr.8: 60280822-60280822 on Build GRCh38
rs1043084160 Chr.8: 60280787-60280787 on Build GRCh38
rs752215372 Chr.8: 60280708-60280708 on Build GRCh38
rs771259928 Chr.8: 60281020-60281020 on Build GRCh38
rs888303295 Chr.8: 60281059-60281059 on Build GRCh38
rs896796415 Chr.8: 60280845-60280845 on Build GRCh38
rs113572519 Chr.8: 60280837-60280837 on Build GRCh38
rs752760505 Chr.8: 60281087-60281087 on Build GRCh38
rs1001608474 Chr.8: 60280810-60280810 on Build GRCh38
rs746415479 Chr.8: 60281107-60281107 on Build GRCh38
rs553144429 Chr.8: 60280935-60280935 on Build GRCh38
rs945676839 Chr.8: 60280936-60280936 on Build GRCh38
rs1055915269 Chr.8: 60281001-60281001 on Build GRCh38
rs147962541 Chr.8: 60281060-60281060 on Build GRCh38
rs760466887 Chr.8: 60281043-60281043 on Build GRCh38
rs994117516 Chr.8: 60281030-60281030 on Build GRCh38
rs763264016 Chr.8: 60281046-60281047 on Build GRCh38
rs528552604 Chr.8: 60280781-60280781 on Build GRCh38
rs1015927157 Chr.8: 60280842-60280842 on Build GRCh38
rs1018035887 Chr.8: 60281076-60281076 on Build GRCh38
rs7002846 Chr.8: 60280894-60280894 on Build GRCh38
rs370751267 Chr.8: 60281014-60281014 on Build GRCh38
rs139012377 Chr.8: 60280914-60280914 on Build GRCh38
rs926801546 Chr.8: 60280754-60280754 on Build GRCh38
rs115668655 Chr.8: 60280731-60280731 on Build GRCh38
rs904625087 Chr.8: 60280801-60280801 on Build GRCh38
rs141820235 Chr.8: 60281067-60281067 on Build GRCh38
rs756129376 Chr.8: 60281092-60281092 on Build GRCh38
rs896965326 Chr.8: 60280834-60280834 on Build GRCh38
rs1049806636 Chr.8: 60281009-60281009 on Build GRCh38
rs1041322636 Chr.8: 60280740-60280740 on Build GRCh38
rs115885226 Chr.8: 60281082-60281082 on Build GRCh38
rs545165610 Chr.8: 60280817-60280817 on Build GRCh38
rs776173702 Chr.8: 60281128-60281128 on Build GRCh38
rs961341320 Chr.8: 60280719-60280719 on Build GRCh38
rs552367128 Chr.8: 60280764-60280764 on Build GRCh38
rs777862901 Chr.8: 60281093-60281093 on Build GRCh38
rs1005389479 Chr.8: 60280825-60280825 on Build GRCh38
rs895841063 Chr.8: 60280815-60280815 on Build GRCh38
rs7003287 Chr.8: 60280967-60280967 on Build GRCh38
rs763820905 Chr.8: 60281050-60281050 on Build GRCh38
rs914152259 Chr.8: 60280925-60280925 on Build GRCh38
rs775962746 Chr.8: 60281022-60281022 on Build GRCh38
rs7002871 Chr.8: 60280963-60280963 on Build GRCh38
rs758301801 Chr.8: 60281079-60281079 on Build GRCh38
rs781459925 Chr.8: 60280926-60280926 on Build GRCh38
rs1041106092 Chr.8: 60280788-60280788 on Build GRCh38
rs563606477 Chr.8: 60280691-60280691 on Build GRCh38
rs546682461 Chr.8: 60281114-60281114 on Build GRCh38
rs977138868 Chr.8: 60280960-60280960 on Build GRCh38
rs143254961 Chr.8: 60280811-60280811 on Build GRCh38
rs74792169 Chr.8: 60281011-60281011 on Build GRCh38
rs112802688 Chr.8: 60280888-60280888 on Build GRCh38
rs573505037 Chr.8: 60280924-60280924 on Build GRCh38
rs535886092 Chr.8: 60280835-60280835 on Build GRCh38
rs946237736 Chr.8: 60280770-60280770 on Build GRCh38
rs1026699952 Chr.8: 60280847-60280847 on Build GRCh38
rs771220749 Chr.8: 60281097-60281097 on Build GRCh38
rs768176485 Chr.8: 60281121-60281121 on Build GRCh38
rs545816687 Chr.8: 60281078-60281078 on Build GRCh38
rs771255096 Chr.8: 60280901-60280901 on Build GRCh38
rs1015203984 Chr.8: 60280712-60280712 on Build GRCh38

View Full Product Details