Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 66128165-66128682 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TRIM55
Entrez Gene ID
84675
Gene Name
tripartite motif containing 55
Gene Aliases
MURF-2, RNF29, muRF2
UniGene
Hs.85524
Gene Chromosome Location
Chr.8: 66112667-66175485 on Build GRCh38
Transcripts
NM_033058.2 NM_184085.1 NM_184086.1 NM_184087.1 XM_005251316.4 XM_011517613.2 XM_011517614.2 XM_011517615.2 XM_011517616.2 XM_011517617.2 XM_017013908.1 XM_017013909.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6808403 c.176A>T p.N59I Chr.8: 66128311-66128311 on Build GRCh38
1314146 c.243T>C p.C81C Chr.8: 66128378-66128378 on Build GRCh38
2720806 c.289A>T p.R97W Chr.8: 66128424-66128424 on Build GRCh38
3901005 c.294C>A p.N98K Chr.8: 66128429-66128429 on Build GRCh38
5824572 c.182A>T p.Y61F Chr.8: 66128317-66128317 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs557767199 Chr.8: 66128304-66128304 on Build GRCh38
rs267601968 Chr.8: 66128313-66128313 on Build GRCh38
rs61760894 Chr.8: 66128314-66128314 on Build GRCh38
rs765212280 Chr.8: 66128455-66128455 on Build GRCh38
rs1007027211 Chr.8: 66128555-66128555 on Build GRCh38
rs773648869 Chr.8: 66128294-66128294 on Build GRCh38
rs769797275 Chr.8: 66128370-66128370 on Build GRCh38
rs755916724 Chr.8: 66128525-66128528 on Build GRCh38
rs748217714 Chr.8: 66128272-66128272 on Build GRCh38
rs952339487 Chr.8: 66128602-66128602 on Build GRCh38
rs527708406 Chr.8: 66128619-66128619 on Build GRCh38
rs781375784 Chr.8: 66128363-66128363 on Build GRCh38
rs542813899 Chr.8: 66128529-66128529 on Build GRCh38
rs749380101 Chr.8: 66128373-66128373 on Build GRCh38
rs752870512 Chr.8: 66128489-66128489 on Build GRCh38
rs777856332 Chr.8: 66128372-66128372 on Build GRCh38
rs562384365 Chr.8: 66128553-66128553 on Build GRCh38
rs778763696 Chr.8: 66128350-66128350 on Build GRCh38
rs531408822 Chr.8: 66128365-66128365 on Build GRCh38
rs760266717 Chr.8: 66128640-66128640 on Build GRCh38
rs560212457 Chr.8: 66128432-66128432 on Build GRCh38
rs556539321 Chr.8: 66128221-66128221 on Build GRCh38
rs551289228 Chr.8: 66128561-66128561 on Build GRCh38
rs918790354 Chr.8: 66128581-66128581 on Build GRCh38
rs571273603 Chr.8: 66128562-66128562 on Build GRCh38
rs774787420 Chr.8: 66128391-66128391 on Build GRCh38
rs763549562 Chr.8: 66128299-66128299 on Build GRCh38
rs758127948 Chr.8: 66128472-66128472 on Build GRCh38
rs900897396 Chr.8: 66128573-66128573 on Build GRCh38
rs576802562 Chr.8: 66128231-66128231 on Build GRCh38
rs781428841 Chr.8: 66128264-66128264 on Build GRCh38
rs750201649 Chr.8: 66128358-66128358 on Build GRCh38
rs61760893 Chr.8: 66128310-66128310 on Build GRCh38
rs759862288 Chr.8: 66128315-66128315 on Build GRCh38
rs539857288 Chr.8: 66128496-66128496 on Build GRCh38
rs536857565 Chr.8: 66128220-66128220 on Build GRCh38
rs763703907 Chr.8: 66128329-66128329 on Build GRCh38
rs949733590 Chr.8: 66128245-66128245 on Build GRCh38
rs750151505 Chr.8: 66128481-66128488 on Build GRCh38
rs958562682 Chr.8: 66128387-66128387 on Build GRCh38
rs767274404 Chr.8: 66128322-66128322 on Build GRCh38
rs766164808 Chr.8: 66128479-66128479 on Build GRCh38
rs756309337 Chr.8: 66128493-66128493 on Build GRCh38
rs905786270 Chr.8: 66128291-66128291 on Build GRCh38
rs1018198709 Chr.8: 66128550-66128550 on Build GRCh38
rs1025428930 Chr.8: 66128583-66128583 on Build GRCh38
rs773909283 Chr.8: 66128214-66128214 on Build GRCh38
rs918718240 Chr.8: 66128211-66128211 on Build GRCh38
rs16932535 Chr.8: 66128234-66128234 on Build GRCh38
rs752456346 Chr.8: 66128326-66128326 on Build GRCh38
rs753676524 Chr.8: 66128340-66128340 on Build GRCh38
rs374673784 Chr.8: 66128498-66128498 on Build GRCh38
rs750382058 Chr.8: 66128468-66128468 on Build GRCh38
rs369600990 Chr.8: 66128554-66128554 on Build GRCh38
rs766497610 Chr.8: 66128191-66128191 on Build GRCh38
rs769916169 Chr.8: 66128283-66128283 on Build GRCh38
rs1005259662 Chr.8: 66128604-66128604 on Build GRCh38
rs766143405 Chr.8: 66128524-66128525 on Build GRCh38
rs746368387 Chr.8: 66128516-66128516 on Build GRCh38
rs746230423 Chr.8: 66128396-66128396 on Build GRCh38
rs868686347 Chr.8: 66128476-66128476 on Build GRCh38
rs775217398 Chr.8: 66128423-66128423 on Build GRCh38
rs760207046 Chr.8: 66128328-66128328 on Build GRCh38
rs974221350 Chr.8: 66128195-66128195 on Build GRCh38
rs191183013 Chr.8: 66128215-66128215 on Build GRCh38
rs983245383 Chr.8: 66128603-66128603 on Build GRCh38
rs374101872 Chr.8: 66128273-66128273 on Build GRCh38
rs367993776 Chr.8: 66128347-66128347 on Build GRCh38
rs771441970 Chr.8: 66128388-66128388 on Build GRCh38
rs1047929211 Chr.8: 66128216-66128216 on Build GRCh38
rs772579473 Chr.8: 66128403-66128403 on Build GRCh38
rs758190155 Chr.8: 66128359-66128359 on Build GRCh38
rs776182574 Chr.8: 66128430-66128430 on Build GRCh38
rs368217551 Chr.8: 66128257-66128257 on Build GRCh38
rs779422045 Chr.8: 66128506-66128506 on Build GRCh38
rs772528133 Chr.8: 66128522-66128522 on Build GRCh38
rs917005036 Chr.8: 66128605-66128605 on Build GRCh38
rs372129244 Chr.8: 66128258-66128258 on Build GRCh38
rs930123479 Chr.8: 66128597-66128597 on Build GRCh38
rs768900297 Chr.8: 66128262-66128262 on Build GRCh38
rs760433737 Chr.8: 66128429-66128429 on Build GRCh38
rs1054021575 Chr.8: 66128552-66128552 on Build GRCh38
rs371406893 Chr.8: 66128444-66128444 on Build GRCh38

View Full Product Details