Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 67433993-67434517 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CPA6
Entrez Gene ID
57094
Gene Name
carboxypeptidase A6
Gene Aliases
CPAH, ETL5, FEB11
UniGene
Hs.658850
Gene Chromosome Location
Chr.8: 67422170-67747114 on Build GRCh38
Transcripts
NM_020361.4 XM_011517570.2 XM_017013646.1 NR_136224.1

Gene Symbol
LOC102724708
Entrez Gene ID
102724708
Gene Name
uncharacterized LOC102724708
Gene Aliases
-
UniGene
Hs.636330
Gene Chromosome Location
Chr.8: 67343834-67491868 on Build GRCh38
Transcripts
NM_020361.4 XM_011517570.2 XM_017013646.1 NR_136224.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6574829 c.909G>C p.V303V Chr.8: 67434170-67434170 on Build GRCh38
5358615 c.1004C>A p.S335Y Chr.8: 67434075-67434075 on Build GRCh38
454777 c.873A>G p.T291T Chr.8: 67434206-67434206 on Build GRCh38
5069260 c.945G>A p.K315K Chr.8: 67434134-67434134 on Build GRCh38
3650384 c.927C>T p.F309F Chr.8: 67434152-67434152 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs138279913 Chr.8: 67434176-67434176 on Build GRCh38
rs1023381016 Chr.8: 67434354-67434354 on Build GRCh38
rs756925537 Chr.8: 67434186-67434186 on Build GRCh38
rs149196398 Chr.8: 67434134-67434134 on Build GRCh38
rs147046973 Chr.8: 67434163-67434163 on Build GRCh38
rs762112759 Chr.8: 67434047-67434047 on Build GRCh38
rs1050905016 Chr.8: 67434345-67434345 on Build GRCh38
rs587780322 Chr.8: 67434151-67434151 on Build GRCh38
rs754796419 Chr.8: 67434229-67434229 on Build GRCh38
rs766177388 Chr.8: 67434058-67434058 on Build GRCh38
rs551174964 Chr.8: 67434188-67434188 on Build GRCh38
rs143107403 Chr.8: 67434159-67434159 on Build GRCh38
rs767858182 Chr.8: 67434160-67434160 on Build GRCh38
rs749823127 Chr.8: 67434316-67434316 on Build GRCh38
rs993930163 Chr.8: 67434293-67434293 on Build GRCh38
rs146662107 Chr.8: 67434037-67434037 on Build GRCh38
rs761221690 Chr.8: 67434264-67434264 on Build GRCh38
rs978823751 Chr.8: 67434437-67434437 on Build GRCh38
rs748738808 Chr.8: 67434016-67434016 on Build GRCh38
rs140734555 Chr.8: 67434055-67434055 on Build GRCh38
rs143321447 Chr.8: 67434147-67434147 on Build GRCh38
rs1029891045 Chr.8: 67434076-67434076 on Build GRCh38
rs752081103 Chr.8: 67434065-67434065 on Build GRCh38
rs774363423 Chr.8: 67434157-67434157 on Build GRCh38
rs771960743 Chr.8: 67434117-67434117 on Build GRCh38
rs772224868 Chr.8: 67434249-67434249 on Build GRCh38
rs780095436 Chr.8: 67434224-67434224 on Build GRCh38
rs372964579 Chr.8: 67434053-67434053 on Build GRCh38
rs139145929 Chr.8: 67434148-67434148 on Build GRCh38
rs764435777 Chr.8: 67434267-67434267 on Build GRCh38
rs764788773 Chr.8: 67434205-67434205 on Build GRCh38
rs775929414 Chr.8: 67434263-67434263 on Build GRCh38
rs1004057331 Chr.8: 67434361-67434361 on Build GRCh38
rs117556189 Chr.8: 67434347-67434347 on Build GRCh38
rs749845096 Chr.8: 67434207-67434207 on Build GRCh38
rs375456613 Chr.8: 67434273-67434273 on Build GRCh38
rs138313759 Chr.8: 67434104-67434104 on Build GRCh38
rs775212726 Chr.8: 67434042-67434042 on Build GRCh38
rs778325977 Chr.8: 67434113-67434113 on Build GRCh38
rs771091901 Chr.8: 67434031-67434034 on Build GRCh38
rs779209758 Chr.8: 67434452-67434452 on Build GRCh38
rs768240710 Chr.8: 67434483-67434483 on Build GRCh38
rs553327896 Chr.8: 67434122-67434122 on Build GRCh38
rs752773496 Chr.8: 67434221-67434221 on Build GRCh38
rs745811365 Chr.8: 67434114-67434114 on Build GRCh38
rs766388803 Chr.8: 67434060-67434060 on Build GRCh38
rs552521766 Chr.8: 67434247-67434247 on Build GRCh38
rs749519696 Chr.8: 67434235-67434235 on Build GRCh38
rs889576352 Chr.8: 67434189-67434189 on Build GRCh38
rs200622875 Chr.8: 67434015-67434015 on Build GRCh38
rs961314522 Chr.8: 67434052-67434052 on Build GRCh38
rs7837952 Chr.8: 67434458-67434458 on Build GRCh38
rs777929597 Chr.8: 67434296-67434296 on Build GRCh38
rs762251943 Chr.8: 67434280-67434280 on Build GRCh38
rs771118654 Chr.8: 67434241-67434241 on Build GRCh38
rs770433020 Chr.8: 67434020-67434020 on Build GRCh38
rs960870464 Chr.8: 67434325-67434325 on Build GRCh38
rs371199710 Chr.8: 67434226-67434226 on Build GRCh38
rs757929205 Chr.8: 67434217-67434217 on Build GRCh38
rs978961467 Chr.8: 67434140-67434140 on Build GRCh38
rs898435961 Chr.8: 67434426-67434426 on Build GRCh38
rs541703726 Chr.8: 67434424-67434424 on Build GRCh38
rs753122064 Chr.8: 67434081-67434081 on Build GRCh38
rs756485062 Chr.8: 67434088-67434088 on Build GRCh38
rs756744382 Chr.8: 67434105-67434105 on Build GRCh38
rs34407100 Chr.8: 67434052-67434053 on Build GRCh38
rs756289250 Chr.8: 67434242-67434242 on Build GRCh38
rs1038189097 Chr.8: 67434254-67434254 on Build GRCh38
rs150477940 Chr.8: 67434080-67434080 on Build GRCh38
rs372446556 Chr.8: 67434155-67434155 on Build GRCh38
rs777340067 Chr.8: 67434102-67434102 on Build GRCh38
rs36063992 Chr.8: 67434079-67434080 on Build GRCh38
rs778872244 Chr.8: 67434025-67434025 on Build GRCh38
rs545504815 Chr.8: 67434232-67434232 on Build GRCh38
rs970208942 Chr.8: 67434365-67434365 on Build GRCh38
rs768598882 Chr.8: 67434142-67434142 on Build GRCh38
rs753468111 Chr.8: 67434177-67434177 on Build GRCh38
rs760988775 Chr.8: 67434166-67434166 on Build GRCh38
rs781569255 Chr.8: 67434060-67434061 on Build GRCh38
rs375860856 Chr.8: 67434079-67434079 on Build GRCh38

View Full Product Details