Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 71198127-71198641 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EYA1
Entrez Gene ID
2138
Gene Name
EYA transcriptional coactivator and phosphatase 1
Gene Aliases
BOP, BOR, BOS1, OFC1
UniGene
Hs.444971
Gene Chromosome Location
Chr.8: 71197433-71548130 on Build GRCh38
Transcripts
NM_000503.5 NM_001288574.1 NM_001288575.1 NM_172058.3 NM_172059.3 NM_172060.3 XM_011517483.2 XM_011517484.2 XM_017013201.1 XM_017013202.1 XM_017013203.1 XM_017013204.1 XM_017013206.1 XM_017013207.1 XM_017013208.1 XM_017013209.1 XM_017013210.1 XM_017013211.1 XM_017013212.1 XM_017013213.1

SNP Detail

SNP ID Chromosome Location
rs200553501 Chr.8: 71198167-71198171 on Build GRCh38
rs890237582 Chr.8: 71198566-71198576 on Build GRCh38
rs751951072 Chr.8: 71198428-71198428 on Build GRCh38
rs900048085 Chr.8: 71198154-71198154 on Build GRCh38
rs371488223 Chr.8: 71198566-71198567 on Build GRCh38
rs762732793 Chr.8: 71198432-71198432 on Build GRCh38
rs900283982 Chr.8: 71198436-71198436 on Build GRCh38
rs752343925 Chr.8: 71198439-71198439 on Build GRCh38
rs975576164 Chr.8: 71198495-71198495 on Build GRCh38
rs929997002 Chr.8: 71198533-71198533 on Build GRCh38
rs896440972 Chr.8: 71198615-71198615 on Build GRCh38
rs183978164 Chr.8: 71198212-71198212 on Build GRCh38
rs1052091244 Chr.8: 71198603-71198603 on Build GRCh38
rs73684710 Chr.8: 71198301-71198301 on Build GRCh38
rs187719222 Chr.8: 71198511-71198511 on Build GRCh38
rs991971276 Chr.8: 71198204-71198204 on Build GRCh38
rs1021985839 Chr.8: 71198460-71198460 on Build GRCh38
rs974549319 Chr.8: 71198522-71198522 on Build GRCh38
rs1030870747 Chr.8: 71198193-71198193 on Build GRCh38
rs948126238 Chr.8: 71198228-71198229 on Build GRCh38
rs974138582 Chr.8: 71198228-71198228 on Build GRCh38
rs1017006534 Chr.8: 71198332-71198332 on Build GRCh38
rs919992165 Chr.8: 71198523-71198523 on Build GRCh38
rs930270938 Chr.8: 71198294-71198294 on Build GRCh38
rs964821408 Chr.8: 71198493-71198493 on Build GRCh38
rs1020849426 Chr.8: 71198458-71198458 on Build GRCh38
rs960544603 Chr.8: 71198200-71198200 on Build GRCh38
rs903580936 Chr.8: 71198592-71198592 on Build GRCh38
rs74862574 Chr.8: 71198540-71198540 on Build GRCh38
rs903505902 Chr.8: 71198465-71198465 on Build GRCh38
rs1002003805 Chr.8: 71198374-71198374 on Build GRCh38
rs887125661 Chr.8: 71198313-71198313 on Build GRCh38
rs570995924 Chr.8: 71198438-71198439 on Build GRCh38
rs866497958 Chr.8: 71198356-71198356 on Build GRCh38
rs899981376 Chr.8: 71198558-71198559 on Build GRCh38
rs1030944146 Chr.8: 71198485-71198485 on Build GRCh38
rs898609785 Chr.8: 71198347-71198347 on Build GRCh38
rs891355243 Chr.8: 71198372-71198372 on Build GRCh38
rs999197071 Chr.8: 71198598-71198598 on Build GRCh38
rs1051066669 Chr.8: 71198227-71198227 on Build GRCh38
rs1047314149 Chr.8: 71198330-71198330 on Build GRCh38
rs992024363 Chr.8: 71198498-71198498 on Build GRCh38
rs563687259 Chr.8: 71198473-71198473 on Build GRCh38
rs77243350 Chr.8: 71198556-71198556 on Build GRCh38
rs940030319 Chr.8: 71198601-71198601 on Build GRCh38
rs886063085 Chr.8: 71198201-71198202 on Build GRCh38
rs766352837 Chr.8: 71198463-71198463 on Build GRCh38
rs1039814492 Chr.8: 71198429-71198429 on Build GRCh38
rs979538866 Chr.8: 71198474-71198474 on Build GRCh38
rs764200686 Chr.8: 71198582-71198582 on Build GRCh38
rs775498240 Chr.8: 71198492-71198492 on Build GRCh38
rs530293492 Chr.8: 71198320-71198320 on Build GRCh38
rs375646172 Chr.8: 71198249-71198250 on Build GRCh38
rs774873177 Chr.8: 71198548-71198548 on Build GRCh38
rs1004647052 Chr.8: 71198314-71198314 on Build GRCh38
rs115794887 Chr.8: 71198397-71198397 on Build GRCh38
rs532364833 Chr.8: 71198499-71198499 on Build GRCh38
rs73284340 Chr.8: 71198539-71198539 on Build GRCh38
rs919900872 Chr.8: 71198243-71198243 on Build GRCh38
rs796305539 Chr.8: 71198243-71198244 on Build GRCh38

View Full Product Details