Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 73008987-73009521 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TERF1
Entrez Gene ID
7013
Gene Name
telomeric repeat binding factor 1
Gene Aliases
PIN2, TRBF1, TRF, TRF1, hTRF1-AS, t-TRF1
UniGene
Hs.442707
Gene Chromosome Location
Chr.8: 73008862-73047752 on Build GRCh38
Transcripts
NM_003218.3 NM_017489.2 XM_005251291.3 XM_005251292.3 XM_011517582.2 XM_017013792.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3395401 c.230G>T p.W77L Chr.8: 73009116-73009116 on Build GRCh38
3432553 c.234G>A p.M78I Chr.8: 73009120-73009120 on Build GRCh38
2875325 c.200_201insGGCCGA p.A72_V73insEA Chr.8: 73009086-73009087 on Build GRCh38
2875329 c.310A>T p.S104C Chr.8: 73009196-73009196 on Build GRCh38
3650814 c.106G>A p.E36K Chr.8: 73008992-73008992 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs960020069 Chr.8: 73009211-73009211 on Build GRCh38
rs748891923 Chr.8: 73009141-73009144 on Build GRCh38
rs544688936 Chr.8: 73009246-73009246 on Build GRCh38
rs753391990 Chr.8: 73009062-73009062 on Build GRCh38
rs777764676 Chr.8: 73009007-73009007 on Build GRCh38
rs750911781 Chr.8: 73009049-73009057 on Build GRCh38
rs754605256 Chr.8: 73009239-73009240 on Build GRCh38
rs1054065269 Chr.8: 73009433-73009433 on Build GRCh38
rs757354613 Chr.8: 73009215-73009215 on Build GRCh38
rs746722487 Chr.8: 73009136-73009136 on Build GRCh38
rs749862674 Chr.8: 73009017-73009017 on Build GRCh38
rs574649908 Chr.8: 73009324-73009324 on Build GRCh38
rs1035571208 Chr.8: 73009201-73009201 on Build GRCh38
rs1035456603 Chr.8: 73009453-73009453 on Build GRCh38
rs200166029 Chr.8: 73009197-73009197 on Build GRCh38
rs201816582 Chr.8: 73009302-73009303 on Build GRCh38
rs188756616 Chr.8: 73009487-73009487 on Build GRCh38
rs768464397 Chr.8: 73009243-73009243 on Build GRCh38
rs113373855 Chr.8: 73009299-73009300 on Build GRCh38
rs746370012 Chr.8: 73009225-73009225 on Build GRCh38
rs1019406888 Chr.8: 73009241-73009241 on Build GRCh38
rs1030703815 Chr.8: 73009332-73009332 on Build GRCh38
rs771416590 Chr.8: 73009021-73009021 on Build GRCh38
rs944266673 Chr.8: 73009368-73009368 on Build GRCh38
rs374169616 Chr.8: 73009194-73009194 on Build GRCh38
rs751939129 Chr.8: 73009049-73009054 on Build GRCh38
rs373985315 Chr.8: 73009024-73009024 on Build GRCh38
rs759792293 Chr.8: 73009037-73009037 on Build GRCh38
rs752867787 Chr.8: 73009084-73009084 on Build GRCh38
rs777889745 Chr.8: 73009095-73009095 on Build GRCh38
rs374662171 Chr.8: 73009189-73009189 on Build GRCh38
rs577204133 Chr.8: 73009229-73009229 on Build GRCh38
rs527993664 Chr.8: 73009421-73009421 on Build GRCh38
rs998640494 Chr.8: 73009419-73009419 on Build GRCh38
rs55882538 Chr.8: 73009254-73009254 on Build GRCh38
rs916365839 Chr.8: 73009318-73009318 on Build GRCh38
rs779841708 Chr.8: 73009078-73009078 on Build GRCh38
rs991398248 Chr.8: 73009496-73009496 on Build GRCh38
rs747357861 Chr.8: 73009240-73009240 on Build GRCh38
rs894879967 Chr.8: 73009020-73009020 on Build GRCh38
rs556421563 Chr.8: 73009290-73009290 on Build GRCh38
rs3832552 Chr.8: 73009072-73009073 on Build GRCh38
rs769191042 Chr.8: 73009252-73009252 on Build GRCh38
rs764711833 Chr.8: 73009415-73009415 on Build GRCh38
rs149294115 Chr.8: 73009048-73009049 on Build GRCh38
rs1040361324 Chr.8: 73009376-73009376 on Build GRCh38
rs894058014 Chr.8: 73009447-73009447 on Build GRCh38
rs890212400 Chr.8: 73009066-73009066 on Build GRCh38
rs987048655 Chr.8: 73009236-73009236 on Build GRCh38
rs758151093 Chr.8: 73009076-73009076 on Build GRCh38
rs866788033 Chr.8: 73009295-73009295 on Build GRCh38
rs747745639 Chr.8: 73009248-73009248 on Build GRCh38
rs796907651 Chr.8: 73009258-73009258 on Build GRCh38
rs71267925 Chr.8: 73009303-73009304 on Build GRCh38
rs776328276 Chr.8: 73009153-73009153 on Build GRCh38
rs772332822 Chr.8: 73009129-73009129 on Build GRCh38
rs762854177 Chr.8: 73009255-73009255 on Build GRCh38
rs891376205 Chr.8: 73009364-73009364 on Build GRCh38
rs199759499 Chr.8: 73009298-73009299 on Build GRCh38
rs766315992 Chr.8: 73009257-73009257 on Build GRCh38
rs760314774 Chr.8: 73009047-73009047 on Build GRCh38
rs563078591 Chr.8: 73009471-73009471 on Build GRCh38
rs755588334 Chr.8: 73009086-73009087 on Build GRCh38
rs999758009 Chr.8: 73009158-73009158 on Build GRCh38
rs761374911 Chr.8: 73009167-73009167 on Build GRCh38
rs1012003158 Chr.8: 73009407-73009407 on Build GRCh38
rs758943132 Chr.8: 73009227-73009227 on Build GRCh38
rs990444165 Chr.8: 73009316-73009316 on Build GRCh38
rs746332601 Chr.8: 73009122-73009122 on Build GRCh38
rs56288812 Chr.8: 73009171-73009171 on Build GRCh38
rs1047330828 Chr.8: 73009123-73009123 on Build GRCh38
rs764799999 Chr.8: 73009168-73009168 on Build GRCh38
rs757766340 Chr.8: 73009049-73009051 on Build GRCh38
rs775756503 Chr.8: 73009080-73009080 on Build GRCh38
rs778997056 Chr.8: 73009223-73009223 on Build GRCh38
rs779324009 Chr.8: 73009114-73009114 on Build GRCh38
rs991823376 Chr.8: 73009230-73009230 on Build GRCh38
rs764273904 Chr.8: 73009195-73009195 on Build GRCh38
rs560386013 Chr.8: 73009410-73009410 on Build GRCh38
rs763699674 Chr.8: 73009051-73009051 on Build GRCh38
rs56349932 Chr.8: 73009147-73009147 on Build GRCh38
rs542018813 Chr.8: 73009400-73009400 on Build GRCh38
rs749350509 Chr.8: 73009097-73009097 on Build GRCh38
rs750271429 Chr.8: 73009073-73009073 on Build GRCh38
rs766155659 Chr.8: 73009180-73009180 on Build GRCh38
rs761898094 Chr.8: 73009068-73009068 on Build GRCh38
rs973148636 Chr.8: 73009292-73009292 on Build GRCh38
rs779461763 Chr.8: 73009091-73009091 on Build GRCh38
rs891407231 Chr.8: 73009124-73009124 on Build GRCh38
rs765243417 Chr.8: 73009070-73009070 on Build GRCh38
rs775024422 Chr.8: 73009046-73009046 on Build GRCh38
rs756327332 Chr.8: 73009094-73009094 on Build GRCh38
rs763836268 Chr.8: 73009295-73009296 on Build GRCh38
rs762882340 Chr.8: 73009173-73009173 on Build GRCh38
rs893839558 Chr.8: 73009188-73009188 on Build GRCh38
rs960148295 Chr.8: 73009482-73009482 on Build GRCh38
rs931005425 Chr.8: 73009305-73009305 on Build GRCh38
rs775854291 Chr.8: 73009135-73009135 on Build GRCh38
rs757804917 Chr.8: 73009112-73009112 on Build GRCh38
rs1047299766 Chr.8: 73009345-73009345 on Build GRCh38
rs201464789 Chr.8: 73009077-73009077 on Build GRCh38
rs756380562 Chr.8: 73009193-73009193 on Build GRCh38
rs996211509 Chr.8: 73009450-73009450 on Build GRCh38
rs554864170 Chr.8: 73009044-73009044 on Build GRCh38
rs112760903 Chr.8: 73009299-73009299 on Build GRCh38
rs1021647391 Chr.8: 73009413-73009413 on Build GRCh38

View Full Product Details