Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 22161464-22161961 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SFTPC
Entrez Gene ID
6440
Gene Name
surfactant protein C
Gene Aliases
BRICD6, PSP-C, SFTP2, SMDP2, SP-C
UniGene
Hs.1074
Gene Chromosome Location
Chr.8: 22161671-22164479 on Build GRCh38
Transcripts
NM_001172357.1 NM_001172410.1 NM_001317778.1 NM_001317779.1 NM_001317780.1 NM_003018.3 XM_011544613.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1098041 c.21G>A p.E7E Chr.8: 22161849-22161849 on Build GRCh38
6776466 c.39G>A p.P13P Chr.8: 22161867-22161867 on Build GRCh38
7389538 c.42G>A p.P14P Chr.8: 22161870-22161870 on Build GRCh38
6112787 c.10G>A p.G4S Chr.8: 22161838-22161838 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs763225526 Chr.8: 22161772-22161772 on Build GRCh38
rs8192339 Chr.8: 22161558-22161558 on Build GRCh38
rs753248473 Chr.8: 22161785-22161785 on Build GRCh38
rs74639360 Chr.8: 22161729-22161729 on Build GRCh38
rs369413166 Chr.8: 22161864-22161864 on Build GRCh38
rs756372282 Chr.8: 22161795-22161795 on Build GRCh38
rs575634399 Chr.8: 22161904-22161904 on Build GRCh38
rs199905878 Chr.8: 22161870-22161870 on Build GRCh38
rs149756879 Chr.8: 22161909-22161909 on Build GRCh38
rs1001448911 Chr.8: 22161650-22161650 on Build GRCh38
rs80139123 Chr.8: 22161538-22161538 on Build GRCh38
rs758095773 Chr.8: 22161812-22161812 on Build GRCh38
rs78177348 Chr.8: 22161907-22161907 on Build GRCh38
rs373338880 Chr.8: 22161777-22161777 on Build GRCh38
rs778358826 Chr.8: 22161734-22161734 on Build GRCh38
rs77253713 Chr.8: 22161738-22161738 on Build GRCh38
rs545529507 Chr.8: 22161772-22161773 on Build GRCh38
rs964212946 Chr.8: 22161902-22161902 on Build GRCh38
rs754211081 Chr.8: 22161887-22161887 on Build GRCh38
rs781209749 Chr.8: 22161828-22161828 on Build GRCh38
rs767594462 Chr.8: 22161879-22161879 on Build GRCh38
rs757721748 Chr.8: 22161897-22161897 on Build GRCh38
rs768313265 Chr.8: 22161827-22161827 on Build GRCh38
rs1025317801 Chr.8: 22161705-22161705 on Build GRCh38
rs749041331 Chr.8: 22161918-22161918 on Build GRCh38
rs573825268 Chr.8: 22161611-22161611 on Build GRCh38
rs78201008 Chr.8: 22161811-22161811 on Build GRCh38
rs773026639 Chr.8: 22161856-22161856 on Build GRCh38
rs761236918 Chr.8: 22161885-22161885 on Build GRCh38
rs886062814 Chr.8: 22161774-22161775 on Build GRCh38
rs980837686 Chr.8: 22161911-22161911 on Build GRCh38
rs532360636 Chr.8: 22161867-22161867 on Build GRCh38
rs986985258 Chr.8: 22161730-22161730 on Build GRCh38
rs755444690 Chr.8: 22161728-22161728 on Build GRCh38
rs919711140 Chr.8: 22161592-22161592 on Build GRCh38
rs765668378 Chr.8: 22161930-22161930 on Build GRCh38
rs772183067 Chr.8: 22161792-22161799 on Build GRCh38
rs767840525 Chr.8: 22161519-22161519 on Build GRCh38
rs773257848 Chr.8: 22161803-22161805 on Build GRCh38
rs925536586 Chr.8: 22161745-22161745 on Build GRCh38
rs1001777224 Chr.8: 22161809-22161809 on Build GRCh38
rs201896861 Chr.8: 22161838-22161838 on Build GRCh38
rs76324979 Chr.8: 22161668-22161668 on Build GRCh38
rs543709575 Chr.8: 22161557-22161557 on Build GRCh38
rs564964303 Chr.8: 22161840-22161840 on Build GRCh38
rs898454266 Chr.8: 22161641-22161641 on Build GRCh38
rs935292025 Chr.8: 22161750-22161750 on Build GRCh38
rs905718380 Chr.8: 22161616-22161616 on Build GRCh38
rs762050676 Chr.8: 22161751-22161751 on Build GRCh38
rs746560865 Chr.8: 22161746-22161746 on Build GRCh38
rs1012802925 Chr.8: 22161933-22161933 on Build GRCh38
rs549699699 Chr.8: 22161752-22161752 on Build GRCh38
rs764551500 Chr.8: 22161886-22161886 on Build GRCh38
rs751703149 Chr.8: 22161727-22161727 on Build GRCh38
rs986998821 Chr.8: 22161504-22161504 on Build GRCh38
rs79647630 Chr.8: 22161737-22161737 on Build GRCh38
rs11552815 Chr.8: 22161854-22161854 on Build GRCh38
rs1008471359 Chr.8: 22161686-22161686 on Build GRCh38
rs533083304 Chr.8: 22161669-22161669 on Build GRCh38
rs759541738 Chr.8: 22161875-22161875 on Build GRCh38
rs8192340 Chr.8: 22161905-22161905 on Build GRCh38
rs935322754 Chr.8: 22161555-22161555 on Build GRCh38
rs367559125 Chr.8: 22161763-22161763 on Build GRCh38
rs200665803 Chr.8: 22161798-22161798 on Build GRCh38
rs925273505 Chr.8: 22161550-22161550 on Build GRCh38
rs746473012 Chr.8: 22161826-22161826 on Build GRCh38
rs766625842 Chr.8: 22161776-22161776 on Build GRCh38
rs879569099 Chr.8: 22161532-22161532 on Build GRCh38
rs1016842547 Chr.8: 22161651-22161651 on Build GRCh38
rs144603526 Chr.8: 22161852-22161852 on Build GRCh38
rs750070143 Chr.8: 22161759-22161759 on Build GRCh38
rs764439267 Chr.8: 22161565-22161565 on Build GRCh38
rs74868576 Chr.8: 22161556-22161556 on Build GRCh38
rs375113623 Chr.8: 22161884-22161884 on Build GRCh38
rs145625779 Chr.8: 22161924-22161924 on Build GRCh38
rs758318330 Chr.8: 22161765-22161765 on Build GRCh38
rs557343687 Chr.8: 22161547-22161547 on Build GRCh38
rs545131719 Chr.8: 22161931-22161931 on Build GRCh38
rs140610971 Chr.8: 22161908-22161908 on Build GRCh38
rs779710215 Chr.8: 22161740-22161740 on Build GRCh38
rs567738879 Chr.8: 22161761-22161761 on Build GRCh38
rs977717997 Chr.8: 22161494-22161494 on Build GRCh38
rs541104547 Chr.8: 22161664-22161664 on Build GRCh38
rs767971320 Chr.8: 22161778-22161778 on Build GRCh38
rs1039934164 Chr.8: 22161903-22161904 on Build GRCh38
rs879184908 Chr.8: 22161631-22161631 on Build GRCh38
rs1035379553 Chr.8: 22161760-22161760 on Build GRCh38
rs28438700 Chr.8: 22161695-22161695 on Build GRCh38
rs756529000 Chr.8: 22161731-22161731 on Build GRCh38
rs886062815 Chr.8: 22161806-22161808 on Build GRCh38
rs760698424 Chr.8: 22161819-22161820 on Build GRCh38
rs147884807 Chr.8: 22161906-22161906 on Build GRCh38
rs766093752 Chr.8: 22161900-22161900 on Build GRCh38
rs371755328 Chr.8: 22161819-22161819 on Build GRCh38
rs377267823 Chr.8: 22161762-22161762 on Build GRCh38
rs1018443678 Chr.8: 22161890-22161890 on Build GRCh38

View Full Product Details