Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 22247131-22247631 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
POLR3D
Entrez Gene ID
661
Gene Name
RNA polymerase III subunit D
Gene Aliases
BN51T, RPC4, RPC53, TSBN51
UniGene
Hs.148342
Gene Chromosome Location
Chr.8: 22245104-22251167 on Build GRCh38
Transcripts
NM_001722.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4188473 c.168A>C p.K56N Chr.8: 22247223-22247223 on Build GRCh38
6752424 c.194G>A p.R65Q Chr.8: 22247249-22247249 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1011499613 Chr.8: 22247269-22247269 on Build GRCh38
rs900904406 Chr.8: 22247180-22247180 on Build GRCh38
rs540765735 Chr.8: 22247241-22247241 on Build GRCh38
rs922331459 Chr.8: 22247474-22247474 on Build GRCh38
rs201039416 Chr.8: 22247192-22247192 on Build GRCh38
rs776202500 Chr.8: 22247262-22247262 on Build GRCh38
rs542061996 Chr.8: 22247534-22247534 on Build GRCh38
rs567515164 Chr.8: 22247361-22247361 on Build GRCh38
rs530861183 Chr.8: 22247327-22247327 on Build GRCh38
rs939642260 Chr.8: 22247177-22247177 on Build GRCh38
rs373646193 Chr.8: 22247182-22247182 on Build GRCh38
rs185242902 Chr.8: 22247386-22247386 on Build GRCh38
rs771312556 Chr.8: 22247225-22247225 on Build GRCh38
rs777779865 Chr.8: 22247315-22247315 on Build GRCh38
rs1031657665 Chr.8: 22247551-22247551 on Build GRCh38
rs919557599 Chr.8: 22247338-22247338 on Build GRCh38
rs1022402289 Chr.8: 22247477-22247477 on Build GRCh38
rs952550151 Chr.8: 22247383-22247383 on Build GRCh38
rs902765862 Chr.8: 22247587-22247587 on Build GRCh38
rs549189282 Chr.8: 22247346-22247346 on Build GRCh38
rs370360179 Chr.8: 22247194-22247194 on Build GRCh38
rs555566565 Chr.8: 22247450-22247450 on Build GRCh38
rs1035019348 Chr.8: 22247323-22247323 on Build GRCh38
rs908474654 Chr.8: 22247174-22247174 on Build GRCh38
rs544295702 Chr.8: 22247304-22247304 on Build GRCh38
rs563714172 Chr.8: 22247407-22247407 on Build GRCh38
rs149905010 Chr.8: 22247431-22247432 on Build GRCh38
rs976405102 Chr.8: 22247469-22247469 on Build GRCh38
rs983970750 Chr.8: 22247399-22247399 on Build GRCh38
rs145048190 Chr.8: 22247314-22247314 on Build GRCh38
rs999550733 Chr.8: 22247548-22247548 on Build GRCh38
rs893559285 Chr.8: 22247554-22247554 on Build GRCh38
rs762534474 Chr.8: 22247410-22247410 on Build GRCh38
rs34392777 Chr.8: 22247178-22247178 on Build GRCh38
rs932310271 Chr.8: 22247535-22247535 on Build GRCh38
rs1042562468 Chr.8: 22247579-22247579 on Build GRCh38
rs946868213 Chr.8: 22247570-22247570 on Build GRCh38
rs968018274 Chr.8: 22247546-22247546 on Build GRCh38
rs761487828 Chr.8: 22247264-22247264 on Build GRCh38
rs113911804 Chr.8: 22247442-22247442 on Build GRCh38
rs990881220 Chr.8: 22247337-22247337 on Build GRCh38
rs756221897 Chr.8: 22247190-22247190 on Build GRCh38
rs1049456427 Chr.8: 22247539-22247539 on Build GRCh38
rs192862658 Chr.8: 22247279-22247279 on Build GRCh38
rs893934539 Chr.8: 22247223-22247223 on Build GRCh38
rs939947374 Chr.8: 22247454-22247454 on Build GRCh38
rs754876187 Chr.8: 22247176-22247176 on Build GRCh38
rs766210860 Chr.8: 22247289-22247289 on Build GRCh38
rs992368487 Chr.8: 22247295-22247295 on Build GRCh38
rs1039581382 Chr.8: 22247412-22247412 on Build GRCh38
rs1047397349 Chr.8: 22247393-22247393 on Build GRCh38
rs992733508 Chr.8: 22247576-22247576 on Build GRCh38
rs373388070 Chr.8: 22247244-22247244 on Build GRCh38
rs139417336 Chr.8: 22247285-22247285 on Build GRCh38

View Full Product Details