Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 22408228-22408733 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC39A14
Entrez Gene ID
23516
Gene Name
solute carrier family 39 member 14
Gene Aliases
HMNDYT2, LZT-Hs4, NET34, ZIP14, cig19
UniGene
Hs.491232
Gene Chromosome Location
Chr.8: 22367249-22434129 on Build GRCh38
Transcripts
NM_001128431.2 NM_001135153.1 NM_001135154.1 NM_015359.4 XM_005273465.2 XM_005273466.4 XM_006716323.2 XM_006716324.2 XM_011544478.2 XM_017013292.1 XM_017013293.1 XM_017013294.1 XM_017013295.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6237481 c.395C>T p.S132L Chr.8: 22408434-22408434 on Build GRCh38
4366729 c.395C>T p.S132L Chr.8: 22408434-22408434 on Build GRCh38
3898933 c.393C>A p.T131T Chr.8: 22408432-22408432 on Build GRCh38
1098092 c.373C>T p.L125L Chr.8: 22408412-22408412 on Build GRCh38
5874369 c.428C>A p.T143K Chr.8: 22408467-22408467 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749258460 Chr.8: 22408532-22408532 on Build GRCh38
rs1002499320 Chr.8: 22408683-22408683 on Build GRCh38
rs13266541 Chr.8: 22408622-22408622 on Build GRCh38
rs753991288 Chr.8: 22408327-22408327 on Build GRCh38
rs772125874 Chr.8: 22408468-22408470 on Build GRCh38
rs753123757 Chr.8: 22408467-22408467 on Build GRCh38
rs376452266 Chr.8: 22408290-22408290 on Build GRCh38
rs201522442 Chr.8: 22408314-22408314 on Build GRCh38
rs1033582703 Chr.8: 22408562-22408562 on Build GRCh38
rs549131410 Chr.8: 22408257-22408257 on Build GRCh38
rs920470308 Chr.8: 22408488-22408488 on Build GRCh38
rs892842882 Chr.8: 22408579-22408579 on Build GRCh38
rs747005078 Chr.8: 22408422-22408422 on Build GRCh38
rs13258982 Chr.8: 22408558-22408558 on Build GRCh38
rs768650948 Chr.8: 22408526-22408526 on Build GRCh38
rs781664946 Chr.8: 22408375-22408375 on Build GRCh38
rs752102367 Chr.8: 22408451-22408451 on Build GRCh38
rs775381592 Chr.8: 22408414-22408414 on Build GRCh38
rs746666663 Chr.8: 22408322-22408322 on Build GRCh38
rs181530530 Chr.8: 22408560-22408560 on Build GRCh38
rs763095255 Chr.8: 22408443-22408443 on Build GRCh38
rs757888834 Chr.8: 22408487-22408487 on Build GRCh38
rs755579926 Chr.8: 22408458-22408458 on Build GRCh38
rs1034284865 Chr.8: 22408456-22408456 on Build GRCh38
rs112687724 Chr.8: 22408305-22408305 on Build GRCh38
rs946240202 Chr.8: 22408617-22408617 on Build GRCh38
rs745566721 Chr.8: 22408486-22408486 on Build GRCh38
rs750529883 Chr.8: 22408351-22408351 on Build GRCh38
rs930581455 Chr.8: 22408295-22408295 on Build GRCh38
rs1034339062 Chr.8: 22408637-22408637 on Build GRCh38
rs755375986 Chr.8: 22408370-22408370 on Build GRCh38
rs567262593 Chr.8: 22408268-22408268 on Build GRCh38
rs3817445 Chr.8: 22408541-22408541 on Build GRCh38
rs988525718 Chr.8: 22408342-22408342 on Build GRCh38
rs190153898 Chr.8: 22408435-22408435 on Build GRCh38
rs779604913 Chr.8: 22408525-22408525 on Build GRCh38
rs748892669 Chr.8: 22408376-22408376 on Build GRCh38
rs995913384 Chr.8: 22408355-22408355 on Build GRCh38
rs1015155815 Chr.8: 22408710-22408710 on Build GRCh38
rs534747726 Chr.8: 22408283-22408283 on Build GRCh38
rs756815173 Chr.8: 22408468-22408468 on Build GRCh38
rs17060832 Chr.8: 22408639-22408639 on Build GRCh38
rs966012516 Chr.8: 22408714-22408714 on Build GRCh38
rs778234130 Chr.8: 22408390-22408390 on Build GRCh38
rs763327826 Chr.8: 22408537-22408537 on Build GRCh38
rs765162706 Chr.8: 22408350-22408350 on Build GRCh38
rs951521587 Chr.8: 22408705-22408705 on Build GRCh38
rs756475794 Chr.8: 22408545-22408545 on Build GRCh38
rs756942789 Chr.8: 22408383-22408383 on Build GRCh38
rs138114438 Chr.8: 22408360-22408360 on Build GRCh38
rs951906800 Chr.8: 22408507-22408507 on Build GRCh38
rs1047639333 Chr.8: 22408297-22408297 on Build GRCh38
rs13281466 Chr.8: 22408559-22408559 on Build GRCh38
rs749020500 Chr.8: 22408262-22408262 on Build GRCh38
rs565036720 Chr.8: 22408623-22408623 on Build GRCh38
rs143724214 Chr.8: 22408434-22408434 on Build GRCh38
rs781485871 Chr.8: 22408362-22408362 on Build GRCh38
rs367623498 Chr.8: 22408405-22408405 on Build GRCh38
rs995585354 Chr.8: 22408646-22408646 on Build GRCh38
rs907568888 Chr.8: 22408301-22408301 on Build GRCh38
rs958813430 Chr.8: 22408476-22408476 on Build GRCh38
rs775223692 Chr.8: 22408299-22408299 on Build GRCh38
rs745409989 Chr.8: 22408393-22408393 on Build GRCh38
rs778069079 Chr.8: 22408450-22408450 on Build GRCh38
rs536468517 Chr.8: 22408543-22408543 on Build GRCh38
rs140464999 Chr.8: 22408706-22408706 on Build GRCh38
rs371767499 Chr.8: 22408531-22408531 on Build GRCh38
rs1040037505 Chr.8: 22408338-22408338 on Build GRCh38
rs1027080443 Chr.8: 22408687-22408687 on Build GRCh38
rs762162034 Chr.8: 22408348-22408348 on Build GRCh38
rs921579731 Chr.8: 22408554-22408554 on Build GRCh38
rs557695720 Chr.8: 22408536-22408536 on Build GRCh38
rs900263790 Chr.8: 22408354-22408354 on Build GRCh38
rs370599682 Chr.8: 22408409-22408409 on Build GRCh38
rs76054297 Chr.8: 22408569-22408569 on Build GRCh38
rs372429540 Chr.8: 22408440-22408440 on Build GRCh38
rs879253764 Chr.8: 22408352-22408352 on Build GRCh38
rs779515680 Chr.8: 22408542-22408542 on Build GRCh38
rs186225449 Chr.8: 22408431-22408431 on Build GRCh38
rs776241676 Chr.8: 22408529-22408529 on Build GRCh38
rs958581422 Chr.8: 22408703-22408703 on Build GRCh38
rs1003066200 Chr.8: 22408634-22408634 on Build GRCh38
rs767710689 Chr.8: 22408465-22408465 on Build GRCh38
rs1006974564 Chr.8: 22408258-22408258 on Build GRCh38
rs766565831 Chr.8: 22408540-22408540 on Build GRCh38
rs974112153 Chr.8: 22408479-22408479 on Build GRCh38
rs772746995 Chr.8: 22408621-22408621 on Build GRCh38
rs879253763 Chr.8: 22408331-22408331 on Build GRCh38
rs377005073 Chr.8: 22408478-22408478 on Build GRCh38

View Full Product Details