Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 144356628-144357124 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FBXL6
Entrez Gene ID
26233
Gene Name
F-box and leucine rich repeat protein 6
Gene Aliases
FBL6, FBL6A, PP14630
UniGene
Hs.12271
Gene Chromosome Location
Chr.8: 144355428-144358523 on Build GRCh38
Transcripts
NM_012162.3 NM_024555.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5672962 c.693C>T p.H231H Chr.8: 144357068-144357068 on Build GRCh38
3898188 c.696T>C p.G232G Chr.8: 144357065-144357065 on Build GRCh38
1455614 c.834G>A p.W278* Chr.8: 144356853-144356853 on Build GRCh38
5531100 c.745C>T p.H249Y Chr.8: 144357016-144357016 on Build GRCh38
6050112 c.662G>T p.R221L Chr.8: 144357099-144357099 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs191055486 Chr.8: 144356737-144356737 on Build GRCh38
rs782471645 Chr.8: 144356671-144356671 on Build GRCh38
rs782112940 Chr.8: 144356886-144356886 on Build GRCh38
rs371484539 Chr.8: 144356739-144356739 on Build GRCh38
rs782111680 Chr.8: 144356956-144356957 on Build GRCh38
rs782737277 Chr.8: 144356723-144356724 on Build GRCh38
rs781999908 Chr.8: 144356710-144356710 on Build GRCh38
rs782222157 Chr.8: 144356934-144356934 on Build GRCh38
rs782776424 Chr.8: 144356982-144356982 on Build GRCh38
rs1031898859 Chr.8: 144356999-144356999 on Build GRCh38
rs369704521 Chr.8: 144356927-144356927 on Build GRCh38
rs782094488 Chr.8: 144356703-144356703 on Build GRCh38
rs61746974 Chr.8: 144356875-144356875 on Build GRCh38
rs782662505 Chr.8: 144356928-144356928 on Build GRCh38
rs782464019 Chr.8: 144357024-144357025 on Build GRCh38
rs532082480 Chr.8: 144356755-144356755 on Build GRCh38
rs375647211 Chr.8: 144356925-144356925 on Build GRCh38
rs576727660 Chr.8: 144356654-144356654 on Build GRCh38
rs782506336 Chr.8: 144356735-144356735 on Build GRCh38
rs782625066 Chr.8: 144356924-144356924 on Build GRCh38
rs781905056 Chr.8: 144357080-144357080 on Build GRCh38
rs781885156 Chr.8: 144356896-144356896 on Build GRCh38
rs201711347 Chr.8: 144356899-144356899 on Build GRCh38
rs782005596 Chr.8: 144356813-144356813 on Build GRCh38
rs149208512 Chr.8: 144357100-144357100 on Build GRCh38
rs372382267 Chr.8: 144356980-144356980 on Build GRCh38
rs199547847 Chr.8: 144356892-144356892 on Build GRCh38
rs781839444 Chr.8: 144357043-144357043 on Build GRCh38
rs368962350 Chr.8: 144356970-144356970 on Build GRCh38
rs146812670 Chr.8: 144357055-144357055 on Build GRCh38
rs552998165 Chr.8: 144356734-144356734 on Build GRCh38
rs1048025880 Chr.8: 144356698-144356698 on Build GRCh38
rs955068843 Chr.8: 144357009-144357009 on Build GRCh38
rs552672053 Chr.8: 144356941-144356941 on Build GRCh38
rs782155053 Chr.8: 144356714-144356714 on Build GRCh38
rs781947849 Chr.8: 144356701-144356701 on Build GRCh38
rs781887898 Chr.8: 144356992-144356993 on Build GRCh38
rs910283330 Chr.8: 144356841-144356841 on Build GRCh38
rs987643189 Chr.8: 144356834-144356834 on Build GRCh38
rs781805773 Chr.8: 144356830-144356830 on Build GRCh38
rs781950205 Chr.8: 144356877-144356877 on Build GRCh38
rs200807701 Chr.8: 144356816-144356816 on Build GRCh38
rs537010969 Chr.8: 144356731-144356731 on Build GRCh38
rs141380444 Chr.8: 144356686-144356686 on Build GRCh38
rs201162690 Chr.8: 144357083-144357083 on Build GRCh38
rs782099692 Chr.8: 144356778-144356778 on Build GRCh38
rs148685592 Chr.8: 144356648-144356648 on Build GRCh38
rs973040915 Chr.8: 144356869-144356869 on Build GRCh38
rs782628611 Chr.8: 144357006-144357006 on Build GRCh38
rs781956596 Chr.8: 144357040-144357040 on Build GRCh38
rs373543815 Chr.8: 144356935-144356935 on Build GRCh38
rs782163549 Chr.8: 144356817-144356817 on Build GRCh38
rs368813425 Chr.8: 144356847-144356847 on Build GRCh38
rs782768251 Chr.8: 144356889-144356889 on Build GRCh38
rs782301861 Chr.8: 144356855-144356855 on Build GRCh38
rs782182511 Chr.8: 144356770-144356770 on Build GRCh38
rs782332873 Chr.8: 144356774-144356774 on Build GRCh38
rs782076645 Chr.8: 144357067-144357067 on Build GRCh38
rs564523873 Chr.8: 144356949-144356949 on Build GRCh38
rs143646016 Chr.8: 144357015-144357015 on Build GRCh38
rs781846827 Chr.8: 144356842-144356842 on Build GRCh38
rs782070332 Chr.8: 144356953-144356953 on Build GRCh38
rs781795704 Chr.8: 144356724-144356724 on Build GRCh38
rs782383000 Chr.8: 144357044-144357044 on Build GRCh38
rs376850993 Chr.8: 144356759-144356759 on Build GRCh38
rs201204346 Chr.8: 144356662-144356662 on Build GRCh38
rs782155174 Chr.8: 144357052-144357052 on Build GRCh38
rs201026916 Chr.8: 144356776-144356776 on Build GRCh38
rs782249729 Chr.8: 144356676-144356676 on Build GRCh38
rs150030093 Chr.8: 144357096-144357096 on Build GRCh38
rs782480866 Chr.8: 144357004-144357004 on Build GRCh38
rs782703101 Chr.8: 144356912-144356912 on Build GRCh38
rs782559616 Chr.8: 144357086-144357086 on Build GRCh38
rs199851775 Chr.8: 144356736-144356736 on Build GRCh38
rs139330417 Chr.8: 144357076-144357076 on Build GRCh38
rs782724713 Chr.8: 144356881-144356881 on Build GRCh38
rs373298729 Chr.8: 144357068-144357068 on Build GRCh38
rs200079529 Chr.8: 144357011-144357011 on Build GRCh38
rs185753888 Chr.8: 144356708-144356708 on Build GRCh38
rs968956255 Chr.8: 144356769-144356769 on Build GRCh38
rs782567879 Chr.8: 144356661-144356661 on Build GRCh38
rs181960705 Chr.8: 144356772-144356772 on Build GRCh38
rs782173457 Chr.8: 144357020-144357020 on Build GRCh38
rs782210639 Chr.8: 144356745-144356745 on Build GRCh38
rs782474389 Chr.8: 144356917-144356917 on Build GRCh38
rs782262513 Chr.8: 144357008-144357008 on Build GRCh38
rs782808542 Chr.8: 144356717-144356717 on Build GRCh38
rs376025805 Chr.8: 144356983-144356983 on Build GRCh38
rs782326012 Chr.8: 144357033-144357033 on Build GRCh38
rs190545783 Chr.8: 144356882-144356882 on Build GRCh38
rs147241187 Chr.8: 144356864-144356864 on Build GRCh38
rs782472849 Chr.8: 144356747-144356747 on Build GRCh38
rs536952980 Chr.8: 144356871-144356871 on Build GRCh38
rs782697742 Chr.8: 144356822-144356822 on Build GRCh38
rs781922743 Chr.8: 144356951-144356951 on Build GRCh38
rs369106175 Chr.8: 144356768-144356768 on Build GRCh38
rs1009060977 Chr.8: 144357039-144357039 on Build GRCh38
rs372391839 Chr.8: 144356732-144356732 on Build GRCh38
rs139706818 Chr.8: 144356943-144356943 on Build GRCh38
rs782210774 Chr.8: 144356868-144356868 on Build GRCh38
rs782752956 Chr.8: 144356833-144356833 on Build GRCh38
rs920276321 Chr.8: 144357074-144357074 on Build GRCh38
rs146049985 Chr.8: 144356832-144356832 on Build GRCh38
rs782173327 Chr.8: 144356931-144356931 on Build GRCh38
rs782259584 Chr.8: 144356757-144356757 on Build GRCh38
rs782011297 Chr.8: 144357051-144357051 on Build GRCh38
rs782409770 Chr.8: 144356874-144356874 on Build GRCh38
rs782589898 Chr.8: 144356990-144356990 on Build GRCh38
rs144139611 Chr.8: 144356749-144356749 on Build GRCh38
rs782574735 Chr.8: 144356744-144356744 on Build GRCh38
rs781816444 Chr.8: 144356994-144356994 on Build GRCh38
rs541935911 Chr.8: 144356948-144356948 on Build GRCh38
rs782688782 Chr.8: 144356689-144356689 on Build GRCh38
rs374186025 Chr.8: 144356670-144356670 on Build GRCh38
rs781930248 Chr.8: 144357059-144357059 on Build GRCh38
rs782526062 Chr.8: 144357102-144357102 on Build GRCh38
rs548024038 Chr.8: 144356863-144356863 on Build GRCh38
rs186501318 Chr.8: 144356800-144356800 on Build GRCh38
rs781876727 Chr.8: 144356731-144356731 on Build GRCh38
rs1021918406 Chr.8: 144356746-144356746 on Build GRCh38
rs1012907528 Chr.8: 144356741-144356741 on Build GRCh38
rs782730125 Chr.8: 144356957-144356957 on Build GRCh38
rs369988113 Chr.8: 144356729-144356730 on Build GRCh38
rs782820466 Chr.8: 144356908-144356908 on Build GRCh38
rs904490383 Chr.8: 144356987-144356987 on Build GRCh38
rs368399680 Chr.8: 144356687-144356687 on Build GRCh38
rs781811695 Chr.8: 144356914-144356914 on Build GRCh38
rs782820063 Chr.8: 144356957-144356957 on Build GRCh38
rs964381457 Chr.8: 144356844-144356844 on Build GRCh38
rs544748472 Chr.8: 144356647-144356647 on Build GRCh38
rs782622446 Chr.8: 144356675-144356675 on Build GRCh38
rs782174322 Chr.8: 144356690-144356690 on Build GRCh38

View Full Product Details