Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 144358085-144358580 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FBXL6
Entrez Gene ID
26233
Gene Name
F-box and leucine rich repeat protein 6
Gene Aliases
FBL6, FBL6A, PP14630
UniGene
Hs.12271
Gene Chromosome Location
Chr.8: 144355428-144358523 on Build GRCh38
Transcripts
NM_012162.3 NM_024555.5 NM_001253816.1 XM_017013822.1

Gene Symbol
SLC52A2
Entrez Gene ID
79581
Gene Name
solute carrier family 52 member 2
Gene Aliases
BVVLS2, D15Ertd747e, GPCR41, GPR172A, PAR1, RFT3, RFVT2, hRFT3
UniGene
Hs.6459
Gene Chromosome Location
Chr.8: 144358547-144361286 on Build GRCh38
Transcripts
NM_012162.3 NM_024555.5 NM_001253816.1 XM_017013822.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6661982 c.361G>A p.V121M Chr.8: 144358087-144358087 on Build GRCh38
6193835 c.305C>A p.T102K Chr.8: 144358143-144358143 on Build GRCh38
5626328 c.55C>T p.P19S Chr.8: 144358393-144358393 on Build GRCh38
4646205 c.9C>T p.A3A Chr.8: 144358439-144358439 on Build GRCh38
6661984 c.277C>G p.P93A Chr.8: 144358171-144358171 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs952021230 Chr.8: 144358479-144358479 on Build GRCh38
rs551163570 Chr.8: 144358519-144358519 on Build GRCh38
rs782640194 Chr.8: 144358331-144358331 on Build GRCh38
rs782336300 Chr.8: 144358163-144358163 on Build GRCh38
rs113552670 Chr.8: 144358428-144358428 on Build GRCh38
rs947799880 Chr.8: 144358515-144358515 on Build GRCh38
rs782742094 Chr.8: 144358116-144358116 on Build GRCh38
rs782280291 Chr.8: 144358439-144358439 on Build GRCh38
rs782527871 Chr.8: 144358407-144358407 on Build GRCh38
rs782602801 Chr.8: 144358427-144358427 on Build GRCh38
rs782222413 Chr.8: 144358323-144358323 on Build GRCh38
rs561994701 Chr.8: 144358342-144358342 on Build GRCh38
rs782690748 Chr.8: 144358408-144358408 on Build GRCh38
rs570360612 Chr.8: 144358364-144358364 on Build GRCh38
rs1052399633 Chr.8: 144358537-144358537 on Build GRCh38
rs782079584 Chr.8: 144358451-144358451 on Build GRCh38
rs782805336 Chr.8: 144358297-144358297 on Build GRCh38
rs918057011 Chr.8: 144358508-144358508 on Build GRCh38
rs1056245359 Chr.8: 144358370-144358370 on Build GRCh38
rs782203191 Chr.8: 144358338-144358338 on Build GRCh38
rs35266279 Chr.8: 144358260-144358260 on Build GRCh38
rs782743551 Chr.8: 144358282-144358282 on Build GRCh38
rs782053106 Chr.8: 144358477-144358477 on Build GRCh38
rs375550621 Chr.8: 144358140-144358140 on Build GRCh38
rs781782800 Chr.8: 144358314-144358314 on Build GRCh38
rs770883571 Chr.8: 144358119-144358119 on Build GRCh38
rs34763875 Chr.8: 144358292-144358292 on Build GRCh38
rs782750055 Chr.8: 144358398-144358398 on Build GRCh38
rs112549558 Chr.8: 144358179-144358179 on Build GRCh38
rs782274379 Chr.8: 144358151-144358151 on Build GRCh38
rs550638473 Chr.8: 144358434-144358434 on Build GRCh38
rs200282665 Chr.8: 144358290-144358290 on Build GRCh38
rs1019069468 Chr.8: 144358465-144358465 on Build GRCh38
rs564308034 Chr.8: 144358171-144358171 on Build GRCh38
rs782526479 Chr.8: 144358291-144358291 on Build GRCh38
rs782509129 Chr.8: 144358326-144358326 on Build GRCh38
rs782424510 Chr.8: 144358335-144358335 on Build GRCh38
rs781903714 Chr.8: 144358135-144358135 on Build GRCh38
rs13265792 Chr.8: 144358400-144358400 on Build GRCh38
rs781826346 Chr.8: 144358142-144358142 on Build GRCh38
rs782132446 Chr.8: 144358125-144358125 on Build GRCh38
rs183286864 Chr.8: 144358462-144358462 on Build GRCh38
rs782712572 Chr.8: 144358367-144358367 on Build GRCh38
rs529178281 Chr.8: 144358344-144358344 on Build GRCh38
rs782130850 Chr.8: 144358459-144358459 on Build GRCh38
rs528243989 Chr.8: 144358204-144358204 on Build GRCh38
rs113885618 Chr.8: 144358155-144358155 on Build GRCh38
rs903545945 Chr.8: 144358522-144358522 on Build GRCh38
rs782597001 Chr.8: 144358320-144358320 on Build GRCh38
rs896214929 Chr.8: 144358406-144358406 on Build GRCh38
rs563338259 Chr.8: 144358149-144358149 on Build GRCh38
rs782300700 Chr.8: 144358446-144358446 on Build GRCh38
rs903493584 Chr.8: 144358423-144358423 on Build GRCh38
rs974762470 Chr.8: 144358471-144358471 on Build GRCh38
rs782237825 Chr.8: 144358429-144358429 on Build GRCh38
rs782648310 Chr.8: 144358435-144358435 on Build GRCh38
rs781790906 Chr.8: 144358411-144358411 on Build GRCh38
rs781921440 Chr.8: 144358336-144358336 on Build GRCh38
rs1057031728 Chr.8: 144358506-144358506 on Build GRCh38
rs782459581 Chr.8: 144358413-144358413 on Build GRCh38
rs782187802 Chr.8: 144358159-144358159 on Build GRCh38
rs782098443 Chr.8: 144358379-144358379 on Build GRCh38
rs782643057 Chr.8: 144358150-144358150 on Build GRCh38
rs781976449 Chr.8: 144358343-144358343 on Build GRCh38
rs985084225 Chr.8: 144358483-144358483 on Build GRCh38
rs782800101 Chr.8: 144358359-144358359 on Build GRCh38
rs781900084 Chr.8: 144358467-144358467 on Build GRCh38
rs571040659 Chr.8: 144358559-144358559 on Build GRCh38
rs200381094 Chr.8: 144358158-144358158 on Build GRCh38
rs782240396 Chr.8: 144358169-144358169 on Build GRCh38
rs35633164 Chr.8: 144358376-144358376 on Build GRCh38
rs782272254 Chr.8: 144358333-144358333 on Build GRCh38
rs782730397 Chr.8: 144358141-144358141 on Build GRCh38
rs781930050 Chr.8: 144358108-144358108 on Build GRCh38
rs782355251 Chr.8: 144358454-144358454 on Build GRCh38
rs782168336 Chr.8: 144358178-144358178 on Build GRCh38
rs782149168 Chr.8: 144358166-144358167 on Build GRCh38
rs782500969 Chr.8: 144358143-144358143 on Build GRCh38
rs781967081 Chr.8: 144358164-144358164 on Build GRCh38
rs534941041 Chr.8: 144358560-144358560 on Build GRCh38
rs782364876 Chr.8: 144358138-144358139 on Build GRCh38
rs781980167 Chr.8: 144358456-144358456 on Build GRCh38
rs779099074 Chr.8: 144358496-144358496 on Build GRCh38
rs936310719 Chr.8: 144358534-144358534 on Build GRCh38
rs782707898 Chr.8: 144358488-144358488 on Build GRCh38
rs782024812 Chr.8: 144358363-144358363 on Build GRCh38
rs940902751 Chr.8: 144358505-144358505 on Build GRCh38
rs782087192 Chr.8: 144358261-144358261 on Build GRCh38
rs782448687 Chr.8: 144358315-144358315 on Build GRCh38
rs545935518 Chr.8: 144358170-144358170 on Build GRCh38
rs369400816 Chr.8: 144358120-144358120 on Build GRCh38
rs782078974 Chr.8: 144358111-144358111 on Build GRCh38
rs568833806 Chr.8: 144358450-144358450 on Build GRCh38
rs782795427 Chr.8: 144358127-144358127 on Build GRCh38
rs782703374 Chr.8: 144358186-144358186 on Build GRCh38
rs782375884 Chr.8: 144358151-144358152 on Build GRCh38

View Full Product Details