Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 5908498-5909035 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KIAA2026
Entrez Gene ID
158358
Gene Name
KIAA2026
Gene Aliases
-
UniGene
Hs.535060
Gene Chromosome Location
Chr.9: 5860254-6008479 on Build GRCh38
Transcripts
XM_011517759.2 XM_011517760.2 XM_017014335.1 NM_005511.1

Gene Symbol
MLANA
Entrez Gene ID
2315
Gene Name
melan-A
Gene Aliases
MART-1, MART1
UniGene
Hs.154069
Gene Chromosome Location
Chr.9: 5890909-5909822 on Build GRCh38
Transcripts
XM_011517759.2 XM_011517760.2 XM_017014335.1 NM_005511.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2777443 c.320T>A p.L107H Chr.9: 5908671-5908671 on Build GRCh38
3780036 c.313G>C p.E105Q Chr.9: 5908664-5908664 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs949238034 Chr.9: 5908918-5908918 on Build GRCh38
rs1022704561 Chr.9: 5909004-5909004 on Build GRCh38
rs886798444 Chr.9: 5909005-5909005 on Build GRCh38
rs543854573 Chr.9: 5908845-5908845 on Build GRCh38
rs868183543 Chr.9: 5908527-5908527 on Build GRCh38
rs924582525 Chr.9: 5908867-5908867 on Build GRCh38
rs1039786922 Chr.9: 5908538-5908538 on Build GRCh38
rs777989222 Chr.9: 5908633-5908633 on Build GRCh38
rs529289261 Chr.9: 5908883-5908883 on Build GRCh38
rs957497266 Chr.9: 5908957-5908957 on Build GRCh38
rs546101193 Chr.9: 5908933-5908933 on Build GRCh38
rs1027031712 Chr.9: 5908771-5908771 on Build GRCh38
rs777670729 Chr.9: 5908726-5908726 on Build GRCh38
rs944155272 Chr.9: 5908529-5908529 on Build GRCh38
rs539693485 Chr.9: 5908531-5908531 on Build GRCh38
rs749425206 Chr.9: 5908754-5908754 on Build GRCh38
rs768000466 Chr.9: 5908694-5908694 on Build GRCh38
rs942768504 Chr.9: 5908889-5908889 on Build GRCh38
rs1033615446 Chr.9: 5908944-5908944 on Build GRCh38
rs560795480 Chr.9: 5908873-5908873 on Build GRCh38
rs547385238 Chr.9: 5908928-5908928 on Build GRCh38
rs372485841 Chr.9: 5908699-5908699 on Build GRCh38
rs866966055 Chr.9: 5908909-5908909 on Build GRCh38
rs1008939913 Chr.9: 5908884-5908884 on Build GRCh38
rs201214079 Chr.9: 5908688-5908688 on Build GRCh38
rs778939612 Chr.9: 5908596-5908596 on Build GRCh38
rs555890108 Chr.9: 5908610-5908610 on Build GRCh38
rs947437786 Chr.9: 5908853-5908853 on Build GRCh38
rs774946264 Chr.9: 5908670-5908670 on Build GRCh38
rs1038203278 Chr.9: 5908607-5908607 on Build GRCh38
rs921389989 Chr.9: 5908828-5908828 on Build GRCh38
rs2233180 Chr.9: 5908772-5908772 on Build GRCh38
rs770056915 Chr.9: 5908624-5908624 on Build GRCh38
rs746799196 Chr.9: 5908613-5908613 on Build GRCh38
rs2233179 Chr.9: 5908581-5908581 on Build GRCh38
rs866307934 Chr.9: 5908646-5908646 on Build GRCh38
rs1023422953 Chr.9: 5908725-5908725 on Build GRCh38
rs903772429 Chr.9: 5908552-5908552 on Build GRCh38
rs775556272 Chr.9: 5908637-5908637 on Build GRCh38
rs898304379 Chr.9: 5908869-5908869 on Build GRCh38
rs764044269 Chr.9: 5908655-5908655 on Build GRCh38
rs577979836 Chr.9: 5908881-5908881 on Build GRCh38
rs377636376 Chr.9: 5908737-5908737 on Build GRCh38
rs140942317 Chr.9: 5908647-5908647 on Build GRCh38
rs771864243 Chr.9: 5908603-5908603 on Build GRCh38
rs896395827 Chr.9: 5908904-5908904 on Build GRCh38
rs770952264 Chr.9: 5908679-5908679 on Build GRCh38
rs772419510 Chr.9: 5908822-5908822 on Build GRCh38
rs558310703 Chr.9: 5908546-5908546 on Build GRCh38
rs762353589 Chr.9: 5908671-5908671 on Build GRCh38
rs912796369 Chr.9: 5908780-5908780 on Build GRCh38
rs376848033 Chr.9: 5908710-5908710 on Build GRCh38
rs991176936 Chr.9: 5908787-5908787 on Build GRCh38
rs768933181 Chr.9: 5908967-5908967 on Build GRCh38
rs375529491 Chr.9: 5908526-5908526 on Build GRCh38
rs969123202 Chr.9: 5908728-5908728 on Build GRCh38
rs368534927 Chr.9: 5908648-5908648 on Build GRCh38
rs758394222 Chr.9: 5908723-5908723 on Build GRCh38
rs192396843 Chr.9: 5908779-5908779 on Build GRCh38
rs773606117 Chr.9: 5908966-5908966 on Build GRCh38
rs1055414069 Chr.9: 5908534-5908534 on Build GRCh38
rs904949800 Chr.9: 5908949-5908949 on Build GRCh38
rs908495237 Chr.9: 5908831-5908831 on Build GRCh38
rs149543135 Chr.9: 5908786-5908786 on Build GRCh38
rs375775687 Chr.9: 5908598-5908598 on Build GRCh38
rs753392792 Chr.9: 5908716-5908716 on Build GRCh38
rs981065977 Chr.9: 5908823-5908823 on Build GRCh38
rs899275069 Chr.9: 5908608-5908608 on Build GRCh38
rs930716366 Chr.9: 5908870-5908870 on Build GRCh38
rs975547879 Chr.9: 5908806-5908806 on Build GRCh38
rs921337259 Chr.9: 5908543-5908543 on Build GRCh38
rs775817500 Chr.9: 5908741-5908741 on Build GRCh38
rs778225719 Chr.9: 5908719-5908719 on Build GRCh38
rs1013409878 Chr.9: 5908687-5908687 on Build GRCh38
rs1033170792 Chr.9: 5908783-5908783 on Build GRCh38
rs1026670926 Chr.9: 5908985-5908985 on Build GRCh38
rs763310379 Chr.9: 5908923-5908923 on Build GRCh38
rs1052808374 Chr.9: 5908891-5908891 on Build GRCh38
rs138188973 Chr.9: 5908740-5908740 on Build GRCh38
rs991207923 Chr.9: 5909002-5909002 on Build GRCh38
rs768795850 Chr.9: 5908759-5908759 on Build GRCh38
rs939940361 Chr.9: 5908835-5908835 on Build GRCh38
rs748116895 Chr.9: 5908722-5908722 on Build GRCh38
rs374261380 Chr.9: 5908717-5908717 on Build GRCh38
rs756524213 Chr.9: 5908705-5908705 on Build GRCh38
rs531736398 Chr.9: 5908799-5908799 on Build GRCh38
rs758145281 Chr.9: 5908545-5908545 on Build GRCh38
rs770330206 Chr.9: 5908861-5908861 on Build GRCh38

View Full Product Details