Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 5909736-5910283 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KIAA2026
Entrez Gene ID
158358
Gene Name
KIAA2026
Gene Aliases
-
UniGene
Hs.535060
Gene Chromosome Location
Chr.9: 5860254-6008479 on Build GRCh38
Transcripts
XM_011517759.2 XM_011517760.2 XM_017014335.1 NM_005511.1

Gene Symbol
MLANA
Entrez Gene ID
2315
Gene Name
melan-A
Gene Aliases
MART-1, MART1
UniGene
Hs.154069
Gene Chromosome Location
Chr.9: 5890909-5909822 on Build GRCh38
Transcripts
XM_011517759.2 XM_011517760.2 XM_017014335.1 NM_005511.1

SNP Detail

SNP ID Chromosome Location
rs1005282874 Chr.9: 5910179-5910179 on Build GRCh38
rs935647166 Chr.9: 5910000-5910000 on Build GRCh38
rs1015549465 Chr.9: 5910186-5910186 on Build GRCh38
rs146855925 Chr.9: 5910018-5910018 on Build GRCh38
rs371674820 Chr.9: 5909848-5909848 on Build GRCh38
rs970095766 Chr.9: 5910202-5910202 on Build GRCh38
rs560926462 Chr.9: 5910035-5910035 on Build GRCh38
rs1001594257 Chr.9: 5910136-5910136 on Build GRCh38
rs755035837 Chr.9: 5909835-5909835 on Build GRCh38
rs878927409 Chr.9: 5910178-5910178 on Build GRCh38
rs960336752 Chr.9: 5910235-5910235 on Build GRCh38
rs932368406 Chr.9: 5909946-5909946 on Build GRCh38
rs976549386 Chr.9: 5909868-5909868 on Build GRCh38
rs578001632 Chr.9: 5909960-5909960 on Build GRCh38
rs200065321 Chr.9: 5909781-5909781 on Build GRCh38
rs140114971 Chr.9: 5909874-5909874 on Build GRCh38
rs961267203 Chr.9: 5909856-5909856 on Build GRCh38
rs1019577939 Chr.9: 5909854-5909854 on Build GRCh38
rs1031240201 Chr.9: 5909869-5909869 on Build GRCh38
rs980068692 Chr.9: 5909909-5909909 on Build GRCh38
rs928595072 Chr.9: 5910218-5910218 on Build GRCh38
rs1021702664 Chr.9: 5910196-5910196 on Build GRCh38
rs952753617 Chr.9: 5909807-5909807 on Build GRCh38
rs117399323 Chr.9: 5910229-5910229 on Build GRCh38
rs879003240 Chr.9: 5909767-5909767 on Build GRCh38
rs1030836019 Chr.9: 5910099-5910099 on Build GRCh38
rs1027625599 Chr.9: 5909791-5909791 on Build GRCh38
rs1029421971 Chr.9: 5910250-5910250 on Build GRCh38
rs766624580 Chr.9: 5909797-5909797 on Build GRCh38
rs1048141300 Chr.9: 5910010-5910010 on Build GRCh38
rs770690348 Chr.9: 5909851-5909851 on Build GRCh38
rs573817975 Chr.9: 5909881-5909881 on Build GRCh38
rs889418649 Chr.9: 5909819-5909819 on Build GRCh38
rs143813071 Chr.9: 5909963-5909963 on Build GRCh38
rs561441159 Chr.9: 5910128-5910128 on Build GRCh38
rs753799741 Chr.9: 5909833-5909833 on Build GRCh38
rs189961084 Chr.9: 5909938-5909938 on Build GRCh38
rs931055476 Chr.9: 5909994-5909994 on Build GRCh38
rs566287575 Chr.9: 5909956-5909956 on Build GRCh38
rs984034245 Chr.9: 5909808-5909808 on Build GRCh38
rs999821631 Chr.9: 5910052-5910052 on Build GRCh38
rs948755979 Chr.9: 5909899-5909899 on Build GRCh38
rs976560732 Chr.9: 5910247-5910247 on Build GRCh38
rs531093909 Chr.9: 5910036-5910036 on Build GRCh38
rs764688174 Chr.9: 5910011-5910011 on Build GRCh38
rs965672554 Chr.9: 5909864-5909864 on Build GRCh38
rs372852671 Chr.9: 5910217-5910217 on Build GRCh38
rs956162665 Chr.9: 5909872-5909872 on Build GRCh38
rs1001540471 Chr.9: 5909785-5909785 on Build GRCh38
rs186246511 Chr.9: 5910146-5910146 on Build GRCh38
rs1006600754 Chr.9: 5910020-5910020 on Build GRCh38
rs376744127 Chr.9: 5910035-5910036 on Build GRCh38
rs180806679 Chr.9: 5910085-5910085 on Build GRCh38
rs1026672225 Chr.9: 5909787-5909787 on Build GRCh38
rs775451923 Chr.9: 5910148-5910148 on Build GRCh38
rs950236697 Chr.9: 5910068-5910068 on Build GRCh38
rs887810052 Chr.9: 5910163-5910163 on Build GRCh38
rs1037213024 Chr.9: 5909962-5909962 on Build GRCh38
rs762780061 Chr.9: 5910162-5910162 on Build GRCh38

View Full Product Details