Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 88571041-88571569 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NXNL2
Entrez Gene ID
158046
Gene Name
nucleoredoxin-like 2
Gene Aliases
C9orf121, RDCVF2
UniGene
Hs.734507
Gene Chromosome Location
Chr.9: 88535088-88584510 on Build GRCh38
Transcripts
NM_145283.2 XM_005251727.3 XM_011518276.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7213685 c.395C>A p.A132D Chr.9: 88571179-88571179 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs562072495 Chr.9: 88571222-88571222 on Build GRCh38
rs1027878240 Chr.9: 88571132-88571132 on Build GRCh38
rs756383056 Chr.9: 88571097-88571097 on Build GRCh38
rs998439375 Chr.9: 88571220-88571220 on Build GRCh38
rs934126451 Chr.9: 88571460-88571460 on Build GRCh38
rs397718045 Chr.9: 88571083-88571084 on Build GRCh38
rs962021991 Chr.9: 88571113-88571113 on Build GRCh38
rs755555777 Chr.9: 88571068-88571071 on Build GRCh38
rs951377388 Chr.9: 88571350-88571350 on Build GRCh38
rs777422642 Chr.9: 88571146-88571146 on Build GRCh38
rs573565515 Chr.9: 88571148-88571148 on Build GRCh38
rs1014971443 Chr.9: 88571348-88571348 on Build GRCh38
rs752589691 Chr.9: 88571089-88571089 on Build GRCh38
rs541822686 Chr.9: 88571193-88571193 on Build GRCh38
rs1028306101 Chr.9: 88571408-88571408 on Build GRCh38
rs927023583 Chr.9: 88571183-88571183 on Build GRCh38
rs750650200 Chr.9: 88571088-88571088 on Build GRCh38
rs192591693 Chr.9: 88571096-88571096 on Build GRCh38
rs78289831 Chr.9: 88571475-88571475 on Build GRCh38
rs185032238 Chr.9: 88571324-88571324 on Build GRCh38
rs866183514 Chr.9: 88571236-88571236 on Build GRCh38
rs921466686 Chr.9: 88571402-88571402 on Build GRCh38
rs980988288 Chr.9: 88571179-88571179 on Build GRCh38
rs372857071 Chr.9: 88571330-88571330 on Build GRCh38
rs187405393 Chr.9: 88571068-88571068 on Build GRCh38
rs945705704 Chr.9: 88571214-88571214 on Build GRCh38
rs529566799 Chr.9: 88571542-88571542 on Build GRCh38
rs34373470 Chr.9: 88571465-88571466 on Build GRCh38
rs983157933 Chr.9: 88571375-88571375 on Build GRCh38
rs1023246799 Chr.9: 88571289-88571289 on Build GRCh38
rs1054559393 Chr.9: 88571513-88571513 on Build GRCh38
rs934342422 Chr.9: 88571499-88571499 on Build GRCh38
rs969697454 Chr.9: 88571437-88571437 on Build GRCh38
rs868344089 Chr.9: 88571340-88571340 on Build GRCh38
rs527683762 Chr.9: 88571126-88571126 on Build GRCh38
rs757810462 Chr.9: 88571516-88571516 on Build GRCh38
rs752042040 Chr.9: 88571427-88571427 on Build GRCh38
rs766023613 Chr.9: 88571212-88571212 on Build GRCh38
rs963311549 Chr.9: 88571393-88571393 on Build GRCh38
rs200212505 Chr.9: 88571088-88571088 on Build GRCh38
rs564550564 Chr.9: 88571381-88571381 on Build GRCh38
rs796119169 Chr.9: 88571072-88571073 on Build GRCh38
rs189414673 Chr.9: 88571477-88571477 on Build GRCh38
rs753814571 Chr.9: 88571072-88571072 on Build GRCh38
rs960870834 Chr.9: 88571541-88571542 on Build GRCh38
rs893014520 Chr.9: 88571261-88571261 on Build GRCh38
rs938483899 Chr.9: 88571196-88571196 on Build GRCh38
rs568668266 Chr.9: 88571071-88571072 on Build GRCh38
rs60872685 Chr.9: 88571072-88571073 on Build GRCh38
rs533552831 Chr.9: 88571406-88571406 on Build GRCh38
rs1050794056 Chr.9: 88571202-88571202 on Build GRCh38
rs975688981 Chr.9: 88571398-88571398 on Build GRCh38
rs556823309 Chr.9: 88571127-88571127 on Build GRCh38
rs1016920189 Chr.9: 88571386-88571386 on Build GRCh38
rs755322975 Chr.9: 88571328-88571328 on Build GRCh38
rs924341401 Chr.9: 88571478-88571478 on Build GRCh38
rs1050308419 Chr.9: 88571322-88571322 on Build GRCh38
rs55737525 Chr.9: 88571276-88571276 on Build GRCh38
rs907035479 Chr.9: 88571321-88571321 on Build GRCh38
rs969940100 Chr.9: 88571167-88571167 on Build GRCh38

View Full Product Details